Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48852728-48853286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALCOCO2
Entrez Gene ID
10241
Gene Name
calcium binding and coiled-coil domain 2
Gene Aliases
NDP52
UniGene
Hs.514920
Gene Chromosome Location
Chr.17: 48830988-48865245 on Build GRCh38
Transcripts
NM_001261390.1 NM_001261391.1 NM_001261393.1 NM_001261395.1 NM_005831.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5414169 c.826A>G p.R276G Chr.17: 48852926-48852926 on Build GRCh38
4668886 c.846C>T p.L282L Chr.17: 48852946-48852946 on Build GRCh38
4767074 c.862C>A p.Q288K Chr.17: 48852962-48852962 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs530384933 Chr.17: 48853145-48853145 on Build GRCh38
rs903157336 Chr.17: 48852772-48852772 on Build GRCh38
rs748520003 Chr.17: 48853221-48853221 on Build GRCh38
rs770702143 Chr.17: 48853031-48853031 on Build GRCh38
rs532650385 Chr.17: 48852864-48852864 on Build GRCh38
rs1045054903 Chr.17: 48853160-48853160 on Build GRCh38
rs1021404066 Chr.17: 48852967-48852967 on Build GRCh38
rs752582123 Chr.17: 48852909-48852909 on Build GRCh38
rs142857064 Chr.17: 48853192-48853201 on Build GRCh38
rs139933630 Chr.17: 48852943-48852943 on Build GRCh38
rs76906433 Chr.17: 48852889-48852889 on Build GRCh38
rs200799401 Chr.17: 48853035-48853035 on Build GRCh38
rs909361727 Chr.17: 48853138-48853138 on Build GRCh38
rs184769672 Chr.17: 48853049-48853049 on Build GRCh38
rs111443380 Chr.17: 48853203-48853203 on Build GRCh38
rs1027520411 Chr.17: 48853115-48853115 on Build GRCh38
rs758698987 Chr.17: 48853000-48853000 on Build GRCh38
rs918321387 Chr.17: 48852785-48852785 on Build GRCh38
rs1053631419 Chr.17: 48852897-48852897 on Build GRCh38
rs765125596 Chr.17: 48852907-48852907 on Build GRCh38
rs748615784 Chr.17: 48852959-48852959 on Build GRCh38
rs185262043 Chr.17: 48853224-48853224 on Build GRCh38
rs917350690 Chr.17: 48853154-48853154 on Build GRCh38
rs374653369 Chr.17: 48853011-48853011 on Build GRCh38
rs918341596 Chr.17: 48853088-48853088 on Build GRCh38
rs369797247 Chr.17: 48852803-48852803 on Build GRCh38
rs762641191 Chr.17: 48852924-48852924 on Build GRCh38
rs997012627 Chr.17: 48852826-48852826 on Build GRCh38
rs1044426527 Chr.17: 48852871-48852871 on Build GRCh38
rs928879513 Chr.17: 48853165-48853165 on Build GRCh38
rs199665295 Chr.17: 48852914-48852914 on Build GRCh38
rs566089014 Chr.17: 48853077-48853077 on Build GRCh38
rs112308313 Chr.17: 48852981-48852981 on Build GRCh38
rs372125448 Chr.17: 48853039-48853039 on Build GRCh38
rs900011858 Chr.17: 48852806-48852806 on Build GRCh38
rs201961359 Chr.17: 48852937-48852937 on Build GRCh38
rs535540895 Chr.17: 48852775-48852775 on Build GRCh38
rs576695844 Chr.17: 48852906-48852906 on Build GRCh38
rs776097051 Chr.17: 48852808-48852808 on Build GRCh38
rs1006742035 Chr.17: 48853032-48853032 on Build GRCh38
rs747256547 Chr.17: 48853029-48853029 on Build GRCh38
rs575698593 Chr.17: 48852842-48852842 on Build GRCh38
rs984998244 Chr.17: 48853093-48853093 on Build GRCh38
rs907185379 Chr.17: 48852885-48852885 on Build GRCh38
rs952433609 Chr.17: 48853089-48853089 on Build GRCh38
rs368214911 Chr.17: 48852879-48852879 on Build GRCh38
rs189402292 Chr.17: 48853143-48853143 on Build GRCh38
rs867493896 Chr.17: 48853014-48853014 on Build GRCh38
rs181679791 Chr.17: 48853193-48853193 on Build GRCh38
rs958571252 Chr.17: 48853117-48853117 on Build GRCh38
rs73989219 Chr.17: 48853167-48853167 on Build GRCh38
rs971519870 Chr.17: 48853155-48853156 on Build GRCh38
rs921495757 Chr.17: 48853202-48853202 on Build GRCh38
rs749923376 Chr.17: 48852947-48852947 on Build GRCh38
rs745438847 Chr.17: 48853191-48853192 on Build GRCh38
rs772515245 Chr.17: 48853241-48853241 on Build GRCh38
rs780882076 Chr.17: 48852946-48852946 on Build GRCh38
rs537841605 Chr.17: 48852856-48852856 on Build GRCh38
rs771013935 Chr.17: 48852880-48852880 on Build GRCh38
rs764157879 Chr.17: 48852942-48852942 on Build GRCh38
rs779275146 Chr.17: 48852951-48852951 on Build GRCh38

View Full Product Details