Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 69153795-69154347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA10
Entrez Gene ID
10349
Gene Name
ATP binding cassette subfamily A member 10
Gene Aliases
EST698739
UniGene
Hs.25377
Gene Chromosome Location
Chr.17: 69148007-69244815 on Build GRCh38
Transcripts
NM_080282.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7298869 c.3803G>A p.S1268N Chr.17: 69153993-69153993 on Build GRCh38
1735140 c.3879G>T p.W1293C Chr.17: 69153917-69153917 on Build GRCh38
7321576 c.3853T>C p.Y1285H Chr.17: 69153943-69153943 on Build GRCh38
148329 c.3819G>A p.R1273R Chr.17: 69153977-69153977 on Build GRCh38
148328 c.3964C>T p.R1322* Chr.17: 69153832-69153832 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773893541 Chr.17: 69153906-69153906 on Build GRCh38
rs148189434 Chr.17: 69153824-69153824 on Build GRCh38
rs369400674 Chr.17: 69154173-69154173 on Build GRCh38
rs766333221 Chr.17: 69154195-69154195 on Build GRCh38
rs771427156 Chr.17: 69154011-69154011 on Build GRCh38
rs904434814 Chr.17: 69154142-69154142 on Build GRCh38
rs16973656 Chr.17: 69153977-69153977 on Build GRCh38
rs765488334 Chr.17: 69154045-69154045 on Build GRCh38
rs940079536 Chr.17: 69154079-69154079 on Build GRCh38
rs773503109 Chr.17: 69154274-69154274 on Build GRCh38
rs779093748 Chr.17: 69153833-69153833 on Build GRCh38
rs757098853 Chr.17: 69154309-69154309 on Build GRCh38
rs764900379 Chr.17: 69154310-69154310 on Build GRCh38
rs750541228 Chr.17: 69154050-69154050 on Build GRCh38
rs762268158 Chr.17: 69154258-69154258 on Build GRCh38
rs951613427 Chr.17: 69154123-69154123 on Build GRCh38
rs750128232 Chr.17: 69154317-69154317 on Build GRCh38
rs764024092 Chr.17: 69154307-69154307 on Build GRCh38
rs772876949 Chr.17: 69153877-69153877 on Build GRCh38
rs556543958 Chr.17: 69153937-69153937 on Build GRCh38
rs756471261 Chr.17: 69153823-69153823 on Build GRCh38
rs765281292 Chr.17: 69153971-69153971 on Build GRCh38
rs375144849 Chr.17: 69154246-69154246 on Build GRCh38
rs555577333 Chr.17: 69154043-69154043 on Build GRCh38
rs577453947 Chr.17: 69154121-69154121 on Build GRCh38
rs763057090 Chr.17: 69154284-69154284 on Build GRCh38
rs937091663 Chr.17: 69153876-69153876 on Build GRCh38
rs753094771 Chr.17: 69153822-69153822 on Build GRCh38
rs781192073 Chr.17: 69153978-69153978 on Build GRCh38
rs866432129 Chr.17: 69154174-69154174 on Build GRCh38
rs370574737 Chr.17: 69154183-69154183 on Build GRCh38
rs527370037 Chr.17: 69154218-69154218 on Build GRCh38
rs369143410 Chr.17: 69153910-69153910 on Build GRCh38
rs778354658 Chr.17: 69154003-69154003 on Build GRCh38
rs574002350 Chr.17: 69154083-69154083 on Build GRCh38
rs145891525 Chr.17: 69154130-69154130 on Build GRCh38
rs771642187 Chr.17: 69154236-69154236 on Build GRCh38
rs373370338 Chr.17: 69153912-69153912 on Build GRCh38
rs10491178 Chr.17: 69153832-69153832 on Build GRCh38
rs796080863 Chr.17: 69153871-69153871 on Build GRCh38
rs750661377 Chr.17: 69154020-69154020 on Build GRCh38
rs776085553 Chr.17: 69154018-69154018 on Build GRCh38
rs760676147 Chr.17: 69154299-69154299 on Build GRCh38
rs747568314 Chr.17: 69154243-69154243 on Build GRCh38
rs777199696 Chr.17: 69154247-69154247 on Build GRCh38
rs1057220471 Chr.17: 69154102-69154102 on Build GRCh38
rs937019879 Chr.17: 69154141-69154144 on Build GRCh38
rs185372264 Chr.17: 69154115-69154115 on Build GRCh38
rs200172999 Chr.17: 69153980-69153980 on Build GRCh38
rs557508194 Chr.17: 69154170-69154170 on Build GRCh38
rs369404727 Chr.17: 69154220-69154220 on Build GRCh38
rs138422677 Chr.17: 69154206-69154206 on Build GRCh38
rs778643341 Chr.17: 69154224-69154224 on Build GRCh38
rs752674370 Chr.17: 69154199-69154199 on Build GRCh38
rs755937158 Chr.17: 69153986-69153986 on Build GRCh38
rs141590994 Chr.17: 69154010-69154010 on Build GRCh38
rs766532619 Chr.17: 69154288-69154288 on Build GRCh38
rs144160248 Chr.17: 69153953-69153953 on Build GRCh38
rs11657280 Chr.17: 69153840-69153840 on Build GRCh38
rs750499120 Chr.17: 69153973-69153973 on Build GRCh38
rs763104475 Chr.17: 69154059-69154059 on Build GRCh38
rs947090046 Chr.17: 69154217-69154217 on Build GRCh38
rs372025211 Chr.17: 69154225-69154225 on Build GRCh38
rs1013210829 Chr.17: 69153828-69153828 on Build GRCh38
rs766837501 Chr.17: 69154104-69154104 on Build GRCh38
rs779397801 Chr.17: 69154242-69154242 on Build GRCh38
rs770470255 Chr.17: 69153905-69153905 on Build GRCh38
rs142909799 Chr.17: 69154209-69154209 on Build GRCh38
rs754919048 Chr.17: 69153850-69153850 on Build GRCh38
rs1007287662 Chr.17: 69154068-69154068 on Build GRCh38
rs775793828 Chr.17: 69153917-69153917 on Build GRCh38
rs376929280 Chr.17: 69154166-69154166 on Build GRCh38
rs769183262 Chr.17: 69154034-69154034 on Build GRCh38
rs769993558 Chr.17: 69154268-69154268 on Build GRCh38
rs928298431 Chr.17: 69154138-69154138 on Build GRCh38
rs777515195 Chr.17: 69153990-69153990 on Build GRCh38
rs113881859 Chr.17: 69153875-69153875 on Build GRCh38
rs908500288 Chr.17: 69154254-69154254 on Build GRCh38
rs750932909 Chr.17: 69154058-69154058 on Build GRCh38
rs1000081898 Chr.17: 69154151-69154151 on Build GRCh38
rs761135569 Chr.17: 69153933-69153933 on Build GRCh38
rs532320175 Chr.17: 69153870-69153870 on Build GRCh38
rs2886232 Chr.17: 69154035-69154035 on Build GRCh38
rs558944942 Chr.17: 69154114-69154114 on Build GRCh38
rs77718465 Chr.17: 69153995-69153995 on Build GRCh38
rs887131161 Chr.17: 69154062-69154062 on Build GRCh38
rs753719056 Chr.17: 69154308-69154308 on Build GRCh38
rs748904094 Chr.17: 69153883-69153883 on Build GRCh38
rs1048415730 Chr.17: 69154040-69154040 on Build GRCh38
rs770558049 Chr.17: 69154001-69154001 on Build GRCh38
rs866283705 Chr.17: 69153938-69153938 on Build GRCh38
rs200206246 Chr.17: 69154297-69154297 on Build GRCh38
rs72852601 Chr.17: 69153831-69153831 on Build GRCh38
rs376127309 Chr.17: 69153861-69153861 on Build GRCh38
rs757521279 Chr.17: 69153954-69153954 on Build GRCh38

View Full Product Details