Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 35977247-35977835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C18orf21
Entrez Gene ID
83608
Gene Name
chromosome 18 open reading frame 21
Gene Aliases
HsT3108, PNAS-124, PNAS-131, XTP13
UniGene
Hs.37883
Gene Chromosome Location
Chr.18: 35972625-35979287 on Build GRCh38
Transcripts
NM_001201474.1 NM_001201475.1 NM_001201476.1 NM_031446.4 XM_005258364.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
473797 c.474C>A p.S158R Chr.18: 35977583-35977583 on Build GRCh38
1494033 c.301-2A>T p.? Chr.18: 35977408-35977408 on Build GRCh38
1611210 c.404T>G p.L135R Chr.18: 35977513-35977513 on Build GRCh38
3388432 c.321C>G p.C107W Chr.18: 35977430-35977430 on Build GRCh38
988179 c.434C>G p.A145G Chr.18: 35977543-35977543 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748172036 Chr.18: 35977482-35977482 on Build GRCh38
rs774827908 Chr.18: 35977628-35977628 on Build GRCh38
rs1008375326 Chr.18: 35977397-35977397 on Build GRCh38
rs755650426 Chr.18: 35977490-35977491 on Build GRCh38
rs765555167 Chr.18: 35977422-35977422 on Build GRCh38
rs940056929 Chr.18: 35977297-35977297 on Build GRCh38
rs774167045 Chr.18: 35977481-35977481 on Build GRCh38
rs766915346 Chr.18: 35977523-35977523 on Build GRCh38
rs772091862 Chr.18: 35977589-35977589 on Build GRCh38
rs770907932 Chr.18: 35977486-35977486 on Build GRCh38
rs553952215 Chr.18: 35977637-35977637 on Build GRCh38
rs768250533 Chr.18: 35977302-35977302 on Build GRCh38
rs746656059 Chr.18: 35977339-35977339 on Build GRCh38
rs745594273 Chr.18: 35977364-35977364 on Build GRCh38
rs758852843 Chr.18: 35977429-35977429 on Build GRCh38
rs768103282 Chr.18: 35977644-35977644 on Build GRCh38
rs765466245 Chr.18: 35977489-35977489 on Build GRCh38
rs961459490 Chr.18: 35977526-35977526 on Build GRCh38
rs1042552379 Chr.18: 35977631-35977631 on Build GRCh38
rs753991028 Chr.18: 35977406-35977406 on Build GRCh38
rs371277931 Chr.18: 35977568-35977568 on Build GRCh38
rs755791233 Chr.18: 35977533-35977533 on Build GRCh38
rs1011942147 Chr.18: 35977700-35977700 on Build GRCh38
rs148863953 Chr.18: 35977527-35977527 on Build GRCh38
rs773305552 Chr.18: 35977399-35977399 on Build GRCh38
rs1053579128 Chr.18: 35977680-35977680 on Build GRCh38
rs768650028 Chr.18: 35977480-35977480 on Build GRCh38
rs779613081 Chr.18: 35977533-35977535 on Build GRCh38
rs919998932 Chr.18: 35977532-35977532 on Build GRCh38
rs1035037578 Chr.18: 35977725-35977725 on Build GRCh38
rs764707427 Chr.18: 35977417-35977417 on Build GRCh38
rs982178778 Chr.18: 35977716-35977716 on Build GRCh38
rs534448413 Chr.18: 35977633-35977633 on Build GRCh38
rs1000980384 Chr.18: 35977660-35977660 on Build GRCh38
rs763309887 Chr.18: 35977490-35977490 on Build GRCh38
rs751025528 Chr.18: 35977435-35977435 on Build GRCh38
rs762181682 Chr.18: 35977635-35977635 on Build GRCh38
rs764579668 Chr.18: 35977502-35977502 on Build GRCh38
rs548757762 Chr.18: 35977586-35977586 on Build GRCh38
rs570958663 Chr.18: 35977677-35977677 on Build GRCh38
rs761190396 Chr.18: 35977430-35977430 on Build GRCh38
rs552186605 Chr.18: 35977519-35977519 on Build GRCh38
rs147823103 Chr.18: 35977479-35977479 on Build GRCh38
rs973453093 Chr.18: 35977293-35977293 on Build GRCh38
rs759731869 Chr.18: 35977487-35977487 on Build GRCh38
rs915725116 Chr.18: 35977762-35977762 on Build GRCh38
rs759817593 Chr.18: 35977412-35977412 on Build GRCh38
rs766563217 Chr.18: 35977403-35977403 on Build GRCh38
rs745976900 Chr.18: 35977592-35977592 on Build GRCh38
rs894867905 Chr.18: 35977685-35977685 on Build GRCh38
rs150834990 Chr.18: 35977571-35977571 on Build GRCh38
rs537432032 Chr.18: 35977380-35977380 on Build GRCh38
rs768739816 Chr.18: 35977381-35977381 on Build GRCh38
rs749883153 Chr.18: 35977446-35977448 on Build GRCh38
rs919249847 Chr.18: 35977298-35977298 on Build GRCh38
rs370515893 Chr.18: 35977338-35977338 on Build GRCh38
rs1025031274 Chr.18: 35977757-35977757 on Build GRCh38
rs902816091 Chr.18: 35977504-35977504 on Build GRCh38
rs908653820 Chr.18: 35977287-35977287 on Build GRCh38
rs749036138 Chr.18: 35977561-35977561 on Build GRCh38
rs1024914305 Chr.18: 35977477-35977477 on Build GRCh38
rs370831050 Chr.18: 35977378-35977378 on Build GRCh38
rs970649390 Chr.18: 35977805-35977805 on Build GRCh38
rs745485657 Chr.18: 35977447-35977447 on Build GRCh38
rs773076329 Chr.18: 35977624-35977624 on Build GRCh38
rs749888626 Chr.18: 35977652-35977652 on Build GRCh38
rs760677661 Chr.18: 35977400-35977400 on Build GRCh38
rs959433601 Chr.18: 35977732-35977732 on Build GRCh38
rs780822015 Chr.18: 35977446-35977446 on Build GRCh38
rs779949649 Chr.18: 35977466-35977466 on Build GRCh38
rs1007761068 Chr.18: 35977574-35977574 on Build GRCh38
rs769934171 Chr.18: 35977598-35977598 on Build GRCh38
rs775678141 Chr.18: 35977603-35977603 on Build GRCh38
rs991304242 Chr.18: 35977748-35977748 on Build GRCh38
rs947271526 Chr.18: 35977474-35977476 on Build GRCh38
rs2276314 Chr.18: 35977503-35977503 on Build GRCh38
rs201951096 Chr.18: 35977539-35977539 on Build GRCh38
rs769623390 Chr.18: 35977459-35977459 on Build GRCh38
rs143556172 Chr.18: 35977591-35977591 on Build GRCh38
rs200115196 Chr.18: 35977425-35977425 on Build GRCh38
rs368304216 Chr.18: 35977367-35977367 on Build GRCh38
rs756710700 Chr.18: 35977436-35977436 on Build GRCh38
rs568034136 Chr.18: 35977758-35977758 on Build GRCh38

View Full Product Details