Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7959569-7960158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELAVL1
Entrez Gene ID
1994
Gene Name
ELAV like RNA binding protein 1
Gene Aliases
ELAV1, HUR, Hua, MelG
UniGene
Hs.184492
Gene Chromosome Location
Chr.19: 7958573-8005645 on Build GRCh38
Transcripts
NM_001419.2

SNP Detail

SNP ID Chromosome Location
rs3745391 Chr.19: 7960127-7960127 on Build GRCh38
rs944503634 Chr.19: 7959727-7959727 on Build GRCh38
rs1053142581 Chr.19: 7959891-7959891 on Build GRCh38
rs184690108 Chr.19: 7959996-7959996 on Build GRCh38
rs952478331 Chr.19: 7959736-7959736 on Build GRCh38
rs765946658 Chr.19: 7960095-7960095 on Build GRCh38
rs568729970 Chr.19: 7959775-7959775 on Build GRCh38
rs900731994 Chr.19: 7959680-7959680 on Build GRCh38
rs541653144 Chr.19: 7960018-7960018 on Build GRCh38
rs913292545 Chr.19: 7959901-7959901 on Build GRCh38
rs1005306755 Chr.19: 7959740-7959740 on Build GRCh38
rs925838685 Chr.19: 7960073-7960073 on Build GRCh38
rs776283805 Chr.19: 7960126-7960126 on Build GRCh38
rs2042920 Chr.19: 7959800-7959800 on Build GRCh38
rs1040530749 Chr.19: 7959927-7959927 on Build GRCh38
rs570971403 Chr.19: 7959886-7959886 on Build GRCh38
rs775986592 Chr.19: 7959667-7959667 on Build GRCh38
rs186610101 Chr.19: 7959801-7959801 on Build GRCh38
rs985929897 Chr.19: 7959707-7959707 on Build GRCh38
rs182688677 Chr.19: 7959591-7959591 on Build GRCh38
rs552513123 Chr.19: 7959642-7959642 on Build GRCh38
rs532836657 Chr.19: 7959633-7959633 on Build GRCh38
rs925974082 Chr.19: 7959868-7959868 on Build GRCh38
rs571963611 Chr.19: 7960062-7960062 on Build GRCh38
rs538050408 Chr.19: 7959765-7959765 on Build GRCh38
rs1015318046 Chr.19: 7959746-7959746 on Build GRCh38
rs770077493 Chr.19: 7959617-7959617 on Build GRCh38
rs957025186 Chr.19: 7959989-7959989 on Build GRCh38
rs980046617 Chr.19: 7960063-7960063 on Build GRCh38
rs1049736842 Chr.19: 7959973-7959973 on Build GRCh38
rs192658356 Chr.19: 7959893-7959893 on Build GRCh38
rs1008988366 Chr.19: 7959629-7959629 on Build GRCh38
rs970419376 Chr.19: 7960056-7960056 on Build GRCh38
rs200747424 Chr.19: 7960133-7960134 on Build GRCh38
rs368346724 Chr.19: 7960011-7960011 on Build GRCh38
rs896560492 Chr.19: 7960131-7960131 on Build GRCh38
rs1005193904 Chr.19: 7959995-7959995 on Build GRCh38
rs540649524 Chr.19: 7960100-7960100 on Build GRCh38
rs944896365 Chr.19: 7959905-7959905 on Build GRCh38
rs919145717 Chr.19: 7959751-7959751 on Build GRCh38
rs996941960 Chr.19: 7959687-7959687 on Build GRCh38
rs189143756 Chr.19: 7960042-7960042 on Build GRCh38
rs900783308 Chr.19: 7959969-7959969 on Build GRCh38
rs909124969 Chr.19: 7960114-7960114 on Build GRCh38
rs147458010 Chr.19: 7959998-7959998 on Build GRCh38
rs141440324 Chr.19: 7959770-7959770 on Build GRCh38
rs556362520 Chr.19: 7959917-7959917 on Build GRCh38
rs996437886 Chr.19: 7959972-7959972 on Build GRCh38
rs17160080 Chr.19: 7959715-7959715 on Build GRCh38
rs1033081374 Chr.19: 7959987-7959987 on Build GRCh38
rs896593044 Chr.19: 7959900-7959900 on Build GRCh38
rs73008886 Chr.19: 7959638-7959638 on Build GRCh38
rs961212300 Chr.19: 7959769-7959769 on Build GRCh38
rs965779973 Chr.19: 7960013-7960013 on Build GRCh38
rs917176796 Chr.19: 7959788-7959788 on Build GRCh38
rs116715205 Chr.19: 7959605-7959605 on Build GRCh38
rs775211736 Chr.19: 7959741-7959741 on Build GRCh38
rs988751715 Chr.19: 7960097-7960097 on Build GRCh38
rs1014532540 Chr.19: 7960035-7960035 on Build GRCh38
rs888118600 Chr.19: 7959974-7959974 on Build GRCh38
rs1001162434 Chr.19: 7959985-7959985 on Build GRCh38

View Full Product Details