Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45052026-45052307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLASRP
Entrez Gene ID
11129
Gene Name
CLK4 associating serine/arginine rich protein
Gene Aliases
CLASP, SFRS16, SWAP2
UniGene
Hs.466917
Gene Chromosome Location
Chr.19: 45039040-45070956 on Build GRCh38
Transcripts
NM_001278439.1 NM_007056.2 NR_103529.1 XM_011526396.2 XM_011526397.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
218586 c.131A>G p.H44R Chr.19: 45052102-45052102 on Build GRCh38
6629752 c.178G>A p.A60T Chr.19: 45052149-45052149 on Build GRCh38
4926019 c.135C>T p.G45G Chr.19: 45052106-45052106 on Build GRCh38
5571641 c.168C>T p.V56V Chr.19: 45052139-45052139 on Build GRCh38
3103726 c.137G>A p.R46Q Chr.19: 45052108-45052108 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201191094 Chr.19: 45052168-45052168 on Build GRCh38
rs772078763 Chr.19: 45052192-45052192 on Build GRCh38
rs1022808333 Chr.19: 45052220-45052220 on Build GRCh38
rs966356331 Chr.19: 45052193-45052193 on Build GRCh38
rs758270429 Chr.19: 45052072-45052072 on Build GRCh38
rs530832677 Chr.19: 45052279-45052279 on Build GRCh38
rs375029997 Chr.19: 45052276-45052276 on Build GRCh38
rs754500120 Chr.19: 45052121-45052121 on Build GRCh38
rs760807132 Chr.19: 45052215-45052215 on Build GRCh38
rs369845427 Chr.19: 45052280-45052280 on Build GRCh38
rs146072562 Chr.19: 45052094-45052094 on Build GRCh38
rs375710334 Chr.19: 45052187-45052187 on Build GRCh38
rs892309900 Chr.19: 45052055-45052055 on Build GRCh38
rs909660080 Chr.19: 45052288-45052288 on Build GRCh38
rs749360245 Chr.19: 45052217-45052217 on Build GRCh38
rs1043849246 Chr.19: 45052100-45052100 on Build GRCh38
rs138855474 Chr.19: 45052139-45052139 on Build GRCh38
rs573355406 Chr.19: 45052062-45052062 on Build GRCh38
rs377525602 Chr.19: 45052219-45052219 on Build GRCh38
rs751263482 Chr.19: 45052097-45052097 on Build GRCh38
rs771378485 Chr.19: 45052158-45052158 on Build GRCh38
rs541726457 Chr.19: 45052209-45052209 on Build GRCh38
rs142091108 Chr.19: 45052140-45052140 on Build GRCh38
rs750179843 Chr.19: 45052064-45052064 on Build GRCh38
rs903835656 Chr.19: 45052108-45052108 on Build GRCh38
rs1026406849 Chr.19: 45052272-45052272 on Build GRCh38
rs778414624 Chr.19: 45052143-45052143 on Build GRCh38
rs774408352 Chr.19: 45052163-45052163 on Build GRCh38
rs749606966 Chr.19: 45052148-45052148 on Build GRCh38
rs988203223 Chr.19: 45052205-45052205 on Build GRCh38
rs980921894 Chr.19: 45052278-45052278 on Build GRCh38

View Full Product Details