Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 51280675-51281274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM124A
Entrez Gene ID
220108
Gene Name
family with sequence similarity 124 member A
Gene Aliases
-
UniGene
Hs.71913
Gene Chromosome Location
Chr.13: 51222328-51284241 on Build GRCh38
Transcripts
NM_001242312.1 NM_145019.3 XM_011534978.2 XM_017020419.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
266494 c.1211C>T p.T404M Chr.13: 51280718-51280718 on Build GRCh38
4911052 c.1410C>T p.P470P Chr.13: 51280917-51280917 on Build GRCh38
6139071 c.1477C>T p.H493Y Chr.13: 51280984-51280984 on Build GRCh38
4048007 c.1332G>T p.G444G Chr.13: 51280839-51280839 on Build GRCh38
253143 c.1252_1253insT p.S419fs*23 Chr.13: 51280759-51280760 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201596638 Chr.13: 51280719-51280719 on Build GRCh38
rs201482688 Chr.13: 51280871-51280871 on Build GRCh38
rs769117623 Chr.13: 51281210-51281210 on Build GRCh38
rs750523295 Chr.13: 51280965-51280965 on Build GRCh38
rs199526838 Chr.13: 51281232-51281232 on Build GRCh38
rs143315968 Chr.13: 51281224-51281224 on Build GRCh38
rs770502031 Chr.13: 51281014-51281014 on Build GRCh38
rs369888482 Chr.13: 51280934-51280934 on Build GRCh38
rs748080141 Chr.13: 51280955-51280955 on Build GRCh38
rs927790247 Chr.13: 51281027-51281027 on Build GRCh38
rs183181635 Chr.13: 51281168-51281168 on Build GRCh38
rs74087446 Chr.13: 51280978-51280978 on Build GRCh38
rs772701284 Chr.13: 51280955-51280955 on Build GRCh38
rs778049276 Chr.13: 51280756-51280756 on Build GRCh38
rs780939346 Chr.13: 51281197-51281197 on Build GRCh38
rs751391739 Chr.13: 51281091-51281091 on Build GRCh38
rs775249581 Chr.13: 51281131-51281131 on Build GRCh38
rs769453246 Chr.13: 51281249-51281250 on Build GRCh38
rs780052468 Chr.13: 51281213-51281213 on Build GRCh38
rs770263485 Chr.13: 51280884-51280884 on Build GRCh38
rs141853570 Chr.13: 51280824-51280824 on Build GRCh38
rs201425404 Chr.13: 51280967-51280967 on Build GRCh38
rs73492203 Chr.13: 51280907-51280907 on Build GRCh38
rs751372590 Chr.13: 51280724-51280725 on Build GRCh38
rs772751758 Chr.13: 51280943-51280943 on Build GRCh38
rs570496783 Chr.13: 51280883-51280883 on Build GRCh38
rs766435081 Chr.13: 51281084-51281084 on Build GRCh38
rs774013147 Chr.13: 51280825-51280825 on Build GRCh38
rs200544300 Chr.13: 51281223-51281223 on Build GRCh38
rs764691984 Chr.13: 51280946-51280946 on Build GRCh38
rs781035748 Chr.13: 51280796-51280801 on Build GRCh38
rs750386210 Chr.13: 51280841-51280841 on Build GRCh38
rs755357233 Chr.13: 51280972-51280972 on Build GRCh38
rs148792088 Chr.13: 51281135-51281135 on Build GRCh38
rs762589034 Chr.13: 51280717-51280717 on Build GRCh38
rs554761463 Chr.13: 51281124-51281124 on Build GRCh38
rs199695346 Chr.13: 51281059-51281059 on Build GRCh38
rs541454271 Chr.13: 51281121-51281121 on Build GRCh38
rs372660224 Chr.13: 51280732-51280732 on Build GRCh38
rs748572384 Chr.13: 51281098-51281098 on Build GRCh38
rs142444243 Chr.13: 51281140-51281140 on Build GRCh38
rs781562278 Chr.13: 51280810-51280810 on Build GRCh38
rs745846241 Chr.13: 51281123-51281123 on Build GRCh38
rs774102178 Chr.13: 51281226-51281226 on Build GRCh38
rs978087114 Chr.13: 51280918-51280918 on Build GRCh38
rs892922127 Chr.13: 51281114-51281114 on Build GRCh38
rs2297969 Chr.13: 51280956-51280956 on Build GRCh38
rs185038718 Chr.13: 51280950-51280950 on Build GRCh38
rs887008248 Chr.13: 51281062-51281062 on Build GRCh38
rs555897503 Chr.13: 51280951-51280951 on Build GRCh38
rs368002066 Chr.13: 51281248-51281248 on Build GRCh38
rs770974094 Chr.13: 51280805-51280805 on Build GRCh38
rs746552464 Chr.13: 51280996-51280996 on Build GRCh38
rs767321784 Chr.13: 51281233-51281233 on Build GRCh38
rs779986291 Chr.13: 51280735-51280735 on Build GRCh38
rs755480597 Chr.13: 51281090-51281090 on Build GRCh38
rs773890927 Chr.13: 51280924-51280924 on Build GRCh38
rs1039937198 Chr.13: 51281214-51281214 on Build GRCh38
rs779106524 Chr.13: 51280697-51280698 on Build GRCh38
rs369530645 Chr.13: 51281133-51281133 on Build GRCh38
rs756945162 Chr.13: 51281183-51281183 on Build GRCh38
rs200873096 Chr.13: 51280744-51280744 on Build GRCh38
rs767810486 Chr.13: 51281073-51281073 on Build GRCh38
rs754410688 Chr.13: 51281085-51281085 on Build GRCh38
rs761334979 Chr.13: 51280726-51280726 on Build GRCh38
rs772695251 Chr.13: 51280992-51280992 on Build GRCh38
rs139792774 Chr.13: 51281161-51281161 on Build GRCh38
rs563845428 Chr.13: 51281144-51281144 on Build GRCh38
rs771549646 Chr.13: 51280935-51280935 on Build GRCh38
rs143623611 Chr.13: 51280718-51280718 on Build GRCh38
rs912534192 Chr.13: 51281008-51281008 on Build GRCh38
rs937432250 Chr.13: 51280797-51280797 on Build GRCh38
rs138998748 Chr.13: 51280873-51280873 on Build GRCh38
rs1010388341 Chr.13: 51281118-51281118 on Build GRCh38
rs750310277 Chr.13: 51280731-51280731 on Build GRCh38
rs775329106 Chr.13: 51281040-51281040 on Build GRCh38
rs919738940 Chr.13: 51281158-51281158 on Build GRCh38
rs751568692 Chr.13: 51281187-51281187 on Build GRCh38
rs767542969 Chr.13: 51280961-51280961 on Build GRCh38
rs146428564 Chr.13: 51281150-51281150 on Build GRCh38
rs757466454 Chr.13: 51280957-51280957 on Build GRCh38
rs958609229 Chr.13: 51281142-51281142 on Build GRCh38
rs767389460 Chr.13: 51280729-51280729 on Build GRCh38
rs146405973 Chr.13: 51280870-51280870 on Build GRCh38
rs747083506 Chr.13: 51280796-51280796 on Build GRCh38
rs776764205 Chr.13: 51281034-51281034 on Build GRCh38
rs1047502574 Chr.13: 51280839-51280839 on Build GRCh38
rs373223015 Chr.13: 51280854-51280854 on Build GRCh38
rs749738902 Chr.13: 51280968-51280968 on Build GRCh38
rs758663391 Chr.13: 51280753-51280753 on Build GRCh38
rs377315859 Chr.13: 51280891-51280891 on Build GRCh38
rs934947550 Chr.13: 51280750-51280750 on Build GRCh38
rs770628949 Chr.13: 51281116-51281116 on Build GRCh38
rs541142132 Chr.13: 51280716-51280716 on Build GRCh38
rs766321580 Chr.13: 51280855-51280855 on Build GRCh38
rs764976880 Chr.13: 51281039-51281039 on Build GRCh38
rs773483406 Chr.13: 51281138-51281138 on Build GRCh38
rs754782799 Chr.13: 51280792-51280792 on Build GRCh38
rs143119311 Chr.13: 51280881-51280881 on Build GRCh38
rs927098528 Chr.13: 51280738-51280738 on Build GRCh38
rs753666905 Chr.13: 51280857-51280857 on Build GRCh38
rs1002798748 Chr.13: 51281137-51281137 on Build GRCh38
rs758653958 Chr.13: 51281099-51281099 on Build GRCh38
rs770210414 Chr.13: 51280822-51280822 on Build GRCh38
rs1053544243 Chr.13: 51280789-51280789 on Build GRCh38
rs201760116 Chr.13: 51280840-51280840 on Build GRCh38
rs746644484 Chr.13: 51281105-51281105 on Build GRCh38
rs148055539 Chr.13: 51280815-51280815 on Build GRCh38
rs756201060 Chr.13: 51281081-51281081 on Build GRCh38
rs939164230 Chr.13: 51281058-51281058 on Build GRCh38
rs754774123 Chr.13: 51280864-51280864 on Build GRCh38
rs775796682 Chr.13: 51280823-51280823 on Build GRCh38
rs745415065 Chr.13: 51281205-51281205 on Build GRCh38
rs776797685 Chr.13: 51281243-51281243 on Build GRCh38
rs189898463 Chr.13: 51281160-51281160 on Build GRCh38
rs370895532 Chr.13: 51281166-51281166 on Build GRCh38
rs771738151 Chr.13: 51280984-51280984 on Build GRCh38
rs749683400 Chr.13: 51280903-51280903 on Build GRCh38
rs1021965747 Chr.13: 51281195-51281195 on Build GRCh38
rs955808362 Chr.13: 51280697-51280697 on Build GRCh38
rs202111589 Chr.13: 51280945-51280945 on Build GRCh38
rs200189512 Chr.13: 51281035-51281035 on Build GRCh38

View Full Product Details