Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 35013862-35014166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGXT2
Entrez Gene ID
64902
Gene Name
alanine--glyoxylate aminotransferase 2
Gene Aliases
AGT2, DAIBAT
UniGene
Hs.34494
Gene Chromosome Location
Chr.5: 34998101-35048188 on Build GRCh38
Transcripts
NM_001306173.1 NM_031900.3 XM_005248337.2 XM_005248338.2 XM_017009748.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
398279 c.1063G>T p.G355C Chr.5: 35014020-35014020 on Build GRCh38
6210411 c.1046C>T p.A349V Chr.5: 35014037-35014037 on Build GRCh38
7412054 c.1038C>G p.V346V Chr.5: 35014045-35014045 on Build GRCh38
3919941 c.1067T>G p.F356C Chr.5: 35014016-35014016 on Build GRCh38
1182253 c.1024C>G p.L342V Chr.5: 35014059-35014059 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558762456 Chr.5: 35013920-35013920 on Build GRCh38
rs759743337 Chr.5: 35014143-35014143 on Build GRCh38
rs889486698 Chr.5: 35013973-35013973 on Build GRCh38
rs751302445 Chr.5: 35014084-35014084 on Build GRCh38
rs778717425 Chr.5: 35014015-35014015 on Build GRCh38
rs1035002299 Chr.5: 35014113-35014113 on Build GRCh38
rs535008171 Chr.5: 35014058-35014058 on Build GRCh38
rs750751459 Chr.5: 35013989-35013989 on Build GRCh38
rs770796340 Chr.5: 35014014-35014014 on Build GRCh38
rs766572860 Chr.5: 35014002-35014002 on Build GRCh38
rs771306167 Chr.5: 35014132-35014132 on Build GRCh38
rs566211283 Chr.5: 35013948-35013948 on Build GRCh38
rs777778215 Chr.5: 35014012-35014012 on Build GRCh38
rs139842754 Chr.5: 35014000-35014000 on Build GRCh38
rs755015773 Chr.5: 35014004-35014004 on Build GRCh38
rs763224422 Chr.5: 35013993-35013993 on Build GRCh38
rs771508619 Chr.5: 35014025-35014025 on Build GRCh38
rs534977787 Chr.5: 35014050-35014050 on Build GRCh38
rs567919307 Chr.5: 35013947-35013947 on Build GRCh38
rs147570589 Chr.5: 35014063-35014063 on Build GRCh38
rs77088572 Chr.5: 35013971-35013971 on Build GRCh38
rs116605914 Chr.5: 35014066-35014066 on Build GRCh38
rs141235149 Chr.5: 35014117-35014117 on Build GRCh38
rs774667915 Chr.5: 35014140-35014140 on Build GRCh38
rs749755111 Chr.5: 35014126-35014126 on Build GRCh38
rs138173903 Chr.5: 35013970-35013970 on Build GRCh38
rs370116434 Chr.5: 35014043-35014043 on Build GRCh38
rs548488819 Chr.5: 35013941-35013941 on Build GRCh38
rs558868891 Chr.5: 35013980-35013980 on Build GRCh38
rs34222067 Chr.5: 35014114-35014114 on Build GRCh38
rs143062537 Chr.5: 35014110-35014110 on Build GRCh38
rs778805554 Chr.5: 35014103-35014103 on Build GRCh38
rs745977837 Chr.5: 35014094-35014094 on Build GRCh38
rs767535041 Chr.5: 35014071-35014071 on Build GRCh38
rs373358835 Chr.5: 35014091-35014091 on Build GRCh38
rs769505977 Chr.5: 35013943-35013943 on Build GRCh38
rs971966932 Chr.5: 35014056-35014056 on Build GRCh38
rs960296289 Chr.5: 35013915-35013915 on Build GRCh38
rs373380683 Chr.5: 35014039-35014039 on Build GRCh38
rs183850114 Chr.5: 35014080-35014080 on Build GRCh38
rs913301683 Chr.5: 35013930-35013930 on Build GRCh38
rs768095873 Chr.5: 35014121-35014121 on Build GRCh38
rs762356643 Chr.5: 35014065-35014065 on Build GRCh38
rs1008297718 Chr.5: 35013975-35013975 on Build GRCh38
rs376208578 Chr.5: 35014010-35014010 on Build GRCh38
rs776089008 Chr.5: 35014008-35014008 on Build GRCh38
rs775040296 Chr.5: 35014030-35014030 on Build GRCh38
rs186032843 Chr.5: 35013926-35013926 on Build GRCh38
rs749082205 Chr.5: 35014013-35014013 on Build GRCh38
rs759753053 Chr.5: 35014070-35014070 on Build GRCh38
rs369923391 Chr.5: 35014024-35014024 on Build GRCh38
rs755036633 Chr.5: 35014124-35014125 on Build GRCh38
rs770325919 Chr.5: 35014040-35014040 on Build GRCh38
rs546297610 Chr.5: 35014144-35014144 on Build GRCh38
rs1042206641 Chr.5: 35013959-35013959 on Build GRCh38
rs945270372 Chr.5: 35013934-35013934 on Build GRCh38
rs753764849 Chr.5: 35014086-35014086 on Build GRCh38
rs202180854 Chr.5: 35014083-35014083 on Build GRCh38
rs765566655 Chr.5: 35013986-35013986 on Build GRCh38
rs375108770 Chr.5: 35013892-35013892 on Build GRCh38
rs192468496 Chr.5: 35013979-35013979 on Build GRCh38
rs781291251 Chr.5: 35014005-35014005 on Build GRCh38
rs745553045 Chr.5: 35014104-35014104 on Build GRCh38
rs770704391 Chr.5: 35013962-35013962 on Build GRCh38
rs927298965 Chr.5: 35013893-35013893 on Build GRCh38
rs146930891 Chr.5: 35014003-35014003 on Build GRCh38
rs773712097 Chr.5: 35014122-35014122 on Build GRCh38

View Full Product Details