Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 14611356-14611915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM105A
Entrez Gene ID
54491
Gene Name
family with sequence similarity 105 member A
Gene Aliases
NET20
UniGene
Hs.155085
Gene Chromosome Location
Chr.5: 14581782-14616180 on Build GRCh38
Transcripts
NM_019018.2 XM_011514056.2

SNP Detail

SNP ID Chromosome Location
rs58624738 Chr.5: 14611690-14611690 on Build GRCh38
rs890958061 Chr.5: 14611706-14611706 on Build GRCh38
rs898418643 Chr.5: 14611632-14611632 on Build GRCh38
rs200034784 Chr.5: 14611705-14611706 on Build GRCh38
rs551560335 Chr.5: 14611723-14611723 on Build GRCh38
rs139002530 Chr.5: 14611816-14611816 on Build GRCh38
rs896220163 Chr.5: 14611445-14611445 on Build GRCh38
rs936984244 Chr.5: 14611537-14611537 on Build GRCh38
rs779090099 Chr.5: 14611569-14611569 on Build GRCh38
rs914843436 Chr.5: 14611403-14611403 on Build GRCh38
rs917691649 Chr.5: 14611841-14611841 on Build GRCh38
rs75475819 Chr.5: 14611689-14611689 on Build GRCh38
rs761829128 Chr.5: 14611878-14611878 on Build GRCh38
rs957617777 Chr.5: 14611426-14611426 on Build GRCh38
rs969092363 Chr.5: 14611767-14611767 on Build GRCh38
rs78445019 Chr.5: 14611717-14611717 on Build GRCh38
rs1041914591 Chr.5: 14611433-14611433 on Build GRCh38
rs1056161597 Chr.5: 14611441-14611441 on Build GRCh38
rs1044725646 Chr.5: 14611511-14611511 on Build GRCh38
rs1012353442 Chr.5: 14611448-14611448 on Build GRCh38
rs529717226 Chr.5: 14611561-14611561 on Build GRCh38
rs561722949 Chr.5: 14611657-14611657 on Build GRCh38
rs776122186 Chr.5: 14611746-14611746 on Build GRCh38
rs761079930 Chr.5: 14611752-14611752 on Build GRCh38
rs759950882 Chr.5: 14611666-14611666 on Build GRCh38
rs981149277 Chr.5: 14611519-14611519 on Build GRCh38
rs953481065 Chr.5: 14611731-14611731 on Build GRCh38
rs533179444 Chr.5: 14611490-14611490 on Build GRCh38
rs947661330 Chr.5: 14611409-14611409 on Build GRCh38
rs533584685 Chr.5: 14611790-14611790 on Build GRCh38
rs929844234 Chr.5: 14611643-14611643 on Build GRCh38
rs1020443727 Chr.5: 14611866-14611866 on Build GRCh38
rs772960009 Chr.5: 14611514-14611514 on Build GRCh38
rs887458239 Chr.5: 14611880-14611880 on Build GRCh38
rs977658641 Chr.5: 14611777-14611777 on Build GRCh38
rs887728713 Chr.5: 14611525-14611525 on Build GRCh38
rs543456671 Chr.5: 14611641-14611641 on Build GRCh38
rs529240426 Chr.5: 14611683-14611683 on Build GRCh38
rs551995061 Chr.5: 14611794-14611794 on Build GRCh38
rs531210138 Chr.5: 14611847-14611847 on Build GRCh38
rs949844337 Chr.5: 14611479-14611479 on Build GRCh38
rs941662181 Chr.5: 14611884-14611884 on Build GRCh38
rs938878570 Chr.5: 14611796-14611796 on Build GRCh38
rs144621933 Chr.5: 14611702-14611702 on Build GRCh38
rs1038016695 Chr.5: 14611616-14611616 on Build GRCh38
rs28007 Chr.5: 14611610-14611610 on Build GRCh38
rs1019430967 Chr.5: 14611733-14611733 on Build GRCh38
rs900563524 Chr.5: 14611756-14611756 on Build GRCh38
rs924792734 Chr.5: 14611435-14611435 on Build GRCh38
rs17296475 Chr.5: 14611855-14611855 on Build GRCh38
rs11750625 Chr.5: 14611439-14611439 on Build GRCh38

View Full Product Details