Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 74722231-74722805 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFM2
Entrez Gene ID
84340
Gene Name
G elongation factor mitochondrial 2
Gene Aliases
EF-G2mt, EFG2, MRRF2, MST027, MSTP027, RRF2, RRF2mt, hEFG2, mEF-G 2
UniGene
Hs.277154
Gene Chromosome Location
Chr.5: 74721204-74767371 on Build GRCh38
Transcripts
NM_001281302.1 NM_032380.4 NM_170691.2 NR_104006.1 XM_017009986.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6670520 c.2157G>C p.Q719H Chr.5: 74722433-74722433 on Build GRCh38
4414855 c.2175A>C p.K725N Chr.5: 74722415-74722415 on Build GRCh38
1695874 c.2099A>G p.Y700C Chr.5: 74722491-74722491 on Build GRCh38
3828404 c.2093G>T p.R698I Chr.5: 74722497-74722497 on Build GRCh38
4889804 c.2041G>T p.A681S Chr.5: 74722549-74722549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs71600438 Chr.5: 74722252-74722253 on Build GRCh38
rs375372058 Chr.5: 74722508-74722508 on Build GRCh38
rs780138908 Chr.5: 74722366-74722366 on Build GRCh38
rs749490366 Chr.5: 74722378-74722378 on Build GRCh38
rs770911457 Chr.5: 74722384-74722384 on Build GRCh38
rs750022497 Chr.5: 74722337-74722346 on Build GRCh38
rs869320704 Chr.5: 74722562-74722562 on Build GRCh38
rs111819315 Chr.5: 74722315-74722315 on Build GRCh38
rs529153433 Chr.5: 74722321-74722321 on Build GRCh38
rs954683301 Chr.5: 74722649-74722649 on Build GRCh38
rs375349328 Chr.5: 74722600-74722600 on Build GRCh38
rs762523757 Chr.5: 74722610-74722610 on Build GRCh38
rs888938693 Chr.5: 74722736-74722745 on Build GRCh38
rs963430692 Chr.5: 74722314-74722314 on Build GRCh38
rs375258233 Chr.5: 74722352-74722352 on Build GRCh38
rs549225329 Chr.5: 74722350-74722350 on Build GRCh38
rs140072367 Chr.5: 74722762-74722762 on Build GRCh38
rs925670695 Chr.5: 74722777-74722778 on Build GRCh38
rs752041066 Chr.5: 74722445-74722445 on Build GRCh38
rs17856871 Chr.5: 74722386-74722386 on Build GRCh38
rs762565992 Chr.5: 74722438-74722438 on Build GRCh38
rs1029033234 Chr.5: 74722704-74722704 on Build GRCh38
rs780042669 Chr.5: 74722495-74722495 on Build GRCh38
rs534968782 Chr.5: 74722518-74722518 on Build GRCh38
rs746317341 Chr.5: 74722355-74722355 on Build GRCh38
rs367676220 Chr.5: 74722583-74722583 on Build GRCh38
rs774771409 Chr.5: 74722603-74722619 on Build GRCh38
rs776564805 Chr.5: 74722389-74722389 on Build GRCh38
rs186461915 Chr.5: 74722344-74722344 on Build GRCh38
rs988674960 Chr.5: 74722771-74722771 on Build GRCh38
rs746085574 Chr.5: 74722604-74722608 on Build GRCh38
rs768900309 Chr.5: 74722379-74722379 on Build GRCh38
rs1032838417 Chr.5: 74722744-74722744 on Build GRCh38
rs770719415 Chr.5: 74722545-74722545 on Build GRCh38
rs528813726 Chr.5: 74722274-74722274 on Build GRCh38
rs757966559 Chr.5: 74722342-74722344 on Build GRCh38
rs994908243 Chr.5: 74722704-74722706 on Build GRCh38
rs551162666 Chr.5: 74722672-74722672 on Build GRCh38
rs558329606 Chr.5: 74722695-74722695 on Build GRCh38
rs548598663 Chr.5: 74722777-74722777 on Build GRCh38
rs143306569 Chr.5: 74722407-74722407 on Build GRCh38
rs532754381 Chr.5: 74722307-74722307 on Build GRCh38
rs957007057 Chr.5: 74722783-74722783 on Build GRCh38
rs750853129 Chr.5: 74722476-74722476 on Build GRCh38
rs915769978 Chr.5: 74722544-74722544 on Build GRCh38
rs536329331 Chr.5: 74722766-74722766 on Build GRCh38
rs905377762 Chr.5: 74722387-74722387 on Build GRCh38
rs773579064 Chr.5: 74722454-74722456 on Build GRCh38
rs763774517 Chr.5: 74722444-74722444 on Build GRCh38
rs764685887 Chr.5: 74722429-74722429 on Build GRCh38
rs1029128378 Chr.5: 74722323-74722323 on Build GRCh38
rs1036380796 Chr.5: 74722677-74722677 on Build GRCh38
rs1009077400 Chr.5: 74722742-74722742 on Build GRCh38
rs200151202 Chr.5: 74722426-74722426 on Build GRCh38
rs771141000 Chr.5: 74722587-74722590 on Build GRCh38
rs749668551 Chr.5: 74722583-74722603 on Build GRCh38
rs72160488 Chr.5: 74722253-74722254 on Build GRCh38
rs139397531 Chr.5: 74722380-74722380 on Build GRCh38
rs113923552 Chr.5: 74722294-74722294 on Build GRCh38
rs779505164 Chr.5: 74722369-74722371 on Build GRCh38
rs779181943 Chr.5: 74722525-74722525 on Build GRCh38
rs569969133 Chr.5: 74722303-74722304 on Build GRCh38
rs760997924 Chr.5: 74722659-74722662 on Build GRCh38
rs748698060 Chr.5: 74722521-74722522 on Build GRCh38
rs906566879 Chr.5: 74722251-74722251 on Build GRCh38
rs775015994 Chr.5: 74722430-74722430 on Build GRCh38
rs778101294 Chr.5: 74722581-74722585 on Build GRCh38
rs756592166 Chr.5: 74722491-74722491 on Build GRCh38
rs565524136 Chr.5: 74722299-74722299 on Build GRCh38
rs762257781 Chr.5: 74722456-74722456 on Build GRCh38
rs145802268 Chr.5: 74722553-74722553 on Build GRCh38
rs964579316 Chr.5: 74722678-74722678 on Build GRCh38
rs1037962828 Chr.5: 74722428-74722428 on Build GRCh38
rs756264556 Chr.5: 74722360-74722360 on Build GRCh38
rs70976129 Chr.5: 74722260-74722261 on Build GRCh38
rs756664066 Chr.5: 74722537-74722540 on Build GRCh38
rs748309595 Chr.5: 74722542-74722542 on Build GRCh38
rs978970019 Chr.5: 74722691-74722691 on Build GRCh38
rs772321151 Chr.5: 74722608-74722612 on Build GRCh38
rs1009000624 Chr.5: 74722656-74722656 on Build GRCh38
rs866813195 Chr.5: 74722619-74722619 on Build GRCh38
rs1004496217 Chr.5: 74722632-74722632 on Build GRCh38
rs767924691 Chr.5: 74722475-74722475 on Build GRCh38
rs949861101 Chr.5: 74722564-74722567 on Build GRCh38
rs745828669 Chr.5: 74722567-74722567 on Build GRCh38

View Full Product Details