Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 111660267-111660866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FYN
Entrez Gene ID
2534
Gene Name
FYN proto-oncogene, Src family tyrosine kinase
Gene Aliases
SLK, SYN, p59-FYN
UniGene
Hs.390567
Gene Chromosome Location
Chr.6: 111660332-111873452 on Build GRCh38
Transcripts
NM_002037.5 NM_153047.3 NM_153048.3 XM_005266890.3 XM_005266892.3 XM_017010650.1 XM_017010651.1 XM_017010652.1 XM_017010653.1 XM_017010654.1

SNP Detail

SNP ID Chromosome Location
rs942147560 Chr.6: 111660784-111660784 on Build GRCh38
rs758994367 Chr.6: 111660318-111660321 on Build GRCh38
rs377060992 Chr.6: 111660308-111660308 on Build GRCh38
rs554069189 Chr.6: 111660361-111660361 on Build GRCh38
rs79741507 Chr.6: 111660685-111660685 on Build GRCh38
rs553869194 Chr.6: 111660319-111660319 on Build GRCh38
rs181495290 Chr.6: 111660493-111660493 on Build GRCh38
rs535807203 Chr.6: 111660403-111660406 on Build GRCh38
rs990204713 Chr.6: 111660549-111660552 on Build GRCh38
rs963821727 Chr.6: 111660527-111660527 on Build GRCh38
rs1012008325 Chr.6: 111660332-111660332 on Build GRCh38
rs1022219560 Chr.6: 111660662-111660662 on Build GRCh38
rs548117220 Chr.6: 111660822-111660822 on Build GRCh38
rs374318910 Chr.6: 111660608-111660608 on Build GRCh38
rs1016663111 Chr.6: 111660347-111660348 on Build GRCh38
rs1056010139 Chr.6: 111660317-111660317 on Build GRCh38
rs920501801 Chr.6: 111660363-111660363 on Build GRCh38
rs185336969 Chr.6: 111660712-111660712 on Build GRCh38
rs1004048892 Chr.6: 111660552-111660552 on Build GRCh38
rs898268510 Chr.6: 111660838-111660838 on Build GRCh38
rs542612763 Chr.6: 111660619-111660619 on Build GRCh38
rs1003122745 Chr.6: 111660299-111660299 on Build GRCh38
rs532678883 Chr.6: 111660719-111660719 on Build GRCh38
rs955331565 Chr.6: 111660406-111660406 on Build GRCh38
rs946102725 Chr.6: 111660607-111660607 on Build GRCh38
rs894666817 Chr.6: 111660769-111660769 on Build GRCh38
rs1056021527 Chr.6: 111660733-111660733 on Build GRCh38
rs1047596698 Chr.6: 111660490-111660490 on Build GRCh38
rs200969046 Chr.6: 111660321-111660322 on Build GRCh38
rs367630294 Chr.6: 111660407-111660410 on Build GRCh38
rs987648951 Chr.6: 111660786-111660786 on Build GRCh38
rs929173061 Chr.6: 111660476-111660476 on Build GRCh38
rs938923915 Chr.6: 111660687-111660687 on Build GRCh38
rs1010900775 Chr.6: 111660631-111660631 on Build GRCh38
rs943029298 Chr.6: 111660324-111660324 on Build GRCh38
rs906341879 Chr.6: 111660521-111660521 on Build GRCh38
rs993877096 Chr.6: 111660397-111660397 on Build GRCh38
rs1055197334 Chr.6: 111660569-111660569 on Build GRCh38
rs955556947 Chr.6: 111660734-111660734 on Build GRCh38
rs531706781 Chr.6: 111660679-111660679 on Build GRCh38
rs561003544 Chr.6: 111660667-111660667 on Build GRCh38
rs894615834 Chr.6: 111660322-111660322 on Build GRCh38
rs572449019 Chr.6: 111660339-111660339 on Build GRCh38
rs1038263553 Chr.6: 111660801-111660801 on Build GRCh38

View Full Product Details