Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 244422709-244423187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADSS
Entrez Gene ID
159
Gene Name
adenylosuccinate synthase
Gene Aliases
ADEH, ADSS 2
UniGene
Hs.498313
Gene Chromosome Location
Chr.1: 244408492-244452134 on Build GRCh38
Transcripts
NM_001126.3 XM_011544108.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
385012 c.647A>C p.E216A Chr.1: 244422851-244422851 on Build GRCh38
4953913 c.598A>G p.N200D Chr.1: 244422900-244422900 on Build GRCh38
6400391 c.656A>G p.K219R Chr.1: 244422842-244422842 on Build GRCh38
3804605 c.640G>C p.E214Q Chr.1: 244422858-244422858 on Build GRCh38
5470011 c.614T>C p.I205T Chr.1: 244422884-244422884 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562997493 Chr.1: 244423029-244423029 on Build GRCh38
rs777053387 Chr.1: 244422800-244422800 on Build GRCh38
rs770315996 Chr.1: 244422968-244422968 on Build GRCh38
rs999732609 Chr.1: 244423165-244423165 on Build GRCh38
rs746570731 Chr.1: 244422941-244422941 on Build GRCh38
rs542423992 Chr.1: 244423120-244423120 on Build GRCh38
rs374379621 Chr.1: 244422920-244422920 on Build GRCh38
rs894864597 Chr.1: 244423077-244423077 on Build GRCh38
rs779604367 Chr.1: 244422935-244422935 on Build GRCh38
rs116235277 Chr.1: 244422739-244422739 on Build GRCh38
rs989080372 Chr.1: 244423151-244423151 on Build GRCh38
rs140299584 Chr.1: 244422790-244422790 on Build GRCh38
rs909827662 Chr.1: 244422940-244422940 on Build GRCh38
rs748474228 Chr.1: 244422966-244422966 on Build GRCh38
rs906694363 Chr.1: 244423099-244423099 on Build GRCh38
rs377468008 Chr.1: 244422931-244422931 on Build GRCh38
rs1003343702 Chr.1: 244423024-244423024 on Build GRCh38
rs780722985 Chr.1: 244422958-244422958 on Build GRCh38
rs1030708204 Chr.1: 244422757-244422757 on Build GRCh38
rs774342769 Chr.1: 244422866-244422866 on Build GRCh38
rs929928461 Chr.1: 244422999-244422999 on Build GRCh38
rs766537315 Chr.1: 244422864-244422864 on Build GRCh38
rs956568473 Chr.1: 244422771-244422771 on Build GRCh38
rs1003330290 Chr.1: 244423108-244423108 on Build GRCh38
rs74692652 Chr.1: 244423009-244423009 on Build GRCh38
rs770367468 Chr.1: 244422826-244422826 on Build GRCh38
rs752274725 Chr.1: 244422813-244422814 on Build GRCh38
rs1041523841 Chr.1: 244422949-244422949 on Build GRCh38
rs145206930 Chr.1: 244423157-244423157 on Build GRCh38
rs1056239602 Chr.1: 244423042-244423042 on Build GRCh38
rs112819847 Chr.1: 244423053-244423053 on Build GRCh38
rs982252726 Chr.1: 244422744-244422744 on Build GRCh38
rs753601100 Chr.1: 244422887-244422887 on Build GRCh38
rs562222566 Chr.1: 244422774-244422774 on Build GRCh38
rs377457816 Chr.1: 244423018-244423018 on Build GRCh38
rs545919383 Chr.1: 244423010-244423010 on Build GRCh38
rs966140027 Chr.1: 244422762-244422762 on Build GRCh38
rs111332377 Chr.1: 244423012-244423012 on Build GRCh38
rs777569716 Chr.1: 244422946-244422947 on Build GRCh38
rs779529210 Chr.1: 244422791-244422791 on Build GRCh38
rs750388925 Chr.1: 244422933-244422933 on Build GRCh38
rs763089080 Chr.1: 244422857-244422857 on Build GRCh38
rs897187890 Chr.1: 244422975-244422975 on Build GRCh38
rs1031137163 Chr.1: 244423116-244423116 on Build GRCh38
rs576486623 Chr.1: 244423032-244423032 on Build GRCh38
rs748469520 Chr.1: 244422819-244422819 on Build GRCh38
rs1011940298 Chr.1: 244423089-244423089 on Build GRCh38
rs755730977 Chr.1: 244422841-244422866 on Build GRCh38
rs746655176 Chr.1: 244422794-244422794 on Build GRCh38
rs754596567 Chr.1: 244422944-244422944 on Build GRCh38
rs759789521 Chr.1: 244422875-244422875 on Build GRCh38
rs1036905023 Chr.1: 244423016-244423016 on Build GRCh38
rs915119000 Chr.1: 244422777-244422777 on Build GRCh38
rs767698508 Chr.1: 244422886-244422886 on Build GRCh38
rs773878391 Chr.1: 244422844-244422844 on Build GRCh38

View Full Product Details