Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 4842373-4842969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR1E2
Entrez Gene ID
83592
Gene Name
aldo-keto reductase family 1 member E2
Gene Aliases
AKR1CL2, AKRDC1, LoopADR, hTSP, htAKR
UniGene
Hs.657944
Gene Chromosome Location
Chr.10: 4824554-4908421 on Build GRCh38
Transcripts
NM_001040177.2 NM_001271021.1 NM_001271025.1 NR_073125.1 NR_073126.1 NR_073127.1 XM_011519715.2 XM_011519718.2 XM_011519720.2 XM_011519722.2 XM_011519724.2 XM_011519725.2 XM_017016743.1 XM_017016744.1 XM_017016745.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7114730 c.777G>T p.Q259H Chr.10: 4842444-4842444 on Build GRCh38
159006 c.792G>A p.V264V Chr.10: 4842459-4842459 on Build GRCh38
5681487 c.836A>G p.Q279R Chr.10: 4842503-4842503 on Build GRCh38
6988451 c.769C>T p.Q257* Chr.10: 4842436-4842436 on Build GRCh38
5663593 c.800G>A p.G267E Chr.10: 4842467-4842467 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779483522 Chr.10: 4842445-4842445 on Build GRCh38
rs1023266857 Chr.10: 4842761-4842761 on Build GRCh38
rs377060185 Chr.10: 4842418-4842418 on Build GRCh38
rs11351812 Chr.10: 4842942-4842942 on Build GRCh38
rs397847099 Chr.10: 4842944-4842944 on Build GRCh38
rs1053247023 Chr.10: 4842401-4842401 on Build GRCh38
rs1050169455 Chr.10: 4842745-4842745 on Build GRCh38
rs985981086 Chr.10: 4842819-4842819 on Build GRCh38
rs758856697 Chr.10: 4842552-4842552 on Build GRCh38
rs765007557 Chr.10: 4842490-4842490 on Build GRCh38
rs1003576162 Chr.10: 4842883-4842883 on Build GRCh38
rs776351950 Chr.10: 4842466-4842466 on Build GRCh38
rs539997937 Chr.10: 4842732-4842732 on Build GRCh38
rs537856364 Chr.10: 4842924-4842924 on Build GRCh38
rs1000602978 Chr.10: 4842599-4842599 on Build GRCh38
rs930239658 Chr.10: 4842849-4842849 on Build GRCh38
rs1044816146 Chr.10: 4842898-4842898 on Build GRCh38
rs1003132968 Chr.10: 4842908-4842908 on Build GRCh38
rs2961593 Chr.10: 4842771-4842771 on Build GRCh38
rs764286688 Chr.10: 4842818-4842818 on Build GRCh38
rs1011246141 Chr.10: 4842934-4842934 on Build GRCh38
rs763631105 Chr.10: 4842510-4842510 on Build GRCh38
rs1015808182 Chr.10: 4842901-4842901 on Build GRCh38
rs201427808 Chr.10: 4842476-4842476 on Build GRCh38
rs934737363 Chr.10: 4842399-4842399 on Build GRCh38
rs1025085278 Chr.10: 4842875-4842875 on Build GRCh38
rs990411582 Chr.10: 4842610-4842610 on Build GRCh38
rs12573690 Chr.10: 4842616-4842616 on Build GRCh38
rs918815987 Chr.10: 4842848-4842848 on Build GRCh38
rs932653538 Chr.10: 4842733-4842733 on Build GRCh38
rs2961592 Chr.10: 4842608-4842608 on Build GRCh38
rs551678940 Chr.10: 4842833-4842833 on Build GRCh38
rs759418870 Chr.10: 4842396-4842396 on Build GRCh38
rs989288253 Chr.10: 4842589-4842589 on Build GRCh38
rs1026760859 Chr.10: 4842516-4842516 on Build GRCh38
rs923772812 Chr.10: 4842574-4842574 on Build GRCh38
rs544188147 Chr.10: 4842779-4842779 on Build GRCh38
rs906194544 Chr.10: 4842906-4842906 on Build GRCh38
rs772364315 Chr.10: 4842468-4842471 on Build GRCh38
rs763570731 Chr.10: 4842430-4842430 on Build GRCh38
rs982002539 Chr.10: 4842567-4842567 on Build GRCh38
rs755488577 Chr.10: 4842444-4842444 on Build GRCh38
rs186509291 Chr.10: 4842539-4842539 on Build GRCh38
rs911231875 Chr.10: 4842595-4842595 on Build GRCh38
rs556440074 Chr.10: 4842602-4842602 on Build GRCh38
rs139045828 Chr.10: 4842814-4842814 on Build GRCh38
rs940169907 Chr.10: 4842685-4842685 on Build GRCh38
rs1050202313 Chr.10: 4842449-4842449 on Build GRCh38
rs2924269 Chr.10: 4842592-4842592 on Build GRCh38
rs970411854 Chr.10: 4842878-4842878 on Build GRCh38
rs534060027 Chr.10: 4842532-4842532 on Build GRCh38
rs1030903923 Chr.10: 4842787-4842787 on Build GRCh38
rs34722294 Chr.10: 4842652-4842653 on Build GRCh38
rs1003396879 Chr.10: 4842775-4842775 on Build GRCh38
rs151134642 Chr.10: 4842531-4842531 on Build GRCh38
rs80210984 Chr.10: 4842854-4842854 on Build GRCh38
rs561820595 Chr.10: 4842736-4842736 on Build GRCh38
rs745771866 Chr.10: 4842871-4842871 on Build GRCh38
rs911225548 Chr.10: 4842823-4842823 on Build GRCh38
rs12570011 Chr.10: 4842529-4842529 on Build GRCh38
rs981988847 Chr.10: 4842504-4842504 on Build GRCh38
rs376995152 Chr.10: 4842429-4842429 on Build GRCh38
rs368223677 Chr.10: 4842464-4842464 on Build GRCh38
rs745450309 Chr.10: 4842505-4842505 on Build GRCh38
rs1049709119 Chr.10: 4842594-4842594 on Build GRCh38
rs748657385 Chr.10: 4842447-4842447 on Build GRCh38
rs368678588 Chr.10: 4842546-4842546 on Build GRCh38
rs771162913 Chr.10: 4842465-4842465 on Build GRCh38
rs143113841 Chr.10: 4842431-4842431 on Build GRCh38
rs149945552 Chr.10: 4842846-4842846 on Build GRCh38
rs539936669 Chr.10: 4842397-4842397 on Build GRCh38
rs562788615 Chr.10: 4842810-4842810 on Build GRCh38
rs78987748 Chr.10: 4842858-4842858 on Build GRCh38
rs897363814 Chr.10: 4842502-4842502 on Build GRCh38
rs141274888 Chr.10: 4842493-4842493 on Build GRCh38
rs989303793 Chr.10: 4842811-4842811 on Build GRCh38
rs12573669 Chr.10: 4842450-4842450 on Build GRCh38
rs757922663 Chr.10: 4842420-4842420 on Build GRCh38
rs981704081 Chr.10: 4842630-4842630 on Build GRCh38
rs944046089 Chr.10: 4842824-4842824 on Build GRCh38
rs191697759 Chr.10: 4842716-4842716 on Build GRCh38
rs892343410 Chr.10: 4842929-4842929 on Build GRCh38
rs77033958 Chr.10: 4842618-4842618 on Build GRCh38
rs866808717 Chr.10: 4842581-4842581 on Build GRCh38
rs753213534 Chr.10: 4842550-4842550 on Build GRCh38
rs950252366 Chr.10: 4842494-4842494 on Build GRCh38
rs537846133 Chr.10: 4842593-4842593 on Build GRCh38
rs150640934 Chr.10: 4842506-4842506 on Build GRCh38
rs974211717 Chr.10: 4842912-4842912 on Build GRCh38
rs773567423 Chr.10: 4842674-4842674 on Build GRCh38
rs567702611 Chr.10: 4842880-4842880 on Build GRCh38
rs994376843 Chr.10: 4842642-4842642 on Build GRCh38

View Full Product Details