Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 110509891-110510447 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP5
Entrez Gene ID
1847
Gene Name
dual specificity phosphatase 5
Gene Aliases
DUSP, HVH3
UniGene
Hs.2128
Gene Chromosome Location
Chr.10: 110497867-110511544 on Build GRCh38
Transcripts
NM_004419.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
914728 c.759G>C p.R253S Chr.10: 110510030-110510030 on Build GRCh38
4558665 c.775G>A p.V259I Chr.10: 110510046-110510046 on Build GRCh38
6202730 c.995A>G p.E332G Chr.10: 110510266-110510266 on Build GRCh38
7365674 c.879C>G p.I293M Chr.10: 110510150-110510150 on Build GRCh38
4457280 c.1039C>T p.P347S Chr.10: 110510310-110510310 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372005583 Chr.10: 110509994-110509994 on Build GRCh38
rs757769176 Chr.10: 110510022-110510022 on Build GRCh38
rs893025336 Chr.10: 110510420-110510420 on Build GRCh38
rs563414179 Chr.10: 110510193-110510193 on Build GRCh38
rs773603037 Chr.10: 110510251-110510251 on Build GRCh38
rs373968247 Chr.10: 110509934-110509934 on Build GRCh38
rs773687227 Chr.10: 110510348-110510348 on Build GRCh38
rs114603867 Chr.10: 110510234-110510234 on Build GRCh38
rs370107033 Chr.10: 110510384-110510384 on Build GRCh38
rs367814937 Chr.10: 110510167-110510167 on Build GRCh38
rs200101719 Chr.10: 110510317-110510317 on Build GRCh38
rs750324864 Chr.10: 110510318-110510318 on Build GRCh38
rs768380350 Chr.10: 110510331-110510331 on Build GRCh38
rs745402177 Chr.10: 110510116-110510116 on Build GRCh38
rs767380690 Chr.10: 110510166-110510166 on Build GRCh38
rs765385201 Chr.10: 110510365-110510365 on Build GRCh38
rs777615224 Chr.10: 110510090-110510090 on Build GRCh38
rs765477721 Chr.10: 110510311-110510311 on Build GRCh38
rs778351910 Chr.10: 110510069-110510069 on Build GRCh38
rs878999495 Chr.10: 110510245-110510245 on Build GRCh38
rs374165559 Chr.10: 110510137-110510137 on Build GRCh38
rs111816223 Chr.10: 110510141-110510141 on Build GRCh38
rs562848514 Chr.10: 110510134-110510134 on Build GRCh38
rs897067064 Chr.10: 110510101-110510111 on Build GRCh38
rs142629528 Chr.10: 110510376-110510376 on Build GRCh38
rs997703722 Chr.10: 110510120-110510120 on Build GRCh38
rs748846234 Chr.10: 110510330-110510330 on Build GRCh38
rs150805476 Chr.10: 110510359-110510359 on Build GRCh38
rs565671192 Chr.10: 110510339-110510339 on Build GRCh38
rs761067494 Chr.10: 110510349-110510349 on Build GRCh38
rs764839233 Chr.10: 110509976-110509976 on Build GRCh38
rs747409647 Chr.10: 110510419-110510419 on Build GRCh38
rs202234029 Chr.10: 110510241-110510241 on Build GRCh38
rs76989225 Chr.10: 110510150-110510150 on Build GRCh38
rs138113379 Chr.10: 110510374-110510374 on Build GRCh38
rs775205143 Chr.10: 110510211-110510211 on Build GRCh38
rs1007584469 Chr.10: 110510294-110510294 on Build GRCh38
rs759207590 Chr.10: 110509971-110509971 on Build GRCh38
rs571909209 Chr.10: 110510288-110510288 on Build GRCh38
rs1011416689 Chr.10: 110509918-110509918 on Build GRCh38
rs767946209 Chr.10: 110510145-110510145 on Build GRCh38
rs771809739 Chr.10: 110510411-110510411 on Build GRCh38
rs777971851 Chr.10: 110510417-110510417 on Build GRCh38
rs201213844 Chr.10: 110510119-110510119 on Build GRCh38
rs769193161 Chr.10: 110510210-110510210 on Build GRCh38
rs746823964 Chr.10: 110510174-110510174 on Build GRCh38
rs966510010 Chr.10: 110510110-110510110 on Build GRCh38
rs750516594 Chr.10: 110510056-110510056 on Build GRCh38
rs754545204 Chr.10: 110510409-110510409 on Build GRCh38
rs765810015 Chr.10: 110510077-110510077 on Build GRCh38
rs201835011 Chr.10: 110510262-110510262 on Build GRCh38
rs573917228 Chr.10: 110510103-110510103 on Build GRCh38
rs970696621 Chr.10: 110510414-110510414 on Build GRCh38
rs753970682 Chr.10: 110510292-110510292 on Build GRCh38
rs368048350 Chr.10: 110510064-110510064 on Build GRCh38
rs35101549 Chr.10: 110510236-110510236 on Build GRCh38
rs751320813 Chr.10: 110510314-110510314 on Build GRCh38
rs766527682 Chr.10: 110510157-110510157 on Build GRCh38
rs371109451 Chr.10: 110510410-110510410 on Build GRCh38
rs753071501 Chr.10: 110509989-110509989 on Build GRCh38
rs555542768 Chr.10: 110510392-110510392 on Build GRCh38
rs533845858 Chr.10: 110509921-110509921 on Build GRCh38
rs776654845 Chr.10: 110510360-110510360 on Build GRCh38
rs752869412 Chr.10: 110510367-110510367 on Build GRCh38
rs1026457804 Chr.10: 110510124-110510124 on Build GRCh38
rs755166385 Chr.10: 110510308-110510308 on Build GRCh38
rs866893667 Chr.10: 110510227-110510227 on Build GRCh38
rs1054055520 Chr.10: 110510416-110510416 on Build GRCh38
rs781015299 Chr.10: 110510190-110510190 on Build GRCh38
rs771483109 Chr.10: 110510222-110510222 on Build GRCh38
rs201758099 Chr.10: 110510125-110510125 on Build GRCh38
rs372682956 Chr.10: 110510283-110510283 on Build GRCh38
rs949848741 Chr.10: 110510072-110510072 on Build GRCh38
rs201740676 Chr.10: 110510012-110510012 on Build GRCh38
rs186163304 Chr.10: 110510143-110510143 on Build GRCh38
rs940936738 Chr.10: 110510242-110510242 on Build GRCh38
rs763344116 Chr.10: 110510131-110510131 on Build GRCh38
rs763302452 Chr.10: 110510368-110510368 on Build GRCh38
rs984792700 Chr.10: 110510230-110510230 on Build GRCh38
rs141147474 Chr.10: 110510347-110510347 on Build GRCh38
rs760738832 Chr.10: 110510281-110510281 on Build GRCh38
rs574511404 Chr.10: 110510078-110510078 on Build GRCh38
rs768015125 Chr.10: 110510248-110510248 on Build GRCh38
rs747887661 Chr.10: 110510082-110510082 on Build GRCh38
rs746938026 Chr.10: 110510097-110510097 on Build GRCh38
rs138451130 Chr.10: 110510152-110510152 on Build GRCh38
rs756176909 Chr.10: 110510321-110510321 on Build GRCh38
rs373903086 Chr.10: 110510040-110510040 on Build GRCh38
rs752115032 Chr.10: 110510013-110510013 on Build GRCh38
rs549093361 Chr.10: 110510183-110510183 on Build GRCh38
rs371775746 Chr.10: 110510088-110510088 on Build GRCh38
rs774914509 Chr.10: 110510142-110510142 on Build GRCh38
rs376480519 Chr.10: 110510345-110510345 on Build GRCh38
rs142065029 Chr.10: 110510173-110510173 on Build GRCh38
rs148203909 Chr.10: 110510324-110510324 on Build GRCh38
rs748933851 Chr.10: 110510233-110510233 on Build GRCh38
rs909497603 Chr.10: 110509985-110509985 on Build GRCh38
rs980162346 Chr.10: 110510171-110510171 on Build GRCh38
rs770826313 Chr.10: 110510099-110510099 on Build GRCh38

View Full Product Details