Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 122951846-122952162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf88
Entrez Gene ID
80007
Gene Name
chromosome 10 open reading frame 88
Gene Aliases
-
UniGene
Hs.501286
Gene Chromosome Location
Chr.10: 122930903-122954403 on Build GRCh38
Transcripts
NM_024942.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3686556 c.412G>A p.E138K Chr.10: 122951983-122951983 on Build GRCh38
915729 c.376G>C p.E126Q Chr.10: 122952019-122952019 on Build GRCh38
293604 c.401delA p.N134fs*2 Chr.10: 122951994-122951994 on Build GRCh38
243273 c.419C>T p.S140F Chr.10: 122951976-122951976 on Build GRCh38
3435360 c.417C>T p.S139S Chr.10: 122951978-122951978 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs994050127 Chr.10: 122951877-122951877 on Build GRCh38
rs562776319 Chr.10: 122951967-122951967 on Build GRCh38
rs9804347 Chr.10: 122952116-122952116 on Build GRCh38
rs964804729 Chr.10: 122951994-122951994 on Build GRCh38
rs527762238 Chr.10: 122952105-122952105 on Build GRCh38
rs150582565 Chr.10: 122951983-122951983 on Build GRCh38
rs767006447 Chr.10: 122951935-122951935 on Build GRCh38
rs757240069 Chr.10: 122952009-122952009 on Build GRCh38
rs952104934 Chr.10: 122952111-122952111 on Build GRCh38
rs974803109 Chr.10: 122952001-122952001 on Build GRCh38
rs754346974 Chr.10: 122951942-122951942 on Build GRCh38
rs564486207 Chr.10: 122952129-122952129 on Build GRCh38
rs758702501 Chr.10: 122951979-122951979 on Build GRCh38
rs180759084 Chr.10: 122952014-122952014 on Build GRCh38
rs147647614 Chr.10: 122952074-122952079 on Build GRCh38
rs764054770 Chr.10: 122951994-122951994 on Build GRCh38
rs1006201724 Chr.10: 122951937-122951937 on Build GRCh38
rs954868911 Chr.10: 122952110-122952110 on Build GRCh38
rs1014501614 Chr.10: 122952044-122952044 on Build GRCh38
rs201523441 Chr.10: 122951990-122951990 on Build GRCh38
rs879244137 Chr.10: 122951897-122951897 on Build GRCh38
rs1025557967 Chr.10: 122951924-122951924 on Build GRCh38
rs755423297 Chr.10: 122951946-122951946 on Build GRCh38
rs370703984 Chr.10: 122951953-122951953 on Build GRCh38
rs776514590 Chr.10: 122952114-122952114 on Build GRCh38
rs751388819 Chr.10: 122951993-122951994 on Build GRCh38
rs199979287 Chr.10: 122952047-122952048 on Build GRCh38
rs768402613 Chr.10: 122951904-122951904 on Build GRCh38
rs766929672 Chr.10: 122952042-122952045 on Build GRCh38
rs768562196 Chr.10: 122952018-122952018 on Build GRCh38
rs774258496 Chr.10: 122952073-122952074 on Build GRCh38
rs761402450 Chr.10: 122952028-122952033 on Build GRCh38
rs774104116 Chr.10: 122951906-122951906 on Build GRCh38
rs761577940 Chr.10: 122951907-122951907 on Build GRCh38
rs774192106 Chr.10: 122952041-122952041 on Build GRCh38
rs767064780 Chr.10: 122951909-122951909 on Build GRCh38
rs548535561 Chr.10: 122952080-122952080 on Build GRCh38
rs780002117 Chr.10: 122952011-122952011 on Build GRCh38
rs772845810 Chr.10: 122951923-122951923 on Build GRCh38
rs891661621 Chr.10: 122952019-122952019 on Build GRCh38
rs146050277 Chr.10: 122951926-122951926 on Build GRCh38
rs999952057 Chr.10: 122951975-122951975 on Build GRCh38
rs996543503 Chr.10: 122951875-122951875 on Build GRCh38

View Full Product Details