Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 117290358-117290930 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BACE1
Entrez Gene ID
23621
Gene Name
beta-secretase 1
Gene Aliases
ASP2, BACE, HSPC104
UniGene
Hs.504003
Gene Chromosome Location
Chr.11: 117285686-117316256 on Build GRCh38
Transcripts
NM_001207048.1 NM_001207049.1 NM_012104.4 NM_138971.3 NM_138972.3 NM_138973.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3443771 c.1127C>T p.S376F Chr.11: 117290625-117290625 on Build GRCh38
6291341 c.1143C>T p.Y381Y Chr.11: 117290609-117290609 on Build GRCh38
4400030 c.1140T>G p.C380W Chr.11: 117290612-117290612 on Build GRCh38
6862766 c.1126T>C p.S376P Chr.11: 117290626-117290626 on Build GRCh38
7204972 c.1110G>A p.V370V Chr.11: 117290642-117290642 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779869666 Chr.11: 117290893-117290893 on Build GRCh38
rs761381483 Chr.11: 117290856-117290856 on Build GRCh38
rs750140317 Chr.11: 117290884-117290884 on Build GRCh38
rs985291140 Chr.11: 117290487-117290487 on Build GRCh38
rs746214847 Chr.11: 117290644-117290644 on Build GRCh38
rs546758177 Chr.11: 117290558-117290558 on Build GRCh38
rs999177859 Chr.11: 117290701-117290701 on Build GRCh38
rs901908458 Chr.11: 117290849-117290849 on Build GRCh38
rs377169130 Chr.11: 117290591-117290591 on Build GRCh38
rs775285842 Chr.11: 117290708-117290708 on Build GRCh38
rs369598078 Chr.11: 117290872-117290872 on Build GRCh38
rs778429080 Chr.11: 117290850-117290850 on Build GRCh38
rs985075588 Chr.11: 117290798-117290798 on Build GRCh38
rs776132225 Chr.11: 117290514-117290514 on Build GRCh38
rs745392028 Chr.11: 117290897-117290898 on Build GRCh38
rs979746829 Chr.11: 117290764-117290764 on Build GRCh38
rs780406532 Chr.11: 117290663-117290663 on Build GRCh38
rs758012470 Chr.11: 117290885-117290885 on Build GRCh38
rs753376097 Chr.11: 117290625-117290625 on Build GRCh38
rs1012121783 Chr.11: 117290732-117290732 on Build GRCh38
rs3214855 Chr.11: 117290707-117290707 on Build GRCh38
rs758837806 Chr.11: 117290650-117290650 on Build GRCh38
rs568682860 Chr.11: 117290765-117290765 on Build GRCh38
rs886304295 Chr.11: 117290517-117290517 on Build GRCh38
rs940491324 Chr.11: 117290816-117290816 on Build GRCh38
rs569648185 Chr.11: 117290485-117290485 on Build GRCh38
rs139977300 Chr.11: 117290587-117290587 on Build GRCh38
rs759218078 Chr.11: 117290696-117290696 on Build GRCh38
rs747372765 Chr.11: 117290762-117290762 on Build GRCh38
rs773546095 Chr.11: 117290695-117290695 on Build GRCh38
rs778686608 Chr.11: 117290637-117290637 on Build GRCh38
rs755908169 Chr.11: 117290794-117290794 on Build GRCh38
rs548729769 Chr.11: 117290685-117290685 on Build GRCh38
rs748232622 Chr.11: 117290910-117290910 on Build GRCh38
rs374894390 Chr.11: 117290681-117290681 on Build GRCh38
rs75519595 Chr.11: 117290627-117290627 on Build GRCh38
rs896131824 Chr.11: 117290710-117290710 on Build GRCh38
rs370158287 Chr.11: 117290618-117290618 on Build GRCh38
rs533069014 Chr.11: 117290542-117290542 on Build GRCh38
rs938891078 Chr.11: 117290827-117290827 on Build GRCh38
rs988932097 Chr.11: 117290430-117290430 on Build GRCh38
rs150730974 Chr.11: 117290582-117290582 on Build GRCh38
rs878995888 Chr.11: 117290458-117290458 on Build GRCh38
rs748988221 Chr.11: 117290447-117290447 on Build GRCh38
rs1040235994 Chr.11: 117290823-117290823 on Build GRCh38
rs769320303 Chr.11: 117290680-117290680 on Build GRCh38
rs764022176 Chr.11: 117290851-117290851 on Build GRCh38
rs984639856 Chr.11: 117290820-117290820 on Build GRCh38
rs755599259 Chr.11: 117290616-117290616 on Build GRCh38
rs747474350 Chr.11: 117290664-117290664 on Build GRCh38
rs767393450 Chr.11: 117290702-117290702 on Build GRCh38
rs907804275 Chr.11: 117290803-117290803 on Build GRCh38
rs141126434 Chr.11: 117290498-117290498 on Build GRCh38
rs11216349 Chr.11: 117290579-117290579 on Build GRCh38
rs377157871 Chr.11: 117290524-117290524 on Build GRCh38
rs940712849 Chr.11: 117290495-117290495 on Build GRCh38
rs760563868 Chr.11: 117290711-117290711 on Build GRCh38
rs191843656 Chr.11: 117290444-117290444 on Build GRCh38
rs932109755 Chr.11: 117290864-117290864 on Build GRCh38
rs753324652 Chr.11: 117290481-117290482 on Build GRCh38
rs768126905 Chr.11: 117290466-117290466 on Build GRCh38
rs772446535 Chr.11: 117290512-117290512 on Build GRCh38
rs780581180 Chr.11: 117290700-117290700 on Build GRCh38
rs930570652 Chr.11: 117290497-117290497 on Build GRCh38
rs769891197 Chr.11: 117290689-117290689 on Build GRCh38
rs780431224 Chr.11: 117290892-117290892 on Build GRCh38
rs530797591 Chr.11: 117290480-117290480 on Build GRCh38
rs1031262289 Chr.11: 117290628-117290628 on Build GRCh38
rs372804989 Chr.11: 117290903-117290903 on Build GRCh38
rs1048922194 Chr.11: 117290501-117290501 on Build GRCh38
rs932237731 Chr.11: 117290554-117290554 on Build GRCh38
rs916954468 Chr.11: 117290760-117290760 on Build GRCh38
rs199579989 Chr.11: 117290482-117290482 on Build GRCh38
rs376210908 Chr.11: 117290489-117290489 on Build GRCh38
rs1050379258 Chr.11: 117290909-117290909 on Build GRCh38
rs185027294 Chr.11: 117290855-117290855 on Build GRCh38
rs778426625 Chr.11: 117290543-117290543 on Build GRCh38
rs371130142 Chr.11: 117290523-117290523 on Build GRCh38
rs751979949 Chr.11: 117290895-117290895 on Build GRCh38
rs368258749 Chr.11: 117290526-117290526 on Build GRCh38
rs752131557 Chr.11: 117290441-117290441 on Build GRCh38
rs550709603 Chr.11: 117290906-117290906 on Build GRCh38
rs398076273 Chr.11: 117290709-117290709 on Build GRCh38
rs949725468 Chr.11: 117290445-117290445 on Build GRCh38
rs748249027 Chr.11: 117290682-117290682 on Build GRCh38
rs990669430 Chr.11: 117290425-117290425 on Build GRCh38
rs397714166 Chr.11: 117290710-117290710 on Build GRCh38

View Full Product Details