Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7716049-7716542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPPA3
Entrez Gene ID
359787
Gene Name
developmental pluripotency associated 3
Gene Aliases
STELLA
UniGene
Hs.131358
Gene Chromosome Location
Chr.12: 7711454-7717559 on Build GRCh38
Transcripts
NM_199286.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
695750 c.351G>A p.K117K Chr.12: 7716221-7716221 on Build GRCh38
5376051 c.361G>A p.G121R Chr.12: 7716231-7716231 on Build GRCh38
695749 c.364G>T p.V122F Chr.12: 7716234-7716234 on Build GRCh38
5933031 c.362G>A p.G121E Chr.12: 7716232-7716232 on Build GRCh38
3872559 c.369G>A p.K123K Chr.12: 7716239-7716239 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1040255092 Chr.12: 7716417-7716420 on Build GRCh38
rs748272388 Chr.12: 7716291-7716291 on Build GRCh38
rs989627950 Chr.12: 7716134-7716134 on Build GRCh38
rs762737956 Chr.12: 7716196-7716196 on Build GRCh38
rs780491824 Chr.12: 7716178-7716178 on Build GRCh38
rs758892687 Chr.12: 7716282-7716282 on Build GRCh38
rs918618577 Chr.12: 7716315-7716315 on Build GRCh38
rs55889848 Chr.12: 7716188-7716189 on Build GRCh38
rs1011590864 Chr.12: 7716450-7716450 on Build GRCh38
rs746244595 Chr.12: 7716181-7716181 on Build GRCh38
rs202173543 Chr.12: 7716167-7716167 on Build GRCh38
rs767587836 Chr.12: 7716138-7716138 on Build GRCh38
rs758769599 Chr.12: 7716173-7716173 on Build GRCh38
rs781222378 Chr.12: 7716288-7716288 on Build GRCh38
rs550000938 Chr.12: 7716498-7716498 on Build GRCh38
rs978139188 Chr.12: 7716480-7716480 on Build GRCh38
rs767037763 Chr.12: 7716153-7716153 on Build GRCh38
rs766068484 Chr.12: 7716250-7716251 on Build GRCh38
rs548671879 Chr.12: 7716515-7716515 on Build GRCh38
rs397775394 Chr.12: 7716281-7716282 on Build GRCh38
rs34619457 Chr.12: 7716181-7716182 on Build GRCh38
rs386375526 Chr.12: 7716279-7716280 on Build GRCh38
rs553079854 Chr.12: 7716239-7716239 on Build GRCh38
rs767008296 Chr.12: 7716251-7716251 on Build GRCh38
rs981223316 Chr.12: 7716093-7716093 on Build GRCh38
rs751869458 Chr.12: 7716284-7716284 on Build GRCh38
rs1048197838 Chr.12: 7716323-7716323 on Build GRCh38
rs1022441786 Chr.12: 7716451-7716451 on Build GRCh38
rs765843942 Chr.12: 7716263-7716263 on Build GRCh38
rs751106554 Chr.12: 7716255-7716257 on Build GRCh38
rs752277429 Chr.12: 7716246-7716246 on Build GRCh38
rs759229149 Chr.12: 7716236-7716240 on Build GRCh38
rs759197527 Chr.12: 7716218-7716218 on Build GRCh38
rs370671962 Chr.12: 7716268-7716269 on Build GRCh38
rs754872843 Chr.12: 7716261-7716269 on Build GRCh38
rs777800952 Chr.12: 7716270-7716270 on Build GRCh38
rs939387612 Chr.12: 7716200-7716200 on Build GRCh38
rs757213567 Chr.12: 7716270-7716273 on Build GRCh38
rs766981224 Chr.12: 7716125-7716125 on Build GRCh38
rs780546746 Chr.12: 7716283-7716283 on Build GRCh38
rs762322563 Chr.12: 7716203-7716205 on Build GRCh38
rs1028615771 Chr.12: 7716104-7716104 on Build GRCh38
rs1027998924 Chr.12: 7716400-7716400 on Build GRCh38
rs775344971 Chr.12: 7716069-7716069 on Build GRCh38
rs750962852 Chr.12: 7716268-7716268 on Build GRCh38
rs74609065 Chr.12: 7716495-7716495 on Build GRCh38
rs745391704 Chr.12: 7716481-7716481 on Build GRCh38
rs116330721 Chr.12: 7716084-7716084 on Build GRCh38
rs751622847 Chr.12: 7716194-7716194 on Build GRCh38
rs948653886 Chr.12: 7716306-7716306 on Build GRCh38
rs374696025 Chr.12: 7716326-7716326 on Build GRCh38
rs1012208166 Chr.12: 7716472-7716472 on Build GRCh38
rs747223098 Chr.12: 7716179-7716179 on Build GRCh38
rs774045171 Chr.12: 7716209-7716209 on Build GRCh38
rs771204868 Chr.12: 7716166-7716166 on Build GRCh38
rs753690173 Chr.12: 7716073-7716073 on Build GRCh38
rs967309438 Chr.12: 7716466-7716466 on Build GRCh38
rs774375713 Chr.12: 7716179-7716180 on Build GRCh38
rs545582138 Chr.12: 7716325-7716325 on Build GRCh38
rs945455198 Chr.12: 7716162-7716162 on Build GRCh38
rs767040190 Chr.12: 7716223-7716223 on Build GRCh38
rs367876022 Chr.12: 7716248-7716248 on Build GRCh38
rs776739994 Chr.12: 7716181-7716181 on Build GRCh38
rs748136200 Chr.12: 7716192-7716192 on Build GRCh38
rs779257873 Chr.12: 7716277-7716278 on Build GRCh38
rs760050138 Chr.12: 7716257-7716257 on Build GRCh38
rs770705059 Chr.12: 7716201-7716201 on Build GRCh38
rs991701269 Chr.12: 7716170-7716170 on Build GRCh38
rs34547953 Chr.12: 7716269-7716270 on Build GRCh38
rs564926613 Chr.12: 7716362-7716362 on Build GRCh38
rs10772662 Chr.12: 7716442-7716442 on Build GRCh38
rs965066750 Chr.12: 7716080-7716080 on Build GRCh38
rs199813030 Chr.12: 7716087-7716090 on Build GRCh38
rs1007073155 Chr.12: 7716375-7716375 on Build GRCh38
rs562386853 Chr.12: 7716369-7716369 on Build GRCh38
rs997892847 Chr.12: 7716394-7716394 on Build GRCh38
rs752939876 Chr.12: 7716128-7716129 on Build GRCh38
rs770166497 Chr.12: 7716362-7716362 on Build GRCh38
rs779242521 Chr.12: 7716171-7716171 on Build GRCh38
rs201470889 Chr.12: 7716285-7716285 on Build GRCh38

View Full Product Details