Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 111644277-111644725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRAP
Entrez Gene ID
8315
Gene Name
BRCA1 associated protein
Gene Aliases
BRAP2, IMP, RNF52
UniGene
Hs.530940
Gene Chromosome Location
Chr.12: 111642146-111686023 on Build GRCh38
Transcripts
NM_006768.4 XM_005253944.4 XM_011538789.2 XM_017019992.1 XM_017019993.1 XM_017019994.1 XM_017019995.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4666003 c.1601G>A p.R534H Chr.12: 111644377-111644377 on Build GRCh38
6285282 c.1618C>A p.L540M Chr.12: 111644360-111644360 on Build GRCh38
1605574 c.1609A>T p.M537L Chr.12: 111644369-111644369 on Build GRCh38
7113602 c.1608C>T p.V536V Chr.12: 111644370-111644370 on Build GRCh38
4575007 c.1540G>T p.V514L Chr.12: 111644438-111644438 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765162109 Chr.12: 111644479-111644479 on Build GRCh38
rs575715091 Chr.12: 111644660-111644660 on Build GRCh38
rs1037490504 Chr.12: 111644624-111644624 on Build GRCh38
rs751241411 Chr.12: 111644397-111644397 on Build GRCh38
rs768788339 Chr.12: 111644566-111644566 on Build GRCh38
rs140662424 Chr.12: 111644441-111644441 on Build GRCh38
rs543311487 Chr.12: 111644684-111644684 on Build GRCh38
rs145317226 Chr.12: 111644317-111644317 on Build GRCh38
rs759792885 Chr.12: 111644323-111644323 on Build GRCh38
rs137950599 Chr.12: 111644396-111644396 on Build GRCh38
rs369273112 Chr.12: 111644515-111644515 on Build GRCh38
rs200375363 Chr.12: 111644373-111644373 on Build GRCh38
rs1041432035 Chr.12: 111644478-111644478 on Build GRCh38
rs986560522 Chr.12: 111644646-111644646 on Build GRCh38
rs190633865 Chr.12: 111644533-111644533 on Build GRCh38
rs747235004 Chr.12: 111644454-111644454 on Build GRCh38
rs773106574 Chr.12: 111644593-111644593 on Build GRCh38
rs577248230 Chr.12: 111644522-111644523 on Build GRCh38
rs776417310 Chr.12: 111644575-111644575 on Build GRCh38
rs776311293 Chr.12: 111644474-111644474 on Build GRCh38
rs185820400 Chr.12: 111644485-111644485 on Build GRCh38
rs765906865 Chr.12: 111644608-111644609 on Build GRCh38
rs201288855 Chr.12: 111644471-111644471 on Build GRCh38
rs772430535 Chr.12: 111644325-111644325 on Build GRCh38
rs767418635 Chr.12: 111644377-111644377 on Build GRCh38
rs77913600 Chr.12: 111644677-111644677 on Build GRCh38
rs752512186 Chr.12: 111644335-111644335 on Build GRCh38
rs376787096 Chr.12: 111644372-111644372 on Build GRCh38
rs775090047 Chr.12: 111644329-111644329 on Build GRCh38
rs753891708 Chr.12: 111644498-111644499 on Build GRCh38
rs139764163 Chr.12: 111644306-111644306 on Build GRCh38
rs751206358 Chr.12: 111644486-111644486 on Build GRCh38
rs371932680 Chr.12: 111644405-111644405 on Build GRCh38
rs1025704238 Chr.12: 111644681-111644681 on Build GRCh38
rs768473216 Chr.12: 111644472-111644472 on Build GRCh38
rs568588651 Chr.12: 111644494-111644494 on Build GRCh38
rs769632380 Chr.12: 111644586-111644586 on Build GRCh38
rs778050354 Chr.12: 111644425-111644425 on Build GRCh38
rs149777018 Chr.12: 111644497-111644497 on Build GRCh38
rs763862113 Chr.12: 111644338-111644338 on Build GRCh38
rs762432701 Chr.12: 111644483-111644483 on Build GRCh38
rs557541644 Chr.12: 111644622-111644622 on Build GRCh38
rs987478852 Chr.12: 111644339-111644339 on Build GRCh38
rs534723839 Chr.12: 111644452-111644452 on Build GRCh38
rs754158425 Chr.12: 111644500-111644500 on Build GRCh38
rs200653575 Chr.12: 111644298-111644298 on Build GRCh38
rs779550704 Chr.12: 111644517-111644517 on Build GRCh38
rs370119257 Chr.12: 111644493-111644493 on Build GRCh38
rs764340417 Chr.12: 111644327-111644357 on Build GRCh38
rs779071140 Chr.12: 111644438-111644438 on Build GRCh38
rs774597983 Chr.12: 111644318-111644318 on Build GRCh38
rs931997728 Chr.12: 111644618-111644618 on Build GRCh38
rs760506899 Chr.12: 111644330-111644330 on Build GRCh38
rs750838995 Chr.12: 111644701-111644701 on Build GRCh38
rs747947817 Chr.12: 111644580-111644580 on Build GRCh38
rs757974635 Chr.12: 111644378-111644378 on Build GRCh38
rs540162471 Chr.12: 111644662-111644662 on Build GRCh38
rs11541241 Chr.12: 111644513-111644513 on Build GRCh38
rs746444571 Chr.12: 111644523-111644523 on Build GRCh38
rs374914110 Chr.12: 111644444-111644444 on Build GRCh38
rs761696036 Chr.12: 111644475-111644475 on Build GRCh38
rs749756786 Chr.12: 111644426-111644426 on Build GRCh38
rs373778694 Chr.12: 111644341-111644341 on Build GRCh38
rs962768307 Chr.12: 111644299-111644299 on Build GRCh38
rs780046712 Chr.12: 111644536-111644536 on Build GRCh38
rs147643341 Chr.12: 111644433-111644433 on Build GRCh38
rs747205542 Chr.12: 111644539-111644539 on Build GRCh38
rs202001896 Chr.12: 111644481-111644481 on Build GRCh38
rs760873725 Chr.12: 111644362-111644362 on Build GRCh38
rs78238613 Chr.12: 111644689-111644689 on Build GRCh38

View Full Product Details