Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27444576-27445157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT172
Entrez Gene ID
26160
Gene Name
intraflagellar transport 172
Gene Aliases
BBS20, NPHP17, RP71, SLB, SRTD10, osm-1, wim
UniGene
Hs.127401
Gene Chromosome Location
Chr.2: 27444373-27489819 on Build GRCh38
Transcripts
NM_015662.2 XM_005264254.1 XM_006711986.3 XM_006711987.1 XM_011532757.2 XM_011532759.2 XM_011532760.2 XM_017003790.1 XM_017003791.1 XM_017003793.1 XM_017003794.1 XM_017003795.1 NM_001168364.1

Gene Symbol
KRTCAP3
Entrez Gene ID
200634
Gene Name
keratinocyte associated protein 3
Gene Aliases
KCP3
UniGene
Hs.59509
Gene Chromosome Location
Chr.2: 27442366-27446481 on Build GRCh38
Transcripts
NM_015662.2 XM_005264254.1 XM_006711986.3 XM_006711987.1 XM_011532757.2 XM_011532759.2 XM_011532760.2 XM_017003790.1 XM_017003791.1 XM_017003793.1 XM_017003794.1 XM_017003795.1 NM_001168364.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1631955 c.5104C>T p.R1702W Chr.2: 27445070-27445070 on Build GRCh38
2940073 c.5106G>A p.R1702R Chr.2: 27445068-27445068 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs777086910 Chr.2: 27445035-27445036 on Build GRCh38
rs867914878 Chr.2: 27445030-27445030 on Build GRCh38
rs1036436563 Chr.2: 27444710-27444710 on Build GRCh38
rs542662514 Chr.2: 27445046-27445046 on Build GRCh38
rs995056984 Chr.2: 27444740-27444740 on Build GRCh38
rs770527147 Chr.2: 27445101-27445102 on Build GRCh38
rs747606863 Chr.2: 27444764-27444764 on Build GRCh38
rs746289435 Chr.2: 27444837-27444837 on Build GRCh38
rs33989432 Chr.2: 27444654-27444655 on Build GRCh38
rs549230810 Chr.2: 27444700-27444700 on Build GRCh38
rs10635082 Chr.2: 27444653-27444654 on Build GRCh38
rs867122593 Chr.2: 27445029-27445029 on Build GRCh38
rs759323455 Chr.2: 27444975-27444975 on Build GRCh38
rs376789087 Chr.2: 27444808-27444808 on Build GRCh38
rs369150915 Chr.2: 27444998-27444998 on Build GRCh38
rs560831644 Chr.2: 27445070-27445070 on Build GRCh38
rs373421077 Chr.2: 27445125-27445125 on Build GRCh38
rs373184496 Chr.2: 27445031-27445031 on Build GRCh38
rs869025254 Chr.2: 27445041-27445041 on Build GRCh38
rs770335799 Chr.2: 27445037-27445037 on Build GRCh38
rs190642930 Chr.2: 27444756-27444756 on Build GRCh38
rs773957784 Chr.2: 27445105-27445110 on Build GRCh38
rs986859612 Chr.2: 27444952-27444955 on Build GRCh38
rs986806380 Chr.2: 27444668-27444668 on Build GRCh38
rs965724751 Chr.2: 27444795-27444795 on Build GRCh38
rs867814195 Chr.2: 27444832-27444832 on Build GRCh38
rs868748622 Chr.2: 27445050-27445050 on Build GRCh38
rs201572823 Chr.2: 27445131-27445131 on Build GRCh38
rs954176890 Chr.2: 27444952-27444952 on Build GRCh38
rs749020649 Chr.2: 27445020-27445020 on Build GRCh38
rs369598169 Chr.2: 27444813-27444813 on Build GRCh38
rs778684568 Chr.2: 27444729-27444729 on Build GRCh38
rs755256585 Chr.2: 27445015-27445015 on Build GRCh38
rs367679647 Chr.2: 27444989-27444989 on Build GRCh38
rs181461008 Chr.2: 27444980-27444980 on Build GRCh38
rs111756175 Chr.2: 27444951-27444952 on Build GRCh38
rs751676417 Chr.2: 27445130-27445130 on Build GRCh38
rs1021041354 Chr.2: 27445100-27445100 on Build GRCh38
rs950089572 Chr.2: 27444935-27444935 on Build GRCh38
rs754296042 Chr.2: 27444982-27444982 on Build GRCh38
rs375081229 Chr.2: 27445058-27445058 on Build GRCh38
rs912560582 Chr.2: 27444755-27444755 on Build GRCh38
rs566835533 Chr.2: 27444653-27444653 on Build GRCh38
rs774162236 Chr.2: 27444973-27444973 on Build GRCh38
rs766439327 Chr.2: 27445127-27445127 on Build GRCh38
rs185432065 Chr.2: 27444687-27444687 on Build GRCh38
rs894942839 Chr.2: 27444799-27444799 on Build GRCh38
rs772753536 Chr.2: 27444968-27444968 on Build GRCh38
rs373153625 Chr.2: 27444655-27444656 on Build GRCh38
rs554311210 Chr.2: 27444849-27444849 on Build GRCh38
rs1028449352 Chr.2: 27444852-27444852 on Build GRCh38
rs751766272 Chr.2: 27445014-27445014 on Build GRCh38
rs917237367 Chr.2: 27444891-27444891 on Build GRCh38
rs937930866 Chr.2: 27444646-27444646 on Build GRCh38
rs889302546 Chr.2: 27444999-27444999 on Build GRCh38
rs765709301 Chr.2: 27445094-27445094 on Build GRCh38
rs1055686607 Chr.2: 27444682-27444682 on Build GRCh38
rs868208741 Chr.2: 27445047-27445047 on Build GRCh38
rs868318014 Chr.2: 27444937-27444937 on Build GRCh38
rs774073874 Chr.2: 27445042-27445042 on Build GRCh38
rs748778867 Chr.2: 27445036-27445036 on Build GRCh38
rs553900972 Chr.2: 27444768-27444768 on Build GRCh38
rs748959424 Chr.2: 27445051-27445052 on Build GRCh38
rs754565379 Chr.2: 27444713-27444713 on Build GRCh38
rs370108416 Chr.2: 27445108-27445108 on Build GRCh38
rs766529492 Chr.2: 27445004-27445004 on Build GRCh38
rs556885484 Chr.2: 27444796-27444796 on Build GRCh38
rs865894977 Chr.2: 27444809-27444809 on Build GRCh38
rs565409936 Chr.2: 27444836-27444836 on Build GRCh38
rs895832270 Chr.2: 27444686-27444686 on Build GRCh38
rs992062716 Chr.2: 27444885-27444885 on Build GRCh38
rs527554115 Chr.2: 27444833-27444833 on Build GRCh38
rs769890818 Chr.2: 27444840-27444840 on Build GRCh38
rs750474739 Chr.2: 27444996-27444996 on Build GRCh38
rs1037374820 Chr.2: 27444757-27444757 on Build GRCh38
rs747468179 Chr.2: 27444773-27444773 on Build GRCh38
rs375966926 Chr.2: 27444986-27444986 on Build GRCh38
rs777579270 Chr.2: 27445032-27445032 on Build GRCh38
rs762114339 Chr.2: 27445081-27445081 on Build GRCh38
rs762404536 Chr.2: 27444969-27444969 on Build GRCh38
rs775719395 Chr.2: 27445069-27445069 on Build GRCh38
rs747938088 Chr.2: 27445027-27445027 on Build GRCh38
rs56750963 Chr.2: 27444664-27444665 on Build GRCh38
rs902198037 Chr.2: 27444829-27444829 on Build GRCh38
rs1032366724 Chr.2: 27444833-27444833 on Build GRCh38
rs1028242278 Chr.2: 27444751-27444751 on Build GRCh38
rs931169377 Chr.2: 27444790-27444790 on Build GRCh38
rs770554830 Chr.2: 27444970-27444970 on Build GRCh38
rs937899523 Chr.2: 27444861-27444861 on Build GRCh38
rs146309780 Chr.2: 27445025-27445025 on Build GRCh38

View Full Product Details