Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 210024863-210025204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KANSL1L
Entrez Gene ID
151050
Gene Name
KAT8 regulatory NSL complex subunit 1 like
Gene Aliases
C2orf67, MSL1v2
UniGene
Hs.282260
Gene Chromosome Location
Chr.2: 210021421-210171370 on Build GRCh38
Transcripts
NM_001307976.1 NM_152519.3 XM_005246328.4 XM_005246329.4 XM_005246330.4 XM_005246332.4 XM_011510706.2 XM_011510707.2 XM_011510710.2 XM_017003430.1 XM_017003431.1 XM_006712677.3

Gene Symbol
RPE
Entrez Gene ID
6120
Gene Name
ribulose-5-phosphate-3-epimerase
Gene Aliases
RPE2-1
UniGene
Hs.282260
Gene Chromosome Location
Chr.2: 210002565-210029156 on Build GRCh38
Transcripts
NM_001307976.1 NM_152519.3 XM_005246328.4 XM_005246329.4 XM_005246330.4 XM_005246332.4 XM_011510706.2 XM_011510707.2 XM_011510710.2 XM_017003430.1 XM_017003431.1 XM_006712677.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2715456 c.2470G>A p.E824K Chr.2: 210025198-210025198 on Build GRCh38
1735876 c.2508_2511delAGAG p.R836fs*32 Chr.2: 210025157-210025160 on Build GRCh38
1136586 c.2564+2C>T p.? Chr.2: 210025102-210025102 on Build GRCh38
6010625 c.2533G>A p.E845K Chr.2: 210025135-210025135 on Build GRCh38
476845 c.2531G>A p.W844* Chr.2: 210025137-210025137 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs990210970 Chr.2: 210025050-210025050 on Build GRCh38
rs373097635 Chr.2: 210025112-210025112 on Build GRCh38
rs1021725883 Chr.2: 210025173-210025173 on Build GRCh38
rs747995867 Chr.2: 210025159-210025159 on Build GRCh38
rs6435550 Chr.2: 210025035-210025035 on Build GRCh38
rs753389048 Chr.2: 210025081-210025081 on Build GRCh38
rs758906097 Chr.2: 210025109-210025109 on Build GRCh38
rs781365273 Chr.2: 210025113-210025113 on Build GRCh38
rs765522279 Chr.2: 210025078-210025079 on Build GRCh38
rs757545327 Chr.2: 210025069-210025070 on Build GRCh38
rs772970759 Chr.2: 210025169-210025169 on Build GRCh38
rs772108844 Chr.2: 210025167-210025167 on Build GRCh38
rs913289212 Chr.2: 210025072-210025072 on Build GRCh38
rs191091412 Chr.2: 210025007-210025007 on Build GRCh38
rs985898407 Chr.2: 210024935-210024935 on Build GRCh38
rs753122006 Chr.2: 210025089-210025089 on Build GRCh38
rs909086790 Chr.2: 210024884-210024884 on Build GRCh38
rs765530721 Chr.2: 210025085-210025085 on Build GRCh38
rs149071192 Chr.2: 210024988-210024988 on Build GRCh38
rs559885185 Chr.2: 210025044-210025044 on Build GRCh38
rs943215418 Chr.2: 210024900-210024900 on Build GRCh38
rs770197627 Chr.2: 210025125-210025125 on Build GRCh38
rs933303499 Chr.2: 210025042-210025042 on Build GRCh38
rs754666202 Chr.2: 210025083-210025083 on Build GRCh38
rs1042207205 Chr.2: 210025162-210025162 on Build GRCh38
rs760148431 Chr.2: 210025065-210025065 on Build GRCh38
rs767397210 Chr.2: 210025008-210025008 on Build GRCh38
rs544200391 Chr.2: 210024929-210024929 on Build GRCh38
rs1011695284 Chr.2: 210025140-210025140 on Build GRCh38
rs778292831 Chr.2: 210025110-210025110 on Build GRCh38
rs1053146465 Chr.2: 210025062-210025062 on Build GRCh38
rs746016798 Chr.2: 210025116-210025116 on Build GRCh38
rs765933021 Chr.2: 210025074-210025074 on Build GRCh38
rs778846240 Chr.2: 210025139-210025139 on Build GRCh38

View Full Product Details