Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 189661185-189661732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASNSD1
Entrez Gene ID
54529
Gene Name
asparagine synthetase domain containing 1
Gene Aliases
NBLA00058, NS3TP1
UniGene
Hs.101364
Gene Chromosome Location
Chr.2: 189661399-189670831 on Build GRCh38
Transcripts
NM_019048.2 XM_017004382.1

SNP Detail

SNP ID Chromosome Location
rs1038949323 Chr.2: 189661470-189661470 on Build GRCh38
rs879533483 Chr.2: 189661402-189661402 on Build GRCh38
rs937398048 Chr.2: 189661302-189661302 on Build GRCh38
rs917257973 Chr.2: 189661433-189661433 on Build GRCh38
rs144135941 Chr.2: 189661306-189661307 on Build GRCh38
rs891434273 Chr.2: 189661425-189661425 on Build GRCh38
rs979487913 Chr.2: 189661308-189661308 on Build GRCh38
rs780363460 Chr.2: 189661651-189661651 on Build GRCh38
rs754742570 Chr.2: 189661515-189661515 on Build GRCh38
rs943801365 Chr.2: 189661551-189661551 on Build GRCh38
rs529004683 Chr.2: 189661430-189661430 on Build GRCh38
rs760100597 Chr.2: 189661480-189661480 on Build GRCh38
rs566374570 Chr.2: 189661259-189661259 on Build GRCh38
rs971408344 Chr.2: 189661534-189661534 on Build GRCh38
rs74593114 Chr.2: 189661555-189661555 on Build GRCh38
rs369157040 Chr.2: 189661662-189661662 on Build GRCh38
rs144769459 Chr.2: 189661477-189661477 on Build GRCh38
rs890146390 Chr.2: 189661488-189661488 on Build GRCh38
rs772512220 Chr.2: 189661492-189661492 on Build GRCh38
rs557646694 Chr.2: 189661345-189661345 on Build GRCh38
rs16831841 Chr.2: 189661397-189661397 on Build GRCh38
rs921454311 Chr.2: 189661638-189661638 on Build GRCh38
rs887303928 Chr.2: 189661350-189661350 on Build GRCh38
rs746694354 Chr.2: 189661277-189661277 on Build GRCh38
rs756327256 Chr.2: 189661469-189661469 on Build GRCh38
rs1041412887 Chr.2: 189661672-189661673 on Build GRCh38
rs141049165 Chr.2: 189661463-189661463 on Build GRCh38
rs1004579416 Chr.2: 189661360-189661360 on Build GRCh38
rs1008812440 Chr.2: 189661493-189661493 on Build GRCh38
rs1019235385 Chr.2: 189661242-189661242 on Build GRCh38
rs746653270 Chr.2: 189661601-189661601 on Build GRCh38
rs146511798 Chr.2: 189661416-189661416 on Build GRCh38
rs1008302551 Chr.2: 189661233-189661233 on Build GRCh38
rs533235636 Chr.2: 189661458-189661458 on Build GRCh38
rs367762939 Chr.2: 189661307-189661308 on Build GRCh38
rs533932245 Chr.2: 189661624-189661624 on Build GRCh38
rs931735997 Chr.2: 189661646-189661646 on Build GRCh38
rs533872599 Chr.2: 189661295-189661295 on Build GRCh38
rs558205983 Chr.2: 189661606-189661606 on Build GRCh38
rs1033274077 Chr.2: 189661506-189661506 on Build GRCh38
rs765883315 Chr.2: 189661342-189661342 on Build GRCh38
rs889574753 Chr.2: 189661522-189661522 on Build GRCh38
rs992008348 Chr.2: 189661523-189661523 on Build GRCh38
rs968193724 Chr.2: 189661448-189661448 on Build GRCh38
rs988356982 Chr.2: 189661255-189661255 on Build GRCh38
rs373323365 Chr.2: 189661497-189661497 on Build GRCh38
rs1029088363 Chr.2: 189661368-189661368 on Build GRCh38
rs1033795857 Chr.2: 189661647-189661647 on Build GRCh38
rs931101164 Chr.2: 189661388-189661388 on Build GRCh38
rs112098534 Chr.2: 189661466-189661466 on Build GRCh38
rs889464651 Chr.2: 189661224-189661224 on Build GRCh38
rs772923573 Chr.2: 189661565-189661565 on Build GRCh38
rs1132360 Chr.2: 189661498-189661498 on Build GRCh38
rs557314889 Chr.2: 189661693-189661693 on Build GRCh38
rs776296653 Chr.2: 189661636-189661636 on Build GRCh38
rs1014104874 Chr.2: 189661653-189661653 on Build GRCh38
rs573188813 Chr.2: 189661407-189661407 on Build GRCh38
rs535045512 Chr.2: 189661330-189661330 on Build GRCh38
rs1016004294 Chr.2: 189661363-189661363 on Build GRCh38
rs904108766 Chr.2: 189661603-189661603 on Build GRCh38
rs938057593 Chr.2: 189661314-189661314 on Build GRCh38
rs187165993 Chr.2: 189661607-189661607 on Build GRCh38
rs1021093033 Chr.2: 189661302-189661302 on Build GRCh38
rs536059762 Chr.2: 189661337-189661337 on Build GRCh38
rs992200259 Chr.2: 189661474-189661474 on Build GRCh38
rs189592653 Chr.2: 189661431-189661431 on Build GRCh38
rs1049572400 Chr.2: 189661519-189661519 on Build GRCh38
rs540711302 Chr.2: 189661415-189661415 on Build GRCh38
rs1050074916 Chr.2: 189661483-189661483 on Build GRCh38
rs959702632 Chr.2: 189661471-189661471 on Build GRCh38
rs931174371 Chr.2: 189661503-189661503 on Build GRCh38
rs562417196 Chr.2: 189661435-189661435 on Build GRCh38
rs926673512 Chr.2: 189661310-189661310 on Build GRCh38
rs2289226 Chr.2: 189661479-189661479 on Build GRCh38
rs539310673 Chr.2: 189661468-189661468 on Build GRCh38
rs573642262 Chr.2: 189661708-189661708 on Build GRCh38
rs917864361 Chr.2: 189661334-189661334 on Build GRCh38
rs1042651064 Chr.2: 189661602-189661602 on Build GRCh38
rs771780567 Chr.2: 189661219-189661219 on Build GRCh38
rs772908307 Chr.2: 189661276-189661276 on Build GRCh38
rs987409288 Chr.2: 189661392-189661392 on Build GRCh38
rs558108768 Chr.2: 189661398-189661398 on Build GRCh38
rs117249934 Chr.2: 189661702-189661702 on Build GRCh38
rs555542782 Chr.2: 189661304-189661304 on Build GRCh38
rs925939028 Chr.2: 189661292-189661292 on Build GRCh38
rs191549755 Chr.2: 189661655-189661655 on Build GRCh38
rs183313673 Chr.2: 189661690-189661690 on Build GRCh38
rs961710943 Chr.2: 189661676-189661676 on Build GRCh38
rs369425866 Chr.2: 189661487-189661487 on Build GRCh38
rs568668677 Chr.2: 189661319-189661319 on Build GRCh38
rs967239007 Chr.2: 189661405-189661405 on Build GRCh38
rs1047195820 Chr.2: 189661324-189661324 on Build GRCh38
rs995267421 Chr.2: 189661208-189661208 on Build GRCh38
rs573820422 Chr.2: 189661657-189661657 on Build GRCh38
rs1021124372 Chr.2: 189661447-189661447 on Build GRCh38
rs992562993 Chr.2: 189661316-189661316 on Build GRCh38
rs536023125 Chr.2: 189661501-189661501 on Build GRCh38
rs968164599 Chr.2: 189661303-189661303 on Build GRCh38
rs966677366 Chr.2: 189661243-189661243 on Build GRCh38
rs1047333876 Chr.2: 189661457-189661457 on Build GRCh38
rs776168579 Chr.2: 189661361-189661361 on Build GRCh38
rs186477034 Chr.2: 189661364-189661364 on Build GRCh38
rs575750733 Chr.2: 189661494-189661494 on Build GRCh38
rs1008529313 Chr.2: 189661660-189661660 on Build GRCh38
rs181844257 Chr.2: 189661560-189661560 on Build GRCh38
rs1028038290 Chr.2: 189661210-189661210 on Build GRCh38
rs540108534 Chr.2: 189661583-189661583 on Build GRCh38

View Full Product Details