Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 28769387-28769973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXG1
Entrez Gene ID
2290
Gene Name
forkhead box G1
Gene Aliases
BF1, BF2, FHKL3, FKH2, FKHL1, FKHL2, FKHL3, FKHL4, FOXG1A, FOXG1B, FOXG1C, HBF-1, HBF-2, HBF-3, HBF-G2, HBF2, HFK1, HFK2, HFK3, KHL2, QIN
UniGene
Hs.632336
Gene Chromosome Location
Chr.14: 28767072-28770277 on Build GRCh38
Transcripts
NM_005249.4

SNP Detail

SNP ID Chromosome Location
rs1042766 Chr.14: 28769642-28769642 on Build GRCh38
rs962160702 Chr.14: 28769412-28769412 on Build GRCh38
rs936053139 Chr.14: 28769439-28769439 on Build GRCh38
rs1028203328 Chr.14: 28769492-28769492 on Build GRCh38
rs979231296 Chr.14: 28769880-28769880 on Build GRCh38
rs992039706 Chr.14: 28769850-28769850 on Build GRCh38
rs535183687 Chr.14: 28769639-28769639 on Build GRCh38
rs1050837682 Chr.14: 28769503-28769503 on Build GRCh38
rs904596301 Chr.14: 28769423-28769423 on Build GRCh38
rs980487543 Chr.14: 28769687-28769687 on Build GRCh38
rs866078101 Chr.14: 28769935-28769935 on Build GRCh38
rs926082087 Chr.14: 28769923-28769923 on Build GRCh38
rs796158610 Chr.14: 28769938-28769938 on Build GRCh38
rs951884710 Chr.14: 28769673-28769673 on Build GRCh38
rs993632294 Chr.14: 28769609-28769609 on Build GRCh38
rs554445894 Chr.14: 28769586-28769586 on Build GRCh38
rs775044607 Chr.14: 28769735-28769735 on Build GRCh38
rs540893672 Chr.14: 28769483-28769483 on Build GRCh38
rs913785249 Chr.14: 28769851-28769851 on Build GRCh38
rs889402864 Chr.14: 28769509-28769509 on Build GRCh38
rs960378930 Chr.14: 28769829-28769829 on Build GRCh38
rs1027440660 Chr.14: 28769653-28769653 on Build GRCh38
rs1033775745 Chr.14: 28769773-28769773 on Build GRCh38
rs189418563 Chr.14: 28769558-28769558 on Build GRCh38
rs1054223172 Chr.14: 28769812-28769812 on Build GRCh38
rs372395431 Chr.14: 28769860-28769860 on Build GRCh38
rs986766231 Chr.14: 28769548-28769548 on Build GRCh38
rs924280928 Chr.14: 28769714-28769714 on Build GRCh38
rs934374901 Chr.14: 28769723-28769723 on Build GRCh38
rs952260281 Chr.14: 28769526-28769526 on Build GRCh38
rs181934041 Chr.14: 28769918-28769918 on Build GRCh38
rs914366189 Chr.14: 28769822-28769822 on Build GRCh38
rs898000834 Chr.14: 28769603-28769603 on Build GRCh38
rs975756000 Chr.14: 28769602-28769602 on Build GRCh38
rs567312195 Chr.14: 28769865-28769865 on Build GRCh38
rs948604173 Chr.14: 28769858-28769858 on Build GRCh38
rs936070181 Chr.14: 28769934-28769934 on Build GRCh38

View Full Product Details