Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120978412-120979000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNF1A
Entrez Gene ID
6927
Gene Name
HNF1 homeobox A
Gene Aliases
HNF-1A, HNF1, IDDM20, LFB1, MODY3, TCF-1, TCF1
UniGene
Hs.654455
Gene Chromosome Location
Chr.12: 120978515-121002512 on Build GRCh38
Transcripts
NM_000545.6 NM_001306179.1 XM_005253931.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3457088 c.226G>A p.D76N Chr.12: 120978994-120978994 on Build GRCh38
46447 c.97_103delCCGGGGC p.G34fs*119 Chr.12: 120978865-120978871 on Build GRCh38
4526942 c.142G>A p.E48K Chr.12: 120978910-120978910 on Build GRCh38
3457086 c.138G>A p.K46K Chr.12: 120978906-120978906 on Build GRCh38
6909813 c.92G>A p.G31D Chr.12: 120978860-120978860 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1039044797 Chr.12: 120978742-120978765 on Build GRCh38
rs868538299 Chr.12: 120978698-120978698 on Build GRCh38
rs909308709 Chr.12: 120978587-120978587 on Build GRCh38
rs576862555 Chr.12: 120978673-120978673 on Build GRCh38
rs961902902 Chr.12: 120978449-120978449 on Build GRCh38
rs748532681 Chr.12: 120978721-120978721 on Build GRCh38
rs781754738 Chr.12: 120978715-120978715 on Build GRCh38
rs886043942 Chr.12: 120978615-120978616 on Build GRCh38
rs766956862 Chr.12: 120978928-120978928 on Build GRCh38
rs757926841 Chr.12: 120978758-120978758 on Build GRCh38
rs1026580886 Chr.12: 120978726-120978726 on Build GRCh38
rs769666514 Chr.12: 120978922-120978922 on Build GRCh38
rs750801469 Chr.12: 120978972-120978972 on Build GRCh38
rs960859520 Chr.12: 120978448-120978448 on Build GRCh38
rs543092439 Chr.12: 120978596-120978596 on Build GRCh38
rs899708954 Chr.12: 120978644-120978644 on Build GRCh38
rs749730441 Chr.12: 120978895-120978895 on Build GRCh38
rs749624286 Chr.12: 120978724-120978724 on Build GRCh38
rs772214801 Chr.12: 120978809-120978809 on Build GRCh38
rs764174318 Chr.12: 120978834-120978834 on Build GRCh38
rs193922588 Chr.12: 120978937-120978937 on Build GRCh38
rs759730121 Chr.12: 120978739-120978739 on Build GRCh38
rs748882858 Chr.12: 120978600-120978600 on Build GRCh38
rs763117095 Chr.12: 120978671-120978671 on Build GRCh38
rs376961226 Chr.12: 120978740-120978740 on Build GRCh38
rs776710848 Chr.12: 120978920-120978920 on Build GRCh38
rs1169289 Chr.12: 120978819-120978819 on Build GRCh38
rs761779651 Chr.12: 120978921-120978921 on Build GRCh38
rs973336454 Chr.12: 120978560-120978560 on Build GRCh38
rs780586155 Chr.12: 120978644-120978644 on Build GRCh38
rs754470733 Chr.12: 120978643-120978644 on Build GRCh38
rs193922578 Chr.12: 120978898-120978898 on Build GRCh38
rs193922592 Chr.12: 120978769-120978769 on Build GRCh38
rs587778393 Chr.12: 120978923-120978924 on Build GRCh38
rs985167109 Chr.12: 120978564-120978564 on Build GRCh38
rs1038779132 Chr.12: 120978703-120978703 on Build GRCh38
rs1024131753 Chr.12: 120978551-120978551 on Build GRCh38
rs142318174 Chr.12: 120978923-120978923 on Build GRCh38
rs779442858 Chr.12: 120978970-120978970 on Build GRCh38
rs773528592 Chr.12: 120978722-120978722 on Build GRCh38
rs970766228 Chr.12: 120978582-120978582 on Build GRCh38
rs759514603 Chr.12: 120978679-120978679 on Build GRCh38
rs777653568 Chr.12: 120978706-120978706 on Build GRCh38
rs768849887 Chr.12: 120978919-120978919 on Build GRCh38
rs767550584 Chr.12: 120978680-120978680 on Build GRCh38
rs574473281 Chr.12: 120978593-120978593 on Build GRCh38
rs561269721 Chr.12: 120978765-120978765 on Build GRCh38
rs1028023355 Chr.12: 120978601-120978601 on Build GRCh38
rs749712370 Chr.12: 120978768-120978768 on Build GRCh38
rs770046425 Chr.12: 120978651-120978651 on Build GRCh38
rs370979090 Chr.12: 120978764-120978764 on Build GRCh38
rs554400432 Chr.12: 120978581-120978581 on Build GRCh38
rs1047425321 Chr.12: 120978714-120978714 on Build GRCh38
rs146686581 Chr.12: 120978978-120978978 on Build GRCh38
rs188085301 Chr.12: 120978971-120978971 on Build GRCh38
rs755941887 Chr.12: 120978688-120978688 on Build GRCh38
rs371945966 Chr.12: 120978650-120978650 on Build GRCh38
rs545740321 Chr.12: 120978710-120978710 on Build GRCh38
rs565546289 Chr.12: 120978729-120978729 on Build GRCh38
rs772222326 Chr.12: 120978910-120978910 on Build GRCh38
rs757721117 Chr.12: 120978961-120978961 on Build GRCh38
rs773440493 Chr.12: 120978652-120978652 on Build GRCh38
rs770577871 Chr.12: 120978641-120978641 on Build GRCh38
rs755029520 Chr.12: 120978497-120978497 on Build GRCh38
rs903548333 Chr.12: 120978865-120978865 on Build GRCh38
rs377129682 Chr.12: 120978953-120978953 on Build GRCh38
rs747774978 Chr.12: 120978704-120978706 on Build GRCh38
rs587778394 Chr.12: 120978841-120978841 on Build GRCh38
rs775391034 Chr.12: 120978810-120978810 on Build GRCh38
rs538476099 Chr.12: 120978597-120978598 on Build GRCh38
rs776694486 Chr.12: 120978836-120978836 on Build GRCh38
rs769332225 Chr.12: 120978710-120978714 on Build GRCh38
rs756136537 Chr.12: 120978511-120978511 on Build GRCh38
rs781434823 Chr.12: 120978522-120978522 on Build GRCh38
rs1033416844 Chr.12: 120978709-120978709 on Build GRCh38
rs764010995 Chr.12: 120978745-120978745 on Build GRCh38
rs778233597 Chr.12: 120978767-120978767 on Build GRCh38
rs752573339 Chr.12: 120978687-120978687 on Build GRCh38
rs980850048 Chr.12: 120978588-120978588 on Build GRCh38
rs753567412 Chr.12: 120978707-120978707 on Build GRCh38
rs193922583 Chr.12: 120978918-120978918 on Build GRCh38
rs758774357 Chr.12: 120978974-120978974 on Build GRCh38
rs150195625 Chr.12: 120978924-120978924 on Build GRCh38
rs747247486 Chr.12: 120978945-120978945 on Build GRCh38
rs373180062 Chr.12: 120978907-120978907 on Build GRCh38
rs751537080 Chr.12: 120978610-120978616 on Build GRCh38
rs779943097 Chr.12: 120978608-120978608 on Build GRCh38
rs779098731 Chr.12: 120978900-120978900 on Build GRCh38
rs563304627 Chr.12: 120978645-120978645 on Build GRCh38
rs750035345 Chr.12: 120978755-120978755 on Build GRCh38
rs757633119 Chr.12: 120978899-120978899 on Build GRCh38
rs775511041 Chr.12: 120978744-120978744 on Build GRCh38
rs35126805 Chr.12: 120978484-120978484 on Build GRCh38
rs765037128 Chr.12: 120978747-120978747 on Build GRCh38
rs778321377 Chr.12: 120978894-120978894 on Build GRCh38
rs1001854457 Chr.12: 120978866-120978866 on Build GRCh38
rs774637975 Chr.12: 120978797-120978797 on Build GRCh38
rs1169288 Chr.12: 120978847-120978847 on Build GRCh38
rs761764877 Chr.12: 120978844-120978844 on Build GRCh38
rs199945468 Chr.12: 120978746-120978746 on Build GRCh38
rs538619966 Chr.12: 120978867-120978867 on Build GRCh38
rs929341467 Chr.12: 120978550-120978550 on Build GRCh38
rs779387337 Chr.12: 120978763-120978763 on Build GRCh38
rs188016232 Chr.12: 120978782-120978782 on Build GRCh38
rs771466066 Chr.12: 120978712-120978712 on Build GRCh38
rs1026610844 Chr.12: 120978509-120978509 on Build GRCh38
rs746020457 Chr.12: 120978798-120978798 on Build GRCh38
rs1022343054 Chr.12: 120978684-120978684 on Build GRCh38
rs950999882 Chr.12: 120978733-120978733 on Build GRCh38
rs751680667 Chr.12: 120978677-120978677 on Build GRCh38
rs779835903 Chr.12: 120978512-120978512 on Build GRCh38
rs749149857 Chr.12: 120978813-120978813 on Build GRCh38
rs145314420 Chr.12: 120978870-120978870 on Build GRCh38
rs776978662 Chr.12: 120978646-120978646 on Build GRCh38
rs994531774 Chr.12: 120978501-120978501 on Build GRCh38
rs139742280 Chr.12: 120978868-120978868 on Build GRCh38
rs137853247 Chr.12: 120978860-120978860 on Build GRCh38
rs926722426 Chr.12: 120978614-120978614 on Build GRCh38
rs780298807 Chr.12: 120978914-120978914 on Build GRCh38
rs534474388 Chr.12: 120978478-120978478 on Build GRCh38
rs759053189 Chr.12: 120978925-120978925 on Build GRCh38
rs774545180 Chr.12: 120978738-120978738 on Build GRCh38
rs778674831 Chr.12: 120978708-120978708 on Build GRCh38
rs750099601 Chr.12: 120978849-120978849 on Build GRCh38

View Full Product Details