Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 68023154-68023702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX28
Entrez Gene ID
55794
Gene Name
DEAD-box helicase 28
Gene Aliases
MDDX28
UniGene
Hs.458313
Gene Chromosome Location
Chr.16: 68021274-68023867 on Build GRCh38
Transcripts
NM_018380.3 NM_001271762.1 NM_001271763.1 NM_017803.4

Gene Symbol
DUS2
Entrez Gene ID
54920
Gene Name
dihydrouridine synthase 2
Gene Aliases
DUS2L, SMM1, URLC8
UniGene
Hs.534460
Gene Chromosome Location
Chr.16: 68022944-68079323 on Build GRCh38
Transcripts
NM_018380.3 NM_001271762.1 NM_001271763.1 NM_017803.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1379056 c.3G>A p.M1I Chr.16: 68023200-68023200 on Build GRCh38
4832588 c.36C>G p.L12L Chr.16: 68023167-68023167 on Build GRCh38
7163569 c.27C>T p.L9L Chr.16: 68023176-68023176 on Build GRCh38
4845633 c.27C>G p.L9L Chr.16: 68023176-68023176 on Build GRCh38
3667886 c.19G>T p.V7L Chr.16: 68023184-68023184 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567612030 Chr.16: 68023556-68023556 on Build GRCh38
rs1044545517 Chr.16: 68023372-68023372 on Build GRCh38
rs887719684 Chr.16: 68023469-68023469 on Build GRCh38
rs914659667 Chr.16: 68023260-68023260 on Build GRCh38
rs551467589 Chr.16: 68023298-68023298 on Build GRCh38
rs563430626 Chr.16: 68023320-68023320 on Build GRCh38
rs756620466 Chr.16: 68023219-68023219 on Build GRCh38
rs567983097 Chr.16: 68023499-68023499 on Build GRCh38
rs779893323 Chr.16: 68023534-68023534 on Build GRCh38
rs751265571 Chr.16: 68023213-68023213 on Build GRCh38
rs752435052 Chr.16: 68023209-68023209 on Build GRCh38
rs912478977 Chr.16: 68023676-68023676 on Build GRCh38
rs984502183 Chr.16: 68023237-68023237 on Build GRCh38
rs986322968 Chr.16: 68023606-68023606 on Build GRCh38
rs777366429 Chr.16: 68023212-68023212 on Build GRCh38
rs986397908 Chr.16: 68023281-68023281 on Build GRCh38
rs774679672 Chr.16: 68023246-68023246 on Build GRCh38
rs76902372 Chr.16: 68023255-68023255 on Build GRCh38
rs912197546 Chr.16: 68023329-68023330 on Build GRCh38
rs757982492 Chr.16: 68023178-68023178 on Build GRCh38
rs372302713 Chr.16: 68023419-68023419 on Build GRCh38
rs1030059668 Chr.16: 68023572-68023572 on Build GRCh38
rs559505232 Chr.16: 68023258-68023258 on Build GRCh38
rs748448080 Chr.16: 68023247-68023247 on Build GRCh38
rs1050631435 Chr.16: 68023191-68023191 on Build GRCh38
rs781521203 Chr.16: 68023186-68023186 on Build GRCh38
rs141515984 Chr.16: 68023204-68023204 on Build GRCh38
rs953973293 Chr.16: 68023613-68023613 on Build GRCh38
rs979276395 Chr.16: 68023302-68023302 on Build GRCh38
rs1001294787 Chr.16: 68023348-68023348 on Build GRCh38
rs1034206482 Chr.16: 68023228-68023228 on Build GRCh38
rs939684432 Chr.16: 68023345-68023345 on Build GRCh38
rs770507825 Chr.16: 68023185-68023185 on Build GRCh38
rs1006547812 Chr.16: 68023470-68023470 on Build GRCh38
rs891841618 Chr.16: 68023494-68023494 on Build GRCh38
rs936891491 Chr.16: 68023292-68023292 on Build GRCh38
rs758213519 Chr.16: 68023211-68023211 on Build GRCh38
rs749618467 Chr.16: 68023233-68023233 on Build GRCh38
rs996089447 Chr.16: 68023367-68023367 on Build GRCh38
rs237831 Chr.16: 68023193-68023193 on Build GRCh38
rs921194144 Chr.16: 68023307-68023307 on Build GRCh38
rs1052487432 Chr.16: 68023295-68023295 on Build GRCh38
rs755929753 Chr.16: 68023477-68023477 on Build GRCh38
rs747119709 Chr.16: 68023206-68023209 on Build GRCh38
rs752347194 Chr.16: 68023177-68023177 on Build GRCh38
rs769171202 Chr.16: 68023234-68023234 on Build GRCh38
rs140657263 Chr.16: 68023329-68023329 on Build GRCh38
rs150933726 Chr.16: 68023266-68023266 on Build GRCh38
rs1036783156 Chr.16: 68023351-68023351 on Build GRCh38
rs746436723 Chr.16: 68023187-68023187 on Build GRCh38
rs898229095 Chr.16: 68023359-68023359 on Build GRCh38
rs533333487 Chr.16: 68023287-68023287 on Build GRCh38
rs183094375 Chr.16: 68023649-68023649 on Build GRCh38
rs547809178 Chr.16: 68023297-68023297 on Build GRCh38
rs746456268 Chr.16: 68023182-68023182 on Build GRCh38
rs764247926 Chr.16: 68023252-68023252 on Build GRCh38
rs780811590 Chr.16: 68023214-68023214 on Build GRCh38
rs562009202 Chr.16: 68023190-68023190 on Build GRCh38
rs375506962 Chr.16: 68023544-68023544 on Build GRCh38
rs73596400 Chr.16: 68023539-68023539 on Build GRCh38
rs923429737 Chr.16: 68023174-68023174 on Build GRCh38
rs148388647 Chr.16: 68023201-68023201 on Build GRCh38
rs1028119432 Chr.16: 68023611-68023611 on Build GRCh38
rs949505016 Chr.16: 68023308-68023308 on Build GRCh38
rs1001410260 Chr.16: 68023215-68023215 on Build GRCh38
rs780718607 Chr.16: 68023229-68023229 on Build GRCh38
rs778740776 Chr.16: 68023176-68023176 on Build GRCh38
rs777420050 Chr.16: 68023180-68023180 on Build GRCh38
rs953537554 Chr.16: 68023595-68023595 on Build GRCh38
rs754856841 Chr.16: 68023659-68023659 on Build GRCh38
rs761501908 Chr.16: 68023654-68023654 on Build GRCh38
rs549140683 Chr.16: 68023424-68023424 on Build GRCh38
rs774734563 Chr.16: 68023197-68023197 on Build GRCh38
rs923440272 Chr.16: 68023261-68023261 on Build GRCh38
rs765166334 Chr.16: 68023207-68023207 on Build GRCh38
rs1035935495 Chr.16: 68023492-68023492 on Build GRCh38
rs200362568 Chr.16: 68023173-68023173 on Build GRCh38
rs979693807 Chr.16: 68023607-68023607 on Build GRCh38
rs866003090 Chr.16: 68023438-68023438 on Build GRCh38
rs1014624782 Chr.16: 68023515-68023515 on Build GRCh38
rs879108591 Chr.16: 68023314-68023315 on Build GRCh38
rs201938424 Chr.16: 68023198-68023198 on Build GRCh38

View Full Product Details