Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74864292-74864848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FDXR
Entrez Gene ID
2232
Gene Name
ferredoxin reductase
Gene Aliases
ADXR
UniGene
Hs.69745
Gene Chromosome Location
Chr.17: 74862497-74873031 on Build GRCh38
Transcripts
NM_001258012.3 NM_001258013.3 NM_001258014.3 NM_001258015.2 NM_001258016.3 NM_004110.5 NM_024417.4 NR_047576.2 XM_006721772.1 XM_011524527.1 XM_011524528.1 XM_011524529.1 XM_011524531.2 XM_011524532.2 XM_011524533.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4859196 c.658G>T p.D220Y Chr.17: 74864492-74864492 on Build GRCh38
3701275 c.670+8G>T p.? Chr.17: 74864472-74864472 on Build GRCh38
6238614 c.616G>A p.A206T Chr.17: 74864534-74864534 on Build GRCh38
311160 c.770G>T p.R257L Chr.17: 74864530-74864530 on Build GRCh38
6556652 c.834G>A p.P278P Chr.17: 74864334-74864334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34654930 Chr.17: 74864680-74864680 on Build GRCh38
rs970119640 Chr.17: 74864747-74864747 on Build GRCh38
rs746189837 Chr.17: 74864386-74864386 on Build GRCh38
rs953069349 Chr.17: 74864646-74864646 on Build GRCh38
rs750268654 Chr.17: 74864477-74864477 on Build GRCh38
rs781601242 Chr.17: 74864588-74864588 on Build GRCh38
rs779057290 Chr.17: 74864510-74864510 on Build GRCh38
rs1008504764 Chr.17: 74864705-74864705 on Build GRCh38
rs751576831 Chr.17: 74864492-74864492 on Build GRCh38
rs190983508 Chr.17: 74864694-74864694 on Build GRCh38
rs773043685 Chr.17: 74864537-74864537 on Build GRCh38
rs750702385 Chr.17: 74864576-74864576 on Build GRCh38
rs903850582 Chr.17: 74864675-74864675 on Build GRCh38
rs112568790 Chr.17: 74864368-74864368 on Build GRCh38
rs758443678 Chr.17: 74864346-74864346 on Build GRCh38
rs751251788 Chr.17: 74864541-74864541 on Build GRCh38
rs769410057 Chr.17: 74864534-74864534 on Build GRCh38
rs1028613307 Chr.17: 74864414-74864414 on Build GRCh38
rs1043814399 Chr.17: 74864668-74864668 on Build GRCh38
rs944540388 Chr.17: 74864420-74864420 on Build GRCh38
rs575876220 Chr.17: 74864453-74864453 on Build GRCh38
rs749258224 Chr.17: 74864502-74864502 on Build GRCh38
rs767942793 Chr.17: 74864320-74864320 on Build GRCh38
rs775408048 Chr.17: 74864454-74864455 on Build GRCh38
rs765315364 Chr.17: 74864783-74864783 on Build GRCh38
rs763488904 Chr.17: 74864334-74864334 on Build GRCh38
rs201737074 Chr.17: 74864393-74864393 on Build GRCh38
rs1015744872 Chr.17: 74864713-74864713 on Build GRCh38
rs35602227 Chr.17: 74864367-74864367 on Build GRCh38
rs751971921 Chr.17: 74864562-74864562 on Build GRCh38
rs777919675 Chr.17: 74864501-74864501 on Build GRCh38
rs769499990 Chr.17: 74864591-74864591 on Build GRCh38
rs979478271 Chr.17: 74864788-74864788 on Build GRCh38
rs752616185 Chr.17: 74864781-74864781 on Build GRCh38
rs903441534 Chr.17: 74864625-74864625 on Build GRCh38
rs772055603 Chr.17: 74864439-74864465 on Build GRCh38
rs891340953 Chr.17: 74864696-74864696 on Build GRCh38
rs1035765959 Chr.17: 74864760-74864760 on Build GRCh38
rs764945182 Chr.17: 74864460-74864460 on Build GRCh38
rs754853560 Chr.17: 74864498-74864498 on Build GRCh38
rs538104936 Chr.17: 74864508-74864508 on Build GRCh38
rs760182097 Chr.17: 74864330-74864330 on Build GRCh38
rs761082090 Chr.17: 74864390-74864390 on Build GRCh38
rs564282507 Chr.17: 74864726-74864726 on Build GRCh38
rs775011125 Chr.17: 74864326-74864326 on Build GRCh38
rs1018686057 Chr.17: 74864663-74864663 on Build GRCh38
rs374472874 Chr.17: 74864613-74864613 on Build GRCh38
rs966221790 Chr.17: 74864665-74864665 on Build GRCh38
rs997488966 Chr.17: 74864401-74864401 on Build GRCh38
rs34964564 Chr.17: 74864800-74864800 on Build GRCh38
rs369277931 Chr.17: 74864516-74864516 on Build GRCh38
rs377589541 Chr.17: 74864579-74864579 on Build GRCh38
rs34038065 Chr.17: 74864531-74864531 on Build GRCh38
rs753973181 Chr.17: 74864556-74864556 on Build GRCh38
rs747565176 Chr.17: 74864530-74864530 on Build GRCh38
rs749579566 Chr.17: 74864373-74864373 on Build GRCh38
rs1026772364 Chr.17: 74864744-74864744 on Build GRCh38
rs772588733 Chr.17: 74864387-74864387 on Build GRCh38
rs913084954 Chr.17: 74864628-74864628 on Build GRCh38
rs756847640 Chr.17: 74864338-74864338 on Build GRCh38
rs777429435 Chr.17: 74864819-74864819 on Build GRCh38
rs746066884 Chr.17: 74864515-74864515 on Build GRCh38
rs561864175 Chr.17: 74864653-74864653 on Build GRCh38
rs373972990 Chr.17: 74864782-74864782 on Build GRCh38
rs913053748 Chr.17: 74864777-74864777 on Build GRCh38
rs763719128 Chr.17: 74864392-74864392 on Build GRCh38
rs753592918 Chr.17: 74864335-74864335 on Build GRCh38
rs111758978 Chr.17: 74864808-74864808 on Build GRCh38
rs35959449 Chr.17: 74864591-74864592 on Build GRCh38
rs748637680 Chr.17: 74864590-74864590 on Build GRCh38
rs760291395 Chr.17: 74864587-74864587 on Build GRCh38
rs369527938 Chr.17: 74864773-74864773 on Build GRCh38
rs758648690 Chr.17: 74864794-74864794 on Build GRCh38
rs2070921 Chr.17: 74864532-74864532 on Build GRCh38
rs891282851 Chr.17: 74864440-74864440 on Build GRCh38
rs762704784 Chr.17: 74864615-74864615 on Build GRCh38
rs539931119 Chr.17: 74864341-74864341 on Build GRCh38
rs571003894 Chr.17: 74864339-74864339 on Build GRCh38
rs750435603 Chr.17: 74864790-74864790 on Build GRCh38
rs758559421 Chr.17: 74864577-74864577 on Build GRCh38
rs769433044 Chr.17: 74864829-74864829 on Build GRCh38
rs971920607 Chr.17: 74864689-74864689 on Build GRCh38
rs199777183 Chr.17: 74864568-74864568 on Build GRCh38
rs35072974 Chr.17: 74864539-74864539 on Build GRCh38
rs763768416 Chr.17: 74864777-74864778 on Build GRCh38
rs35637152 Chr.17: 74864693-74864693 on Build GRCh38
rs758320615 Chr.17: 74864481-74864481 on Build GRCh38
rs959959699 Chr.17: 74864762-74864762 on Build GRCh38
rs766233753 Chr.17: 74864674-74864674 on Build GRCh38
rs766250055 Chr.17: 74864490-74864490 on Build GRCh38
rs370506060 Chr.17: 74864779-74864779 on Build GRCh38
rs774700651 Chr.17: 74864380-74864380 on Build GRCh38
rs547136156 Chr.17: 74864815-74864815 on Build GRCh38
rs541796147 Chr.17: 74864616-74864616 on Build GRCh38
rs748770358 Chr.17: 74864828-74864828 on Build GRCh38
rs753323215 Chr.17: 74864813-74864813 on Build GRCh38
rs770998084 Chr.17: 74864379-74864379 on Build GRCh38
rs781612378 Chr.17: 74864804-74864804 on Build GRCh38
rs747298107 Chr.17: 74864581-74864581 on Build GRCh38
rs780093358 Chr.17: 74864358-74864358 on Build GRCh38
rs775819257 Chr.17: 74864529-74864529 on Build GRCh38
rs988702605 Chr.17: 74864769-74864769 on Build GRCh38
rs143449689 Chr.17: 74864533-74864533 on Build GRCh38
rs375282282 Chr.17: 74864799-74864799 on Build GRCh38
rs755334781 Chr.17: 74864802-74864802 on Build GRCh38
rs897358847 Chr.17: 74864722-74864722 on Build GRCh38
rs373740363 Chr.17: 74864551-74864551 on Build GRCh38
rs757375278 Chr.17: 74864558-74864558 on Build GRCh38
rs113443078 Chr.17: 74864389-74864389 on Build GRCh38
rs979755554 Chr.17: 74864757-74864757 on Build GRCh38
rs545500381 Chr.17: 74864538-74864538 on Build GRCh38
rs769231215 Chr.17: 74864587-74864587 on Build GRCh38
rs146549848 Chr.17: 74864552-74864552 on Build GRCh38

View Full Product Details