Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41292145-41292557 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPUL1
Entrez Gene ID
11100
Gene Name
heterogeneous nuclear ribonucleoprotein U like 1
Gene Aliases
E1B-AP5, E1BAP5, HNRPUL1
UniGene
Hs.155218
Gene Chromosome Location
Chr.19: 41262476-41307783 on Build GRCh38
Transcripts
NM_001301016.2 NM_001321208.1 NM_001321211.1 NM_007040.5 NM_144732.4 XM_005258459.3 XM_005258461.4 XM_005258463.3 XM_005258464.3 XM_005258465.4 XM_011526392.2 XM_011526393.2 XM_011526394.2 XM_011526395.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1393954 c.1023G>T p.V341V Chr.19: 41292268-41292268 on Build GRCh38
6681434 c.715G>A p.G239S Chr.19: 41292302-41292302 on Build GRCh38
3534380 c.833C>T p.S278F Chr.19: 41292420-41292420 on Build GRCh38
6681453 c.1132G>A p.A378T Chr.19: 41292377-41292377 on Build GRCh38
4078470 c.1073T>A p.I358N Chr.19: 41292318-41292318 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370119348 Chr.19: 41292198-41292198 on Build GRCh38
rs61738510 Chr.19: 41292362-41292362 on Build GRCh38
rs142989078 Chr.19: 41292266-41292266 on Build GRCh38
rs112213182 Chr.19: 41292512-41292512 on Build GRCh38
rs867126206 Chr.19: 41292457-41292457 on Build GRCh38
rs528087616 Chr.19: 41292394-41292394 on Build GRCh38
rs758463831 Chr.19: 41292482-41292482 on Build GRCh38
rs111667636 Chr.19: 41292243-41292243 on Build GRCh38
rs775843098 Chr.19: 41292196-41292196 on Build GRCh38
rs763580084 Chr.19: 41292414-41292414 on Build GRCh38
rs746834749 Chr.19: 41292518-41292518 on Build GRCh38
rs1045731976 Chr.19: 41292332-41292332 on Build GRCh38
rs12974243 Chr.19: 41292407-41292407 on Build GRCh38
rs764242027 Chr.19: 41292202-41292202 on Build GRCh38
rs546128654 Chr.19: 41292205-41292205 on Build GRCh38
rs964707000 Chr.19: 41292178-41292178 on Build GRCh38
rs983613670 Chr.19: 41292240-41292240 on Build GRCh38
rs908333736 Chr.19: 41292315-41292315 on Build GRCh38
rs750729710 Chr.19: 41292216-41292216 on Build GRCh38
rs111858337 Chr.19: 41292305-41292305 on Build GRCh38
rs369738571 Chr.19: 41292533-41292533 on Build GRCh38
rs369129717 Chr.19: 41292207-41292207 on Build GRCh38
rs751968794 Chr.19: 41292425-41292425 on Build GRCh38
rs750740675 Chr.19: 41292536-41292537 on Build GRCh38
rs747216807 Chr.19: 41292373-41292373 on Build GRCh38
rs777241878 Chr.19: 41292281-41292281 on Build GRCh38
rs763764872 Chr.19: 41292455-41292455 on Build GRCh38
rs8100690 Chr.19: 41292201-41292201 on Build GRCh38
rs377300107 Chr.19: 41292376-41292376 on Build GRCh38
rs779949595 Chr.19: 41292487-41292487 on Build GRCh38
rs372936991 Chr.19: 41292334-41292334 on Build GRCh38
rs879077564 Chr.19: 41292335-41292335 on Build GRCh38
rs950889549 Chr.19: 41292219-41292219 on Build GRCh38
rs748818271 Chr.19: 41292296-41292296 on Build GRCh38
rs766697507 Chr.19: 41292417-41292417 on Build GRCh38
rs755713629 Chr.19: 41292433-41292433 on Build GRCh38
rs563533035 Chr.19: 41292175-41292175 on Build GRCh38
rs376980850 Chr.19: 41292197-41292197 on Build GRCh38
rs757176507 Chr.19: 41292317-41292317 on Build GRCh38
rs756710629 Chr.19: 41292471-41292471 on Build GRCh38
rs778553397 Chr.19: 41292473-41292473 on Build GRCh38
rs3752705 Chr.19: 41292206-41292206 on Build GRCh38
rs62120963 Chr.19: 41292193-41292193 on Build GRCh38
rs780452136 Chr.19: 41292220-41292220 on Build GRCh38
rs775466827 Chr.19: 41292347-41292347 on Build GRCh38
rs752338996 Chr.19: 41292225-41292225 on Build GRCh38
rs768935854 Chr.19: 41292525-41292525 on Build GRCh38
rs776815219 Chr.19: 41292385-41292385 on Build GRCh38
rs758701018 Chr.19: 41292218-41292218 on Build GRCh38
rs763871796 Chr.19: 41292481-41292481 on Build GRCh38
rs765715604 Chr.19: 41292409-41292409 on Build GRCh38
rs777003728 Chr.19: 41292532-41292532 on Build GRCh38
rs545165335 Chr.19: 41292410-41292410 on Build GRCh38
rs981851328 Chr.19: 41292350-41292350 on Build GRCh38
rs771760316 Chr.19: 41292338-41292338 on Build GRCh38
rs753332324 Chr.19: 41292467-41292467 on Build GRCh38

View Full Product Details