Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 16589181-16589689 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTPS2
Entrez Gene ID
56474
Gene Name
CTP synthase 2
Gene Aliases
-
UniGene
Hs.227049
Gene Chromosome Location
Chr.X: 16587999-16712979 on Build GRCh38
Transcripts
NM_001144002.1 NM_019857.4 NM_175859.2 XM_005274562.3 XM_006724503.3 XM_011545545.2

SNP Detail

SNP ID Chromosome Location
rs990932377 Chr.X: 16589456-16589456 on Build GRCh38
rs780868462 Chr.X: 16589244-16589244 on Build GRCh38
rs748855029 Chr.X: 16589411-16589411 on Build GRCh38
rs976901824 Chr.X: 16589664-16589664 on Build GRCh38
rs916652397 Chr.X: 16589458-16589458 on Build GRCh38
rs369789635 Chr.X: 16589588-16589588 on Build GRCh38
rs773512952 Chr.X: 16589526-16589533 on Build GRCh38
rs894585036 Chr.X: 16589219-16589219 on Build GRCh38
rs1024842239 Chr.X: 16589275-16589275 on Build GRCh38
rs1052962097 Chr.X: 16589657-16589657 on Build GRCh38
rs780174709 Chr.X: 16589407-16589407 on Build GRCh38
rs994341780 Chr.X: 16589419-16589419 on Build GRCh38
rs142245573 Chr.X: 16589265-16589265 on Build GRCh38
rs750511891 Chr.X: 16589291-16589291 on Build GRCh38
rs1055777576 Chr.X: 16589212-16589212 on Build GRCh38
rs963868158 Chr.X: 16589610-16589610 on Build GRCh38
rs377292018 Chr.X: 16589441-16589441 on Build GRCh38
rs187023858 Chr.X: 16589424-16589424 on Build GRCh38
rs56111615 Chr.X: 16589466-16589466 on Build GRCh38
rs1025533290 Chr.X: 16589467-16589467 on Build GRCh38
rs910870654 Chr.X: 16589606-16589606 on Build GRCh38
rs961618699 Chr.X: 16589364-16589364 on Build GRCh38
rs769334192 Chr.X: 16589268-16589268 on Build GRCh38
rs925869948 Chr.X: 16589567-16589567 on Build GRCh38
rs761052109 Chr.X: 16589632-16589632 on Build GRCh38
rs771404273 Chr.X: 16589494-16589494 on Build GRCh38
rs993975216 Chr.X: 16589438-16589438 on Build GRCh38
rs772703014 Chr.X: 16589310-16589310 on Build GRCh38
rs968471192 Chr.X: 16589546-16589546 on Build GRCh38
rs1017621307 Chr.X: 16589304-16589304 on Build GRCh38
rs12010843 Chr.X: 16589560-16589560 on Build GRCh38
rs952602586 Chr.X: 16589491-16589491 on Build GRCh38
rs756215885 Chr.X: 16589349-16589350 on Build GRCh38

View Full Product Details