Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 192142163-192142713 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF12
Entrez Gene ID
2257
Gene Name
fibroblast growth factor 12
Gene Aliases
FGF12B, FHF1
UniGene
Hs.390250
Gene Chromosome Location
Chr.3: 192139393-192727599 on Build GRCh38
Transcripts
NM_004113.5 NM_021032.4 XM_005247227.2 XM_006713538.3 XM_017005879.1

SNP Detail

SNP ID Chromosome Location
rs1043037018 Chr.3: 192142393-192142393 on Build GRCh38
rs35460777 Chr.3: 192142474-192142474 on Build GRCh38
rs569029847 Chr.3: 192142667-192142667 on Build GRCh38
rs548436307 Chr.3: 192142209-192142209 on Build GRCh38
rs773046696 Chr.3: 192142551-192142551 on Build GRCh38
rs1035575002 Chr.3: 192142522-192142522 on Build GRCh38
rs563079931 Chr.3: 192142300-192142300 on Build GRCh38
rs532920249 Chr.3: 192142386-192142386 on Build GRCh38
rs983702613 Chr.3: 192142221-192142221 on Build GRCh38
rs951090641 Chr.3: 192142449-192142449 on Build GRCh38
rs938781784 Chr.3: 192142280-192142280 on Build GRCh38
rs928557190 Chr.3: 192142214-192142214 on Build GRCh38
rs971982983 Chr.3: 192142189-192142189 on Build GRCh38
rs1041269217 Chr.3: 192142411-192142411 on Build GRCh38
rs772666190 Chr.3: 192142626-192142629 on Build GRCh38
rs998350679 Chr.3: 192142420-192142420 on Build GRCh38
rs562297442 Chr.3: 192142293-192142293 on Build GRCh38
rs548947333 Chr.3: 192142572-192142572 on Build GRCh38
rs370752801 Chr.3: 192142504-192142504 on Build GRCh38
rs983648914 Chr.3: 192142458-192142458 on Build GRCh38
rs934811142 Chr.3: 192142688-192142688 on Build GRCh38
rs1018538584 Chr.3: 192142404-192142404 on Build GRCh38
rs960228947 Chr.3: 192142555-192142555 on Build GRCh38
rs891811738 Chr.3: 192142353-192142353 on Build GRCh38
rs535657945 Chr.3: 192142504-192142504 on Build GRCh38
rs900897514 Chr.3: 192142418-192142418 on Build GRCh38
rs1025300037 Chr.3: 192142429-192142429 on Build GRCh38
rs950932923 Chr.3: 192142186-192142186 on Build GRCh38
rs1016497263 Chr.3: 192142470-192142470 on Build GRCh38
rs915562545 Chr.3: 192142575-192142575 on Build GRCh38
rs942224130 Chr.3: 192142267-192142267 on Build GRCh38
rs397991975 Chr.3: 192142479-192142479 on Build GRCh38
rs374884808 Chr.3: 192142314-192142314 on Build GRCh38
rs370538965 Chr.3: 192142250-192142250 on Build GRCh38
rs566728289 Chr.3: 192142496-192142496 on Build GRCh38
rs924375732 Chr.3: 192142651-192142651 on Build GRCh38
rs191251644 Chr.3: 192142279-192142279 on Build GRCh38
rs748011491 Chr.3: 192142367-192142367 on Build GRCh38
rs571198053 Chr.3: 192142384-192142384 on Build GRCh38
rs927453945 Chr.3: 192142507-192142507 on Build GRCh38
rs183750358 Chr.3: 192142352-192142352 on Build GRCh38
rs139892590 Chr.3: 192142398-192142398 on Build GRCh38
rs972317371 Chr.3: 192142519-192142519 on Build GRCh38
rs377480214 Chr.3: 192142547-192142547 on Build GRCh38
rs550625595 Chr.3: 192142336-192142336 on Build GRCh38
rs946054372 Chr.3: 192142668-192142668 on Build GRCh38
rs200260911 Chr.3: 192142612-192142616 on Build GRCh38
rs934841506 Chr.3: 192142410-192142415 on Build GRCh38
rs913235602 Chr.3: 192142653-192142653 on Build GRCh38
rs867067433 Chr.3: 192142257-192142257 on Build GRCh38
rs776290226 Chr.3: 192142431-192142432 on Build GRCh38
rs1005622054 Chr.3: 192142496-192142496 on Build GRCh38
rs542016358 Chr.3: 192142282-192142282 on Build GRCh38
rs915619677 Chr.3: 192142301-192142301 on Build GRCh38
rs904339144 Chr.3: 192142396-192142396 on Build GRCh38
rs1044859321 Chr.3: 192142269-192142269 on Build GRCh38
rs993358341 Chr.3: 192142640-192142640 on Build GRCh38

View Full Product Details