Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157029602-157030109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAVCR1
Entrez Gene ID
26762
Gene Name
hepatitis A virus cellular receptor 1
Gene Aliases
CD365, HAVCR, HAVCR-1, KIM-1, KIM1, TIM, TIM-1, TIM1, TIMD-1, TIMD1
UniGene
Hs.129711
Gene Chromosome Location
Chr.5: 156979443-157059119 on Build GRCh38
Transcripts
NM_001173393.2 NM_001308156.1 NM_012206.3 XM_011534515.2 XM_017009339.1 XM_017009340.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5910455 c.1039G>A p.E347K Chr.5: 157029789-157029789 on Build GRCh38
449183 c.1044C>T p.V348V Chr.5: 157029784-157029784 on Build GRCh38
737078 c.1030G>T p.V344F Chr.5: 157029798-157029798 on Build GRCh38
1729462 c.1014A>T p.K338N Chr.5: 157029814-157029814 on Build GRCh38
3613047 c.992C>T p.S331L Chr.5: 157029836-157029836 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763732775 Chr.5: 157029783-157029783 on Build GRCh38
rs375338120 Chr.5: 157029862-157029862 on Build GRCh38
rs762367309 Chr.5: 157029878-157029878 on Build GRCh38
rs7716614 Chr.5: 157029851-157029851 on Build GRCh38
rs185380041 Chr.5: 157029739-157029739 on Build GRCh38
rs886322901 Chr.5: 157029917-157029917 on Build GRCh38
rs746516163 Chr.5: 157029669-157029669 on Build GRCh38
rs749667001 Chr.5: 157029634-157029635 on Build GRCh38
rs965382912 Chr.5: 157030026-157030026 on Build GRCh38
rs765369679 Chr.5: 157029742-157029742 on Build GRCh38
rs761010730 Chr.5: 157029636-157029636 on Build GRCh38
rs780983817 Chr.5: 157029696-157029696 on Build GRCh38
rs571014946 Chr.5: 157029662-157029662 on Build GRCh38
rs111759551 Chr.5: 157029720-157029720 on Build GRCh38
rs764346701 Chr.5: 157029642-157029642 on Build GRCh38
rs1005220481 Chr.5: 157029929-157029929 on Build GRCh38
rs1006948106 Chr.5: 157029966-157029966 on Build GRCh38
rs752549782 Chr.5: 157029772-157029772 on Build GRCh38
rs568638686 Chr.5: 157029827-157029827 on Build GRCh38
rs199878594 Chr.5: 157029820-157029820 on Build GRCh38
rs971451690 Chr.5: 157029961-157029961 on Build GRCh38
rs772667379 Chr.5: 157029729-157029729 on Build GRCh38
rs774814370 Chr.5: 157029904-157029904 on Build GRCh38
rs536424242 Chr.5: 157029924-157029924 on Build GRCh38
rs373813178 Chr.5: 157029723-157029723 on Build GRCh38
rs770483110 Chr.5: 157029710-157029710 on Build GRCh38
rs2134230 Chr.5: 157029946-157029946 on Build GRCh38
rs950729585 Chr.5: 157029706-157029706 on Build GRCh38
rs368278246 Chr.5: 157029816-157029816 on Build GRCh38
rs1018300205 Chr.5: 157030009-157030009 on Build GRCh38
rs745340159 Chr.5: 157029797-157029797 on Build GRCh38
rs775949578 Chr.5: 157029632-157029632 on Build GRCh38
rs568070159 Chr.5: 157029993-157029993 on Build GRCh38
rs977153599 Chr.5: 157030042-157030042 on Build GRCh38
rs777473215 Chr.5: 157029703-157029703 on Build GRCh38
rs942742881 Chr.5: 157029866-157029866 on Build GRCh38
rs979866131 Chr.5: 157029732-157029732 on Build GRCh38
rs551851890 Chr.5: 157029826-157029826 on Build GRCh38
rs773225254 Chr.5: 157029766-157029766 on Build GRCh38
rs1048883012 Chr.5: 157029911-157029911 on Build GRCh38
rs768944549 Chr.5: 157029678-157029679 on Build GRCh38
rs1017369783 Chr.5: 157029680-157029680 on Build GRCh38
rs898617229 Chr.5: 157029898-157029898 on Build GRCh38
rs200614570 Chr.5: 157029740-157029740 on Build GRCh38
rs772581266 Chr.5: 157029628-157029628 on Build GRCh38
rs766292669 Chr.5: 157029667-157029667 on Build GRCh38
rs145237072 Chr.5: 157029698-157029698 on Build GRCh38
rs181656381 Chr.5: 157029686-157029686 on Build GRCh38
rs369159990 Chr.5: 157029685-157029685 on Build GRCh38
rs753538218 Chr.5: 157029881-157029881 on Build GRCh38
rs774694582 Chr.5: 157029846-157029849 on Build GRCh38
rs137884244 Chr.5: 157029704-157029704 on Build GRCh38
rs753544741 Chr.5: 157029784-157029784 on Build GRCh38
rs373996191 Chr.5: 157029800-157029800 on Build GRCh38
rs201441165 Chr.5: 157029778-157029778 on Build GRCh38
rs751500388 Chr.5: 157029676-157029676 on Build GRCh38
rs866833306 Chr.5: 157030056-157030056 on Build GRCh38
rs759496799 Chr.5: 157029770-157029770 on Build GRCh38
rs994391699 Chr.5: 157029916-157029916 on Build GRCh38
rs1014326808 Chr.5: 157029890-157029890 on Build GRCh38
rs755774394 Chr.5: 157029700-157029700 on Build GRCh38
rs34283489 Chr.5: 157029996-157029997 on Build GRCh38
rs2173964 Chr.5: 157030070-157030070 on Build GRCh38
rs1051537562 Chr.5: 157029645-157029645 on Build GRCh38
rs188551881 Chr.5: 157029796-157029796 on Build GRCh38
rs150173323 Chr.5: 157029803-157029803 on Build GRCh38
rs376920430 Chr.5: 157029738-157029738 on Build GRCh38
rs763973623 Chr.5: 157029871-157029871 on Build GRCh38
rs927052304 Chr.5: 157029768-157029768 on Build GRCh38
rs780853400 Chr.5: 157029728-157029728 on Build GRCh38
rs769067757 Chr.5: 157029814-157029814 on Build GRCh38
rs767408402 Chr.5: 157029771-157029771 on Build GRCh38
rs753914129 Chr.5: 157029659-157029659 on Build GRCh38
rs368598624 Chr.5: 157029870-157029870 on Build GRCh38
rs772606297 Chr.5: 157029963-157029963 on Build GRCh38
rs776039147 Chr.5: 157029735-157029735 on Build GRCh38
rs1034097846 Chr.5: 157030000-157030000 on Build GRCh38
rs76133899 Chr.5: 157029891-157029891 on Build GRCh38

View Full Product Details