Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 135393496-135394046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT4
Entrez Gene ID
4850
Gene Name
CCR4-NOT transcription complex subunit 4
Gene Aliases
CLONE243, NOT4, NOT4H
UniGene
Hs.490224
Gene Chromosome Location
Chr.7: 135361795-135510127 on Build GRCh38
Transcripts
NM_001008225.2 NM_001190847.1 NM_001190848.1 NM_001190849.1 NM_001190850.1 NM_013316.3 XM_017012235.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1085831 c.1560C>A p.F520L Chr.7: 135393976-135393976 on Build GRCh38
744428 c.1617A>T p.A539A Chr.7: 135393919-135393919 on Build GRCh38
4004303 c.1507T>G p.F503V Chr.7: 135394038-135394038 on Build GRCh38
5900089 c.1573C>T p.L525F Chr.7: 135393963-135393963 on Build GRCh38
1085833 c.1497C>A p.A499A Chr.7: 135394039-135394039 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545080566 Chr.7: 135393977-135393977 on Build GRCh38
rs557922306 Chr.7: 135393601-135393601 on Build GRCh38
rs1017308192 Chr.7: 135393953-135393953 on Build GRCh38
rs763112694 Chr.7: 135393969-135393969 on Build GRCh38
rs778356590 Chr.7: 135393875-135393875 on Build GRCh38
rs747338142 Chr.7: 135393927-135393927 on Build GRCh38
rs397841662 Chr.7: 135393545-135393545 on Build GRCh38
rs372237832 Chr.7: 135394025-135394025 on Build GRCh38
rs1045063460 Chr.7: 135393586-135393586 on Build GRCh38
rs947655338 Chr.7: 135393701-135393701 on Build GRCh38
rs775478778 Chr.7: 135393962-135393962 on Build GRCh38
rs1030420831 Chr.7: 135393880-135393880 on Build GRCh38
rs751835452 Chr.7: 135393978-135393978 on Build GRCh38
rs926597958 Chr.7: 135393598-135393598 on Build GRCh38
rs755914315 Chr.7: 135393988-135393988 on Build GRCh38
rs900727687 Chr.7: 135393817-135393817 on Build GRCh38
rs749708841 Chr.7: 135393934-135393935 on Build GRCh38
rs1029908544 Chr.7: 135393991-135393991 on Build GRCh38
rs28578709 Chr.7: 135393538-135393538 on Build GRCh38
rs1043294097 Chr.7: 135393811-135393811 on Build GRCh38
rs754914456 Chr.7: 135394012-135394012 on Build GRCh38
rs187567413 Chr.7: 135393643-135393643 on Build GRCh38
rs574209635 Chr.7: 135393681-135393681 on Build GRCh38
rs939300926 Chr.7: 135393605-135393605 on Build GRCh38
rs369479270 Chr.7: 135393820-135393820 on Build GRCh38
rs778465540 Chr.7: 135394016-135394016 on Build GRCh38
rs1056367176 Chr.7: 135393608-135393608 on Build GRCh38
rs766333798 Chr.7: 135393992-135393992 on Build GRCh38
rs537919063 Chr.7: 135393587-135393587 on Build GRCh38
rs147452844 Chr.7: 135393617-135393617 on Build GRCh38
rs903623103 Chr.7: 135393703-135393703 on Build GRCh38
rs533525464 Chr.7: 135393622-135393622 on Build GRCh38
rs765326803 Chr.7: 135393961-135393961 on Build GRCh38
rs771293695 Chr.7: 135393933-135393933 on Build GRCh38
rs3841170 Chr.7: 135393538-135393538 on Build GRCh38
rs757378583 Chr.7: 135393604-135393604 on Build GRCh38
rs938378858 Chr.7: 135393592-135393593 on Build GRCh38
rs1007284828 Chr.7: 135393809-135393809 on Build GRCh38
rs374569292 Chr.7: 135393958-135393958 on Build GRCh38
rs753820571 Chr.7: 135393997-135393997 on Build GRCh38
rs754495360 Chr.7: 135393867-135393867 on Build GRCh38
rs771672346 Chr.7: 135393895-135393895 on Build GRCh38
rs1043911426 Chr.7: 135393666-135393666 on Build GRCh38
rs190906616 Chr.7: 135393959-135393959 on Build GRCh38
rs780540697 Chr.7: 135393874-135393875 on Build GRCh38
rs757957704 Chr.7: 135394027-135394027 on Build GRCh38
rs372901306 Chr.7: 135393924-135393924 on Build GRCh38
rs549191256 Chr.7: 135393537-135393537 on Build GRCh38
rs768993320 Chr.7: 135394019-135394021 on Build GRCh38
rs201142434 Chr.7: 135393894-135393894 on Build GRCh38

View Full Product Details