Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28520630-28520993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD3
Entrez Gene ID
7976
Gene Name
frizzled class receptor 3
Gene Aliases
Fz-3
UniGene
Hs.40735
Gene Chromosome Location
Chr.8: 28494205-28574268 on Build GRCh38
Transcripts
NM_017412.3 NM_145866.1 XM_017013841.1 XM_017013842.1 XM_017013843.1 XM_017013844.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5906823 c.190-6C>T p.? Chr.8: 28520632-28520632 on Build GRCh38
281273 c.370G>T p.D124Y Chr.8: 28520818-28520818 on Build GRCh38
6318274 c.352G>T p.G118C Chr.8: 28520800-28520800 on Build GRCh38
4453266 c.272A>C p.E91A Chr.8: 28520720-28520720 on Build GRCh38
6180722 c.285C>G p.V95V Chr.8: 28520733-28520733 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs924100903 Chr.8: 28520936-28520936 on Build GRCh38
rs757928052 Chr.8: 28520704-28520704 on Build GRCh38
rs566937390 Chr.8: 28520890-28520890 on Build GRCh38
rs763767677 Chr.8: 28520681-28520681 on Build GRCh38
rs912770984 Chr.8: 28520907-28520907 on Build GRCh38
rs761638226 Chr.8: 28520802-28520802 on Build GRCh38
rs746607374 Chr.8: 28520718-28520718 on Build GRCh38
rs1003095822 Chr.8: 28520839-28520839 on Build GRCh38
rs768401363 Chr.8: 28520719-28520719 on Build GRCh38
rs199631551 Chr.8: 28520806-28520806 on Build GRCh38
rs34550578 Chr.8: 28520742-28520743 on Build GRCh38
rs748470823 Chr.8: 28520728-28520728 on Build GRCh38
rs113879840 Chr.8: 28520856-28520856 on Build GRCh38
rs756456566 Chr.8: 28520873-28520873 on Build GRCh38
rs758445861 Chr.8: 28520815-28520815 on Build GRCh38
rs879858180 Chr.8: 28520746-28520746 on Build GRCh38
rs73230974 Chr.8: 28520899-28520899 on Build GRCh38
rs780727541 Chr.8: 28520720-28520720 on Build GRCh38
rs372232529 Chr.8: 28520876-28520876 on Build GRCh38
rs112453513 Chr.8: 28520872-28520872 on Build GRCh38
rs1015439073 Chr.8: 28520863-28520863 on Build GRCh38
rs778956876 Chr.8: 28520701-28520701 on Build GRCh38
rs779405901 Chr.8: 28520824-28520824 on Build GRCh38
rs372861034 Chr.8: 28520949-28520949 on Build GRCh38
rs778178225 Chr.8: 28520875-28520875 on Build GRCh38
rs780829817 Chr.8: 28520854-28520854 on Build GRCh38
rs368048184 Chr.8: 28520703-28520703 on Build GRCh38
rs754475290 Chr.8: 28520838-28520838 on Build GRCh38
rs1000049128 Chr.8: 28520908-28520911 on Build GRCh38
rs771519523 Chr.8: 28520881-28520881 on Build GRCh38
rs760665801 Chr.8: 28520872-28520872 on Build GRCh38
rs1042549514 Chr.8: 28520674-28520674 on Build GRCh38
rs1034658953 Chr.8: 28520921-28520921 on Build GRCh38
rs779139892 Chr.8: 28520916-28520917 on Build GRCh38
rs751028612 Chr.8: 28520831-28520831 on Build GRCh38
rs770147193 Chr.8: 28520729-28520729 on Build GRCh38
rs773651025 Chr.8: 28520751-28520751 on Build GRCh38
rs200008265 Chr.8: 28520780-28520780 on Build GRCh38
rs564721361 Chr.8: 28520714-28520714 on Build GRCh38
rs772981106 Chr.8: 28520871-28520871 on Build GRCh38
rs993179857 Chr.8: 28520945-28520945 on Build GRCh38
rs765141407 Chr.8: 28520805-28520805 on Build GRCh38
rs757143113 Chr.8: 28520697-28520697 on Build GRCh38
rs770995177 Chr.8: 28520775-28520775 on Build GRCh38
rs759657609 Chr.8: 28520781-28520781 on Build GRCh38
rs137887908 Chr.8: 28520785-28520785 on Build GRCh38
rs1012271105 Chr.8: 28520762-28520762 on Build GRCh38
rs942969330 Chr.8: 28520794-28520794 on Build GRCh38
rs978204616 Chr.8: 28520918-28520918 on Build GRCh38
rs918752522 Chr.8: 28520694-28520694 on Build GRCh38
rs759871121 Chr.8: 28520671-28520671 on Build GRCh38
rs965657334 Chr.8: 28520908-28520908 on Build GRCh38
rs745923648 Chr.8: 28520882-28520882 on Build GRCh38
rs147828133 Chr.8: 28520878-28520878 on Build GRCh38
rs1041039821 Chr.8: 28520843-28520843 on Build GRCh38

View Full Product Details