Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 85281855-85282400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA13
Entrez Gene ID
377677
Gene Name
carbonic anhydrase 13
Gene Aliases
CAXIII
UniGene
Hs.127189
Gene Chromosome Location
Chr.8: 85245487-85284073 on Build GRCh38
Transcripts
NM_198584.2

SNP Detail

SNP ID Chromosome Location
rs188101124 Chr.8: 85282217-85282217 on Build GRCh38
rs961622405 Chr.8: 85282219-85282219 on Build GRCh38
rs1042318589 Chr.8: 85282134-85282134 on Build GRCh38
rs940942693 Chr.8: 85281983-85281983 on Build GRCh38
rs555510177 Chr.8: 85282123-85282123 on Build GRCh38
rs73265548 Chr.8: 85282272-85282272 on Build GRCh38
rs772315722 Chr.8: 85282365-85282365 on Build GRCh38
rs1018691395 Chr.8: 85281947-85281947 on Build GRCh38
rs866177402 Chr.8: 85281907-85281907 on Build GRCh38
rs181140491 Chr.8: 85282143-85282143 on Build GRCh38
rs4617148 Chr.8: 85282132-85282132 on Build GRCh38
rs752191783 Chr.8: 85282105-85282105 on Build GRCh38
rs953043472 Chr.8: 85282010-85282010 on Build GRCh38
rs56031817 Chr.8: 85282261-85282261 on Build GRCh38
rs538292400 Chr.8: 85281918-85281918 on Build GRCh38
rs748180781 Chr.8: 85282351-85282351 on Build GRCh38
rs893641178 Chr.8: 85282132-85282137 on Build GRCh38
rs915347473 Chr.8: 85282267-85282267 on Build GRCh38
rs55702127 Chr.8: 85282258-85282258 on Build GRCh38
rs973562731 Chr.8: 85281992-85281992 on Build GRCh38
rs908833625 Chr.8: 85282040-85282040 on Build GRCh38
rs982048245 Chr.8: 85282223-85282223 on Build GRCh38
rs927992169 Chr.8: 85282252-85282252 on Build GRCh38
rs575290086 Chr.8: 85282064-85282064 on Build GRCh38
rs943757766 Chr.8: 85282110-85282110 on Build GRCh38
rs931027638 Chr.8: 85282031-85282031 on Build GRCh38
rs1055334351 Chr.8: 85282126-85282126 on Build GRCh38
rs78751054 Chr.8: 85282155-85282155 on Build GRCh38
rs139173356 Chr.8: 85282318-85282318 on Build GRCh38
rs6994520 Chr.8: 85282311-85282311 on Build GRCh38
rs995948461 Chr.8: 85282202-85282202 on Build GRCh38
rs560381676 Chr.8: 85282262-85282262 on Build GRCh38
rs542935838 Chr.8: 85282147-85282147 on Build GRCh38
rs938124933 Chr.8: 85282124-85282124 on Build GRCh38
rs759149925 Chr.8: 85281939-85281939 on Build GRCh38
rs920774837 Chr.8: 85282001-85282001 on Build GRCh38
rs964788215 Chr.8: 85282200-85282200 on Build GRCh38
rs561088600 Chr.8: 85282303-85282303 on Build GRCh38
rs781249780 Chr.8: 85282208-85282208 on Build GRCh38
rs940012260 Chr.8: 85282060-85282061 on Build GRCh38
rs944620180 Chr.8: 85282327-85282327 on Build GRCh38
rs185395782 Chr.8: 85282203-85282203 on Build GRCh38
rs148140280 Chr.8: 85281948-85281948 on Build GRCh38
rs142399076 Chr.8: 85282108-85282108 on Build GRCh38
rs74472386 Chr.8: 85282285-85282285 on Build GRCh38
rs984281920 Chr.8: 85282209-85282209 on Build GRCh38
rs975963079 Chr.8: 85281962-85281962 on Build GRCh38

View Full Product Details