Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 121479758-121479930 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGFR2
Entrez Gene ID
2263
Gene Name
fibroblast growth factor receptor 2
Gene Aliases
BBDS, BEK, BFR-1, CD332, CEK3, CFD1, ECT1, JWS, K-SAM, KGFR, TK14, TK25
UniGene
Hs.533683
Gene Chromosome Location
Chr.10: 121478330-121598656 on Build GRCh38
Transcripts
NM_000141.4 NM_001144914.1 NM_001144915.1 NM_001144916.1 NM_001144917.1 NM_001144918.1 NM_001320654.1 NM_001320658.1 NM_022970.3 NM_023029.2 NR_073009.1 XM_006717708.2 XM_006717710.3 XM_006717711.2 XM_006717712.2 XM_017015920.1 XM_017015921.1 XM_017015922.1 XM_017015923.1 XM_017015924.1 XM_017015925.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5596722 c.2411C>T p.P804L Chr.10: 121479912-121479912 on Build GRCh38
1474408 c.2454C>T p.G818G Chr.10: 121479872-121479872 on Build GRCh38
2056115 c.2440C>A p.P814T Chr.10: 121479886-121479886 on Build GRCh38
2056116 c.2437C>A p.P813T Chr.10: 121479886-121479886 on Build GRCh38
6566451 c.2400A>T p.P800P Chr.10: 121479923-121479923 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140318221 Chr.10: 121479866-121479866 on Build GRCh38
rs571201278 Chr.10: 121479786-121479786 on Build GRCh38
rs1006403490 Chr.10: 121479851-121479851 on Build GRCh38
rs749474548 Chr.10: 121479871-121479871 on Build GRCh38
rs747773151 Chr.10: 121479850-121479851 on Build GRCh38
rs144176428 Chr.10: 121479874-121479874 on Build GRCh38
rs558460047 Chr.10: 121479908-121479908 on Build GRCh38
rs866527033 Chr.10: 121479862-121479862 on Build GRCh38
rs775669188 Chr.10: 121479829-121479829 on Build GRCh38
rs746532522 Chr.10: 121479823-121479823 on Build GRCh38
rs759496942 Chr.10: 121479899-121479899 on Build GRCh38
rs766217118 Chr.10: 121479888-121479888 on Build GRCh38
rs764198102 Chr.10: 121479832-121479832 on Build GRCh38
rs886479146 Chr.10: 121479797-121479797 on Build GRCh38
rs371712759 Chr.10: 121479902-121479902 on Build GRCh38
rs763130972 Chr.10: 121479830-121479830 on Build GRCh38
rs774232306 Chr.10: 121479875-121479875 on Build GRCh38
rs567030847 Chr.10: 121479885-121479885 on Build GRCh38
rs764959117 Chr.10: 121479907-121479907 on Build GRCh38
rs368003279 Chr.10: 121479897-121479897 on Build GRCh38
rs773909650 Chr.10: 121479832-121479832 on Build GRCh38
rs547079762 Chr.10: 121479856-121479856 on Build GRCh38
rs533590230 Chr.10: 121479800-121479800 on Build GRCh38
rs771208561 Chr.10: 121479878-121479879 on Build GRCh38

View Full Product Details