Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20780264-20780768 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSM3
Entrez Gene ID
6296
Gene Name
acyl-CoA synthetase medium-chain family member 3
Gene Aliases
SA, SAH
UniGene
Hs.706754
Gene Chromosome Location
Chr.16: 20763990-20797157 on Build GRCh38
Transcripts
NM_005622.3 NM_202000.2 XM_011545911.1 XM_017023523.1 XM_017023524.1 NM_080663.2

Gene Symbol
ERI2
Entrez Gene ID
112479
Gene Name
ERI1 exoribonuclease family member 2
Gene Aliases
EXOD1, ZGRF5
UniGene
Hs.248437
Gene Chromosome Location
Chr.16: 20780193-20806473 on Build GRCh38
Transcripts
NM_005622.3 NM_202000.2 XM_011545911.1 XM_017023523.1 XM_017023524.1 NM_080663.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3888102 c.923G>A p.R308K Chr.16: 20780706-20780706 on Build GRCh38
7161155 c.924G>C p.R308S Chr.16: 20780705-20780705 on Build GRCh38
1129036 c.907C>G p.L303V Chr.16: 20780722-20780722 on Build GRCh38
4059002 c.964A>G p.T322A Chr.16: 20780665-20780665 on Build GRCh38
5473706 c.654C>T p.S218S Chr.16: 20780729-20780729 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748474607 Chr.16: 20780645-20780645 on Build GRCh38
rs771689378 Chr.16: 20780688-20780688 on Build GRCh38
rs932377665 Chr.16: 20780714-20780714 on Build GRCh38
rs191349449 Chr.16: 20780480-20780480 on Build GRCh38
rs1037298707 Chr.16: 20780477-20780477 on Build GRCh38
rs138743502 Chr.16: 20780721-20780721 on Build GRCh38
rs761804163 Chr.16: 20780717-20780717 on Build GRCh38
rs1020636891 Chr.16: 20780410-20780410 on Build GRCh38
rs753782085 Chr.16: 20780600-20780600 on Build GRCh38
rs747536181 Chr.16: 20780628-20780628 on Build GRCh38
rs763848035 Chr.16: 20780703-20780703 on Build GRCh38
rs1037267717 Chr.16: 20780733-20780733 on Build GRCh38
rs140807467 Chr.16: 20780659-20780659 on Build GRCh38
rs567976003 Chr.16: 20780322-20780322 on Build GRCh38
rs745397760 Chr.16: 20780686-20780686 on Build GRCh38
rs758473133 Chr.16: 20780555-20780555 on Build GRCh38
rs778191262 Chr.16: 20780568-20780568 on Build GRCh38
rs765286234 Chr.16: 20780718-20780718 on Build GRCh38
rs770394616 Chr.16: 20780674-20780674 on Build GRCh38
rs995529546 Chr.16: 20780554-20780554 on Build GRCh38
rs752751049 Chr.16: 20780467-20780467 on Build GRCh38
rs568573861 Chr.16: 20780516-20780516 on Build GRCh38
rs954339682 Chr.16: 20780464-20780464 on Build GRCh38
rs11649520 Chr.16: 20780492-20780492 on Build GRCh38
rs376874839 Chr.16: 20780683-20780683 on Build GRCh38
rs752920558 Chr.16: 20780728-20780728 on Build GRCh38
rs113164614 Chr.16: 20780594-20780594 on Build GRCh38
rs199569669 Chr.16: 20780604-20780604 on Build GRCh38
rs754230915 Chr.16: 20780744-20780744 on Build GRCh38
rs201158405 Chr.16: 20780663-20780663 on Build GRCh38
rs765390975 Chr.16: 20780408-20780408 on Build GRCh38
rs756477969 Chr.16: 20780731-20780731 on Build GRCh38
rs759689506 Chr.16: 20780719-20780719 on Build GRCh38
rs535404012 Chr.16: 20780422-20780422 on Build GRCh38
rs760505175 Chr.16: 20780698-20780698 on Build GRCh38
rs910779608 Chr.16: 20780540-20780540 on Build GRCh38
rs534992698 Chr.16: 20780675-20780675 on Build GRCh38
rs778037946 Chr.16: 20780739-20780739 on Build GRCh38
rs766609034 Chr.16: 20780605-20780605 on Build GRCh38
rs751722896 Chr.16: 20780613-20780613 on Build GRCh38
rs781281668 Chr.16: 20780643-20780643 on Build GRCh38
rs755200190 Chr.16: 20780623-20780623 on Build GRCh38
rs113477819 Chr.16: 20780694-20780694 on Build GRCh38
rs756498670 Chr.16: 20780648-20780648 on Build GRCh38
rs987856172 Chr.16: 20780377-20780377 on Build GRCh38
rs776682176 Chr.16: 20780710-20780710 on Build GRCh38
rs140383492 Chr.16: 20780734-20780734 on Build GRCh38
rs537290265 Chr.16: 20780585-20780585 on Build GRCh38
rs150171056 Chr.16: 20780690-20780690 on Build GRCh38
rs561637380 Chr.16: 20780463-20780463 on Build GRCh38

View Full Product Details