Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20409379-20409868 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSM5
Entrez Gene ID
54988
Gene Name
acyl-CoA synthetase medium-chain family member 5
Gene Aliases
-
UniGene
Hs.659606
Gene Chromosome Location
Chr.16: 20409508-20441333 on Build GRCh38
Transcripts
NM_001324371.1 NM_001324372.1 NM_001324373.1 NM_017888.2 XM_006721056.3

SNP Detail

SNP ID Chromosome Location
rs989803222 Chr.16: 20409713-20409713 on Build GRCh38
rs981230275 Chr.16: 20409824-20409824 on Build GRCh38
rs1051319441 Chr.16: 20409712-20409712 on Build GRCh38
rs763736107 Chr.16: 20409750-20409750 on Build GRCh38
rs998728953 Chr.16: 20409634-20409634 on Build GRCh38
rs62034983 Chr.16: 20409720-20409720 on Build GRCh38
rs748526348 Chr.16: 20409807-20409807 on Build GRCh38
rs541925831 Chr.16: 20409691-20409691 on Build GRCh38
rs550494594 Chr.16: 20409406-20409406 on Build GRCh38
rs7199791 Chr.16: 20409610-20409610 on Build GRCh38
rs912739140 Chr.16: 20409756-20409756 on Build GRCh38
rs186015014 Chr.16: 20409606-20409606 on Build GRCh38
rs7198746 Chr.16: 20409775-20409775 on Build GRCh38
rs565958749 Chr.16: 20409516-20409516 on Build GRCh38
rs945698309 Chr.16: 20409706-20409706 on Build GRCh38
rs1018978385 Chr.16: 20409467-20409467 on Build GRCh38
rs979956968 Chr.16: 20409596-20409596 on Build GRCh38
rs957027280 Chr.16: 20409692-20409692 on Build GRCh38
rs1034159133 Chr.16: 20409844-20409844 on Build GRCh38
rs191730922 Chr.16: 20409607-20409607 on Build GRCh38
rs895190343 Chr.16: 20409841-20409841 on Build GRCh38
rs552899955 Chr.16: 20409611-20409611 on Build GRCh38
rs1024153913 Chr.16: 20409614-20409614 on Build GRCh38
rs940215919 Chr.16: 20409765-20409765 on Build GRCh38
rs1055156350 Chr.16: 20409816-20409816 on Build GRCh38
rs563422417 Chr.16: 20409721-20409721 on Build GRCh38
rs1025518249 Chr.16: 20409790-20409790 on Build GRCh38
rs538975699 Chr.16: 20409469-20409469 on Build GRCh38
rs949503160 Chr.16: 20409842-20409842 on Build GRCh38
rs912783785 Chr.16: 20409486-20409486 on Build GRCh38
rs953839691 Chr.16: 20409725-20409725 on Build GRCh38
rs1050843663 Chr.16: 20409428-20409428 on Build GRCh38
rs890873832 Chr.16: 20409715-20409715 on Build GRCh38
rs536891390 Chr.16: 20409591-20409591 on Build GRCh38
rs895262880 Chr.16: 20409594-20409594 on Build GRCh38
rs920211808 Chr.16: 20409779-20409779 on Build GRCh38
rs554676523 Chr.16: 20409526-20409526 on Build GRCh38
rs141997607 Chr.16: 20409657-20409657 on Build GRCh38
rs931344382 Chr.16: 20409544-20409544 on Build GRCh38
rs758054383 Chr.16: 20409631-20409631 on Build GRCh38
rs957178407 Chr.16: 20409847-20409847 on Build GRCh38
rs183956628 Chr.16: 20409795-20409795 on Build GRCh38
rs7199761 Chr.16: 20409567-20409567 on Build GRCh38
rs147583044 Chr.16: 20409448-20409448 on Build GRCh38
rs7198481 Chr.16: 20409485-20409485 on Build GRCh38
rs921055563 Chr.16: 20409419-20409419 on Build GRCh38
rs1025661357 Chr.16: 20409505-20409505 on Build GRCh38
rs891312271 Chr.16: 20409443-20409443 on Build GRCh38
rs373474249 Chr.16: 20409524-20409524 on Build GRCh38

View Full Product Details