Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20427752-20428279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSM5
Entrez Gene ID
54988
Gene Name
acyl-CoA synthetase medium-chain family member 5
Gene Aliases
-
UniGene
Hs.659606
Gene Chromosome Location
Chr.16: 20409508-20441333 on Build GRCh38
Transcripts
NM_001324371.1 NM_001324372.1 NM_017888.2 XM_006721056.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6585251 c.988G>C p.E330Q Chr.16: 20427854-20427854 on Build GRCh38
5724234 c.936C>T p.F312F Chr.16: 20427802-20427802 on Build GRCh38
6884996 c.930C>T p.S310S Chr.16: 20427796-20427796 on Build GRCh38
434798 c.967A>T p.I323F Chr.16: 20427833-20427833 on Build GRCh38
6274570 c.978G>T p.L326L Chr.16: 20427844-20427844 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373901252 Chr.16: 20427897-20427897 on Build GRCh38
rs749458982 Chr.16: 20427889-20427889 on Build GRCh38
rs759664670 Chr.16: 20428254-20428254 on Build GRCh38
rs973016306 Chr.16: 20427997-20427997 on Build GRCh38
rs148662746 Chr.16: 20427830-20427830 on Build GRCh38
rs147407378 Chr.16: 20428238-20428238 on Build GRCh38
rs867050869 Chr.16: 20427802-20427802 on Build GRCh38
rs997927927 Chr.16: 20427907-20427907 on Build GRCh38
rs772496625 Chr.16: 20427916-20427916 on Build GRCh38
rs1012829940 Chr.16: 20428188-20428188 on Build GRCh38
rs760173518 Chr.16: 20427861-20427861 on Build GRCh38
rs12923177 Chr.16: 20427978-20427978 on Build GRCh38
rs1035282172 Chr.16: 20428179-20428179 on Build GRCh38
rs759989634 Chr.16: 20427845-20427845 on Build GRCh38
rs145689236 Chr.16: 20427840-20427840 on Build GRCh38
rs1005342604 Chr.16: 20428101-20428101 on Build GRCh38
rs138922279 Chr.16: 20427789-20427789 on Build GRCh38
rs960869925 Chr.16: 20428186-20428186 on Build GRCh38
rs760046765 Chr.16: 20427786-20427786 on Build GRCh38
rs762567727 Chr.16: 20427871-20427871 on Build GRCh38
rs376883657 Chr.16: 20427778-20427778 on Build GRCh38
rs776021394 Chr.16: 20427857-20427857 on Build GRCh38
rs117080231 Chr.16: 20427880-20427880 on Build GRCh38
rs763570628 Chr.16: 20427864-20427864 on Build GRCh38
rs1042942804 Chr.16: 20427859-20427859 on Build GRCh38
rs780109290 Chr.16: 20427878-20427878 on Build GRCh38
rs141831659 Chr.16: 20427966-20427966 on Build GRCh38
rs753473263 Chr.16: 20427865-20427865 on Build GRCh38
rs114588253 Chr.16: 20428021-20428021 on Build GRCh38
rs889536260 Chr.16: 20428080-20428080 on Build GRCh38
rs779158150 Chr.16: 20427898-20427898 on Build GRCh38
rs559942147 Chr.16: 20428054-20428054 on Build GRCh38
rs200496913 Chr.16: 20427806-20427806 on Build GRCh38
rs868031800 Chr.16: 20428235-20428235 on Build GRCh38
rs994405313 Chr.16: 20428164-20428164 on Build GRCh38
rs769800964 Chr.16: 20427828-20427828 on Build GRCh38
rs566965339 Chr.16: 20427915-20427915 on Build GRCh38
rs961621743 Chr.16: 20427990-20427990 on Build GRCh38
rs529627648 Chr.16: 20427855-20427855 on Build GRCh38
rs781370867 Chr.16: 20427817-20427817 on Build GRCh38
rs956724983 Chr.16: 20428252-20428252 on Build GRCh38
rs12921643 Chr.16: 20427798-20427798 on Build GRCh38
rs960882102 Chr.16: 20428014-20428014 on Build GRCh38
rs1005637956 Chr.16: 20427985-20427985 on Build GRCh38
rs922417432 Chr.16: 20428045-20428045 on Build GRCh38
rs1049458274 Chr.16: 20428062-20428062 on Build GRCh38
rs147838581 Chr.16: 20427869-20427869 on Build GRCh38
rs66989570 Chr.16: 20428227-20428227 on Build GRCh38
rs577268868 Chr.16: 20428037-20428037 on Build GRCh38
rs750264532 Chr.16: 20427874-20427874 on Build GRCh38
rs12922410 Chr.16: 20428038-20428038 on Build GRCh38
rs373914142 Chr.16: 20428200-20428200 on Build GRCh38
rs946696991 Chr.16: 20428024-20428024 on Build GRCh38
rs765040529 Chr.16: 20427871-20427871 on Build GRCh38
rs12921787 Chr.16: 20427826-20427826 on Build GRCh38
rs540436881 Chr.16: 20428144-20428144 on Build GRCh38
rs757062050 Chr.16: 20427804-20427804 on Build GRCh38
rs1052988299 Chr.16: 20428039-20428039 on Build GRCh38
rs933832233 Chr.16: 20428058-20428058 on Build GRCh38
rs11859961 Chr.16: 20427877-20427877 on Build GRCh38
rs1023842751 Chr.16: 20428217-20428217 on Build GRCh38
rs758439647 Chr.16: 20427808-20427808 on Build GRCh38
rs1035175318 Chr.16: 20428010-20428011 on Build GRCh38
rs746204148 Chr.16: 20427906-20427906 on Build GRCh38
rs370567892 Chr.16: 20427885-20427885 on Build GRCh38
rs922530310 Chr.16: 20428255-20428255 on Build GRCh38
rs765889155 Chr.16: 20427891-20427892 on Build GRCh38
rs774731305 Chr.16: 20427842-20427842 on Build GRCh38
rs896854851 Chr.16: 20428129-20428129 on Build GRCh38
rs1028510529 Chr.16: 20427961-20427961 on Build GRCh38
rs376209637 Chr.16: 20427987-20427987 on Build GRCh38
rs11860025 Chr.16: 20428246-20428246 on Build GRCh38
rs763774602 Chr.16: 20427791-20427791 on Build GRCh38
rs551623680 Chr.16: 20427886-20427886 on Build GRCh38
rs190209283 Chr.16: 20427949-20427949 on Build GRCh38
rs749527279 Chr.16: 20427839-20427839 on Build GRCh38
rs904449877 Chr.16: 20427862-20427862 on Build GRCh38
rs368924557 Chr.16: 20428003-20428003 on Build GRCh38
rs771270639 Chr.16: 20428239-20428239 on Build GRCh38
rs555721530 Chr.16: 20428026-20428026 on Build GRCh38
rs763001296 Chr.16: 20427906-20427907 on Build GRCh38
rs750092770 Chr.16: 20427807-20427813 on Build GRCh38
rs376411975 Chr.16: 20428104-20428104 on Build GRCh38

View Full Product Details