Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24827790-24828297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY3
Entrez Gene ID
109
Gene Name
adenylate cyclase 3
Gene Aliases
AC-III, AC3
UniGene
Hs.467898
Gene Chromosome Location
Chr.2: 24819169-24920017 on Build GRCh38
Transcripts
NM_001320613.1 NM_004036.4 XM_006711925.1 XM_011532489.1 XM_011532490.1 XM_011532491.1 XM_011532492.2 XM_011532493.1 XM_011532494.1 XM_011532496.1 XM_017003186.1 XM_017003187.1 XM_017003188.1 XM_017003189.1 XM_017003190.1 XM_017003191.1 XM_017003192.1 XM_017003193.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6157745 c.2331C>T p.L777L Chr.2: 24828003-24828003 on Build GRCh38
1195507 c.2273C>T p.A758V Chr.2: 24828061-24828061 on Build GRCh38
4583399 c.2274C>T p.A758A Chr.2: 24828060-24828060 on Build GRCh38
6594911 c.2391C>T p.P797P Chr.2: 24827943-24827943 on Build GRCh38
3991219 c.2175C>T p.L725L Chr.2: 24828159-24828159 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991059557 Chr.2: 24828069-24828069 on Build GRCh38
rs751356378 Chr.2: 24828051-24828051 on Build GRCh38
rs146269706 Chr.2: 24827852-24827852 on Build GRCh38
rs765173644 Chr.2: 24827962-24827962 on Build GRCh38
rs917820514 Chr.2: 24828121-24828121 on Build GRCh38
rs7566416 Chr.2: 24828108-24828108 on Build GRCh38
rs757402803 Chr.2: 24828172-24828172 on Build GRCh38
rs759311730 Chr.2: 24828139-24828139 on Build GRCh38
rs778849471 Chr.2: 24828086-24828086 on Build GRCh38
rs375943024 Chr.2: 24828106-24828106 on Build GRCh38
rs775904787 Chr.2: 24827945-24827945 on Build GRCh38
rs767110396 Chr.2: 24827885-24827885 on Build GRCh38
rs146981649 Chr.2: 24828085-24828085 on Build GRCh38
rs780543067 Chr.2: 24828207-24828207 on Build GRCh38
rs565303281 Chr.2: 24827889-24827889 on Build GRCh38
rs759846027 Chr.2: 24827880-24827880 on Build GRCh38
rs754015078 Chr.2: 24828070-24828070 on Build GRCh38
rs1045870709 Chr.2: 24827952-24827952 on Build GRCh38
rs746001072 Chr.2: 24828094-24828094 on Build GRCh38
rs140426660 Chr.2: 24827966-24827966 on Build GRCh38
rs374127927 Chr.2: 24828038-24828038 on Build GRCh38
rs997622455 Chr.2: 24828040-24828040 on Build GRCh38
rs929411578 Chr.2: 24828161-24828161 on Build GRCh38
rs747327769 Chr.2: 24828103-24828103 on Build GRCh38
rs775810904 Chr.2: 24828000-24828000 on Build GRCh38
rs532463063 Chr.2: 24827892-24827892 on Build GRCh38
rs763500351 Chr.2: 24827876-24827876 on Build GRCh38
rs752824508 Chr.2: 24827867-24827870 on Build GRCh38
rs752598507 Chr.2: 24828167-24828167 on Build GRCh38
rs559928401 Chr.2: 24828264-24828264 on Build GRCh38
rs902101859 Chr.2: 24828200-24828200 on Build GRCh38
rs75629912 Chr.2: 24828175-24828175 on Build GRCh38
rs373035912 Chr.2: 24827918-24827918 on Build GRCh38
rs750464171 Chr.2: 24828176-24828176 on Build GRCh38
rs897855322 Chr.2: 24827831-24827831 on Build GRCh38
rs542868330 Chr.2: 24827822-24827822 on Build GRCh38
rs902049647 Chr.2: 24828036-24828036 on Build GRCh38
rs766456461 Chr.2: 24828050-24828050 on Build GRCh38
rs766366885 Chr.2: 24828120-24828120 on Build GRCh38
rs759381730 Chr.2: 24828212-24828212 on Build GRCh38
rs767804097 Chr.2: 24827862-24827864 on Build GRCh38
rs896984111 Chr.2: 24828263-24828263 on Build GRCh38
rs770197739 Chr.2: 24827920-24827920 on Build GRCh38
rs543479985 Chr.2: 24827863-24827863 on Build GRCh38
rs369803323 Chr.2: 24827942-24827942 on Build GRCh38
rs772816661 Chr.2: 24828113-24828113 on Build GRCh38
rs973142839 Chr.2: 24827995-24827995 on Build GRCh38
rs754839662 Chr.2: 24828059-24828059 on Build GRCh38
rs773017407 Chr.2: 24828193-24828193 on Build GRCh38
rs79391846 Chr.2: 24828220-24828220 on Build GRCh38
rs771615369 Chr.2: 24828251-24828251 on Build GRCh38
rs757082218 Chr.2: 24828187-24828188 on Build GRCh38
rs991120544 Chr.2: 24827941-24827941 on Build GRCh38
rs952832634 Chr.2: 24828063-24828063 on Build GRCh38
rs766839931 Chr.2: 24827877-24827877 on Build GRCh38
rs767214067 Chr.2: 24828164-24828164 on Build GRCh38
rs185180064 Chr.2: 24827917-24827917 on Build GRCh38
rs558636041 Chr.2: 24828228-24828228 on Build GRCh38
rs761261963 Chr.2: 24827958-24827958 on Build GRCh38
rs200197892 Chr.2: 24828138-24828138 on Build GRCh38
rs776171716 Chr.2: 24828037-24828037 on Build GRCh38
rs560339786 Chr.2: 24827911-24827911 on Build GRCh38
rs934311006 Chr.2: 24827922-24827924 on Build GRCh38
rs777887022 Chr.2: 24827983-24827983 on Build GRCh38
rs755558661 Chr.2: 24827890-24827890 on Build GRCh38
rs541532773 Chr.2: 24828262-24828262 on Build GRCh38
rs78678013 Chr.2: 24827978-24827978 on Build GRCh38
rs148197917 Chr.2: 24828119-24828119 on Build GRCh38
rs759333413 Chr.2: 24827857-24827857 on Build GRCh38
rs780570885 Chr.2: 24828102-24828102 on Build GRCh38
rs142601620 Chr.2: 24828178-24828178 on Build GRCh38
rs150566853 Chr.2: 24828210-24828210 on Build GRCh38
rs761068478 Chr.2: 24828021-24828021 on Build GRCh38
rs757843051 Chr.2: 24828115-24828115 on Build GRCh38
rs1007829744 Chr.2: 24828001-24828001 on Build GRCh38
rs1000633257 Chr.2: 24828245-24828245 on Build GRCh38
rs1045815907 Chr.2: 24827828-24827828 on Build GRCh38
rs778518036 Chr.2: 24828197-24828198 on Build GRCh38
rs750011135 Chr.2: 24828045-24828045 on Build GRCh38
rs538315098 Chr.2: 24828101-24828101 on Build GRCh38
rs764647486 Chr.2: 24828044-24828044 on Build GRCh38
rs749363565 Chr.2: 24827927-24827927 on Build GRCh38
rs774738640 Chr.2: 24827937-24827937 on Build GRCh38
rs952721494 Chr.2: 24828270-24828270 on Build GRCh38
rs557497923 Chr.2: 24828201-24828201 on Build GRCh38
rs758474445 Chr.2: 24827975-24827975 on Build GRCh38
rs201034328 Chr.2: 24827870-24827870 on Build GRCh38
rs753347971 Chr.2: 24827903-24827903 on Build GRCh38
rs779982153 Chr.2: 24827976-24827976 on Build GRCh38
rs779412964 Chr.2: 24827987-24827987 on Build GRCh38
rs527469043 Chr.2: 24828267-24828267 on Build GRCh38
rs2289092 Chr.2: 24828048-24828048 on Build GRCh38
rs755565638 Chr.2: 24828180-24828180 on Build GRCh38
rs772629343 Chr.2: 24827990-24827990 on Build GRCh38
rs780483152 Chr.2: 24828179-24828179 on Build GRCh38
rs943353473 Chr.2: 24828183-24828183 on Build GRCh38
rs777026103 Chr.2: 24827865-24827865 on Build GRCh38
rs377506465 Chr.2: 24827928-24827928 on Build GRCh38
rs372148762 Chr.2: 24827853-24827853 on Build GRCh38
rs764980386 Chr.2: 24827872-24827872 on Build GRCh38
rs371228102 Chr.2: 24828153-24828153 on Build GRCh38
rs778757630 Chr.2: 24827906-24827906 on Build GRCh38
rs773315426 Chr.2: 24827922-24827922 on Build GRCh38
rs368885005 Chr.2: 24828192-24828192 on Build GRCh38
rs758279809 Chr.2: 24827912-24827912 on Build GRCh38
rs537829337 Chr.2: 24827986-24827986 on Build GRCh38
rs533950715 Chr.2: 24828244-24828244 on Build GRCh38
rs1028116101 Chr.2: 24827824-24827824 on Build GRCh38
rs959092323 Chr.2: 24828265-24828265 on Build GRCh38
rs1032492628 Chr.2: 24828248-24828248 on Build GRCh38
rs772433588 Chr.2: 24827943-24827943 on Build GRCh38
rs1008087534 Chr.2: 24827971-24827971 on Build GRCh38
rs757211680 Chr.2: 24827856-24827856 on Build GRCh38
rs777118828 Chr.2: 24828105-24828105 on Build GRCh38
rs576369380 Chr.2: 24827854-24827854 on Build GRCh38
rs748746044 Chr.2: 24828204-24828204 on Build GRCh38
rs145224170 Chr.2: 24828060-24828060 on Build GRCh38
rs190125602 Chr.2: 24827948-24827948 on Build GRCh38
rs749337506 Chr.2: 24827984-24827984 on Build GRCh38
rs970587123 Chr.2: 24828132-24828132 on Build GRCh38
rs768315783 Chr.2: 24828024-24828024 on Build GRCh38
rs372560995 Chr.2: 24827947-24827947 on Build GRCh38
rs373053710 Chr.2: 24827869-24827869 on Build GRCh38
rs756369092 Chr.2: 24827982-24827982 on Build GRCh38
rs919336331 Chr.2: 24827963-24827963 on Build GRCh38
rs139420185 Chr.2: 24827904-24827904 on Build GRCh38
rs774744041 Chr.2: 24827878-24827878 on Build GRCh38

View Full Product Details