Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 6757403-6757897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGPAT5
Entrez Gene ID
55326
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 5
Gene Aliases
1AGPAT5, LPAATE
UniGene
Hs.624002
Gene Chromosome Location
Chr.8: 6708357-6761503 on Build GRCh38
Transcripts
NM_018361.3 XM_011534751.2

SNP Detail

SNP ID Chromosome Location
rs180785046 Chr.8: 6757869-6757869 on Build GRCh38
rs758325300 Chr.8: 6757576-6757576 on Build GRCh38
rs898120322 Chr.8: 6757825-6757825 on Build GRCh38
rs965466660 Chr.8: 6757468-6757468 on Build GRCh38
rs540596356 Chr.8: 6757648-6757648 on Build GRCh38
rs761982955 Chr.8: 6757425-6757425 on Build GRCh38
rs968792421 Chr.8: 6757725-6757725 on Build GRCh38
rs7830784 Chr.8: 6757644-6757644 on Build GRCh38
rs1036781644 Chr.8: 6757795-6757795 on Build GRCh38
rs922455159 Chr.8: 6757503-6757503 on Build GRCh38
rs897028846 Chr.8: 6757598-6757598 on Build GRCh38
rs896912665 Chr.8: 6757802-6757802 on Build GRCh38
rs552865777 Chr.8: 6757465-6757465 on Build GRCh38
rs550282792 Chr.8: 6757813-6757813 on Build GRCh38
rs979346464 Chr.8: 6757728-6757728 on Build GRCh38
rs73509806 Chr.8: 6757729-6757729 on Build GRCh38
rs1005291692 Chr.8: 6757646-6757646 on Build GRCh38
rs1019644979 Chr.8: 6757664-6757664 on Build GRCh38
rs939334249 Chr.8: 6757797-6757797 on Build GRCh38
rs988871969 Chr.8: 6757529-6757529 on Build GRCh38
rs373971203 Chr.8: 6757730-6757730 on Build GRCh38
rs898005567 Chr.8: 6757606-6757606 on Build GRCh38
rs376715740 Chr.8: 6757424-6757424 on Build GRCh38
rs755343253 Chr.8: 6757724-6757724 on Build GRCh38
rs1019580555 Chr.8: 6757464-6757464 on Build GRCh38
rs765271343 Chr.8: 6757516-6757516 on Build GRCh38
rs1017934729 Chr.8: 6757457-6757457 on Build GRCh38
rs896129987 Chr.8: 6757570-6757570 on Build GRCh38
rs976649064 Chr.8: 6757681-6757681 on Build GRCh38
rs775672688 Chr.8: 6757440-6757440 on Build GRCh38
rs1020376968 Chr.8: 6757697-6757697 on Build GRCh38
rs73661471 Chr.8: 6757546-6757546 on Build GRCh38
rs949047602 Chr.8: 6757771-6757771 on Build GRCh38
rs753918900 Chr.8: 6757547-6757547 on Build GRCh38
rs79089126 Chr.8: 6757857-6757857 on Build GRCh38
rs546218716 Chr.8: 6757861-6757861 on Build GRCh38
rs913426759 Chr.8: 6757530-6757530 on Build GRCh38
rs1055209370 Chr.8: 6757551-6757551 on Build GRCh38
rs984234720 Chr.8: 6757442-6757442 on Build GRCh38
rs560269200 Chr.8: 6757678-6757678 on Build GRCh38
rs985870299 Chr.8: 6757455-6757455 on Build GRCh38
rs575336684 Chr.8: 6757625-6757625 on Build GRCh38
rs749139726 Chr.8: 6757638-6757639 on Build GRCh38
rs566988411 Chr.8: 6757829-6757829 on Build GRCh38
rs780138046 Chr.8: 6757587-6757587 on Build GRCh38
rs987973779 Chr.8: 6757748-6757749 on Build GRCh38
rs1045506541 Chr.8: 6757569-6757569 on Build GRCh38
rs926630416 Chr.8: 6757765-6757765 on Build GRCh38
rs927538415 Chr.8: 6757542-6757542 on Build GRCh38
rs954112747 Chr.8: 6757443-6757443 on Build GRCh38
rs979075314 Chr.8: 6757536-6757536 on Build GRCh38
rs964337303 Chr.8: 6757463-6757463 on Build GRCh38
rs976677196 Chr.8: 6757494-6757494 on Build GRCh38
rs947528516 Chr.8: 6757531-6757531 on Build GRCh38
rs1048716544 Chr.8: 6757847-6757847 on Build GRCh38
rs373336856 Chr.8: 6757471-6757471 on Build GRCh38
rs915068312 Chr.8: 6757749-6757749 on Build GRCh38
rs555050716 Chr.8: 6757642-6757642 on Build GRCh38
rs748331349 Chr.8: 6757758-6757758 on Build GRCh38
rs369644951 Chr.8: 6757439-6757439 on Build GRCh38
rs532888683 Chr.8: 6757690-6757690 on Build GRCh38
rs1006780029 Chr.8: 6757658-6757658 on Build GRCh38
rs766541379 Chr.8: 6757488-6757488 on Build GRCh38
rs558761838 Chr.8: 6757588-6757588 on Build GRCh38
rs7830779 Chr.8: 6757637-6757637 on Build GRCh38
rs531747057 Chr.8: 6757754-6757754 on Build GRCh38

View Full Product Details