Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 968377-968793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID3A
Entrez Gene ID
1820
Gene Name
AT-rich interaction domain 3A
Gene Aliases
BRIGHT, DRIL1, DRIL3, E2FBP1
UniGene
Hs.501296
Gene Chromosome Location
Chr.19: 925733-975934 on Build GRCh38
Transcripts
NM_005224.2 XM_005259513.4 XM_005259514.4 XM_017026445.1 XM_017026446.1 XM_017026447.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6225163 c.1507C>T p.R503C Chr.19: 968416-968416 on Build GRCh38
6604647 c.1544G>A p.G515D Chr.19: 968453-968453 on Build GRCh38
1641283 c.1553G>A p.S518N Chr.19: 968462-968462 on Build GRCh38
6604648 c.1579G>A p.G527S Chr.19: 968488-968488 on Build GRCh38
1004338 c.1496-2A>G p.? Chr.19: 968403-968403 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs78850921 Chr.19: 968623-968623 on Build GRCh38
rs917029634 Chr.19: 968594-968594 on Build GRCh38
rs561725111 Chr.19: 968565-968565 on Build GRCh38
rs199783917 Chr.19: 968547-968547 on Build GRCh38
rs958651550 Chr.19: 968744-968744 on Build GRCh38
rs901840793 Chr.19: 968577-968577 on Build GRCh38
rs140797776 Chr.19: 968451-968451 on Build GRCh38
rs761812985 Chr.19: 968514-968514 on Build GRCh38
rs143256319 Chr.19: 968417-968417 on Build GRCh38
rs142537557 Chr.19: 968441-968441 on Build GRCh38
rs565797164 Chr.19: 968609-968609 on Build GRCh38
rs116347167 Chr.19: 968551-968551 on Build GRCh38
rs755814713 Chr.19: 968420-968420 on Build GRCh38
rs1055638644 Chr.19: 968590-968590 on Build GRCh38
rs553005797 Chr.19: 968753-968753 on Build GRCh38
rs371086068 Chr.19: 968548-968548 on Build GRCh38
rs770082664 Chr.19: 968488-968488 on Build GRCh38
rs1035140289 Chr.19: 968701-968701 on Build GRCh38
rs763245378 Chr.19: 968494-968494 on Build GRCh38
rs529223975 Chr.19: 968576-968576 on Build GRCh38
rs367995649 Chr.19: 968523-968523 on Build GRCh38
rs150959744 Chr.19: 968442-968442 on Build GRCh38
rs916851833 Chr.19: 968716-968716 on Build GRCh38
rs867696627 Chr.19: 968675-968675 on Build GRCh38
rs777065341 Chr.19: 968484-968484 on Build GRCh38
rs747585677 Chr.19: 968474-968474 on Build GRCh38
rs755902919 Chr.19: 968524-968524 on Build GRCh38
rs773529270 Chr.19: 968754-968754 on Build GRCh38
rs113851216 Chr.19: 968714-968714 on Build GRCh38
rs547405769 Chr.19: 968598-968598 on Build GRCh38
rs530706758 Chr.19: 968553-968553 on Build GRCh38
rs781106095 Chr.19: 968416-968416 on Build GRCh38
rs549292986 Chr.19: 968564-968564 on Build GRCh38
rs373835356 Chr.19: 968522-968522 on Build GRCh38
rs978670335 Chr.19: 968561-968561 on Build GRCh38
rs925573573 Chr.19: 968739-968739 on Build GRCh38
rs117438574 Chr.19: 968691-968691 on Build GRCh38
rs775890384 Chr.19: 968493-968493 on Build GRCh38
rs537786817 Chr.19: 968410-968410 on Build GRCh38
rs781508980 Chr.19: 968550-968550 on Build GRCh38
rs770264610 Chr.19: 968708-968708 on Build GRCh38
rs984928406 Chr.19: 968755-968755 on Build GRCh38
rs751596134 Chr.19: 968409-968409 on Build GRCh38
rs779802062 Chr.19: 968527-968527 on Build GRCh38
rs1039061266 Chr.19: 968643-968643 on Build GRCh38
rs370801930 Chr.19: 968510-968510 on Build GRCh38
rs182218059 Chr.19: 968684-968684 on Build GRCh38
rs748908471 Chr.19: 968427-968427 on Build GRCh38
rs1047148562 Chr.19: 968616-968616 on Build GRCh38
rs755376907 Chr.19: 968745-968745 on Build GRCh38
rs7246309 Chr.19: 968669-968669 on Build GRCh38
rs371807095 Chr.19: 968539-968539 on Build GRCh38
rs533472260 Chr.19: 968613-968613 on Build GRCh38
rs940743548 Chr.19: 968556-968556 on Build GRCh38
rs779533318 Chr.19: 968422-968422 on Build GRCh38
rs759767653 Chr.19: 968487-968487 on Build GRCh38
rs534558878 Chr.19: 968711-968711 on Build GRCh38
rs753551629 Chr.19: 968538-968538 on Build GRCh38
rs1022065150 Chr.19: 968460-968460 on Build GRCh38
rs1056321757 Chr.19: 968437-968437 on Build GRCh38
rs998535322 Chr.19: 968608-968608 on Build GRCh38
rs964599951 Chr.19: 968518-968518 on Build GRCh38
rs1025651534 Chr.19: 968453-968453 on Build GRCh38
rs762102678 Chr.19: 968724-968726 on Build GRCh38
rs146132253 Chr.19: 968475-968475 on Build GRCh38
rs1022096410 Chr.19: 968676-968676 on Build GRCh38
rs760788965 Chr.19: 968513-968514 on Build GRCh38
rs773386590 Chr.19: 968443-968451 on Build GRCh38

View Full Product Details