Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 125841447-125841952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCCIP
Entrez Gene ID
56647
Gene Name
BRCA2 and CDKN1A interacting protein
Gene Aliases
TOK-1, TOK1
UniGene
Hs.370292
Gene Chromosome Location
Chr.10: 125823535-125853695 on Build GRCh38
Transcripts
NM_016567.3 NM_078469.2 NM_018180.2 XM_017016404.1 XM_017016405.1

Gene Symbol
DHX32
Entrez Gene ID
55760
Gene Name
DEAH-box helicase 32 (putative)
Gene Aliases
DDX32, DHLP1
UniGene
Hs.370292
Gene Chromosome Location
Chr.10: 125836337-125896483 on Build GRCh38
Transcripts
NM_016567.3 NM_078469.2 NM_018180.2 XM_017016404.1 XM_017016405.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1741442 c.1510G>A p.D504N Chr.10: 125841776-125841776 on Build GRCh38
4012521 c.1529C>T p.A510V Chr.10: 125841757-125841757 on Build GRCh38
6045537 c.1359A>C p.E453D Chr.10: 125841927-125841927 on Build GRCh38
4526180 c.1378G>A p.E460K Chr.10: 125841908-125841908 on Build GRCh38
6014147 c.1440C>T p.I480I Chr.10: 125841846-125841846 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747820971 Chr.10: 125841911-125841911 on Build GRCh38
rs984967202 Chr.10: 125841799-125841799 on Build GRCh38
rs748857701 Chr.10: 125841776-125841776 on Build GRCh38
rs774237813 Chr.10: 125841695-125841695 on Build GRCh38
rs774523614 Chr.10: 125841849-125841849 on Build GRCh38
rs923294385 Chr.10: 125841844-125841844 on Build GRCh38
rs758252946 Chr.10: 125841913-125841913 on Build GRCh38
rs746110433 Chr.10: 125841710-125841711 on Build GRCh38
rs751987538 Chr.10: 125841757-125841757 on Build GRCh38
rs946594750 Chr.10: 125841905-125841905 on Build GRCh38
rs987806344 Chr.10: 125841474-125841474 on Build GRCh38
rs1055792808 Chr.10: 125841680-125841680 on Build GRCh38
rs928142683 Chr.10: 125841636-125841636 on Build GRCh38
rs760541218 Chr.10: 125841551-125841553 on Build GRCh38
rs565496653 Chr.10: 125841608-125841608 on Build GRCh38
rs756666817 Chr.10: 125841869-125841869 on Build GRCh38
rs775182055 Chr.10: 125841792-125841796 on Build GRCh38
rs1052814050 Chr.10: 125841899-125841899 on Build GRCh38
rs1037406849 Chr.10: 125841724-125841724 on Build GRCh38
rs778520434 Chr.10: 125841793-125841793 on Build GRCh38
rs754110250 Chr.10: 125841903-125841903 on Build GRCh38
rs779600996 Chr.10: 125841770-125841770 on Build GRCh38
rs764401766 Chr.10: 125841848-125841848 on Build GRCh38
rs898940891 Chr.10: 125841746-125841746 on Build GRCh38
rs771545282 Chr.10: 125841535-125841538 on Build GRCh38
rs943468181 Chr.10: 125841825-125841825 on Build GRCh38
rs768381279 Chr.10: 125841785-125841785 on Build GRCh38
rs767938640 Chr.10: 125841861-125841861 on Build GRCh38
rs934676983 Chr.10: 125841898-125841898 on Build GRCh38
rs769994781 Chr.10: 125841716-125841718 on Build GRCh38
rs964708151 Chr.10: 125841527-125841527 on Build GRCh38
rs765831395 Chr.10: 125841871-125841871 on Build GRCh38
rs563230784 Chr.10: 125841502-125841505 on Build GRCh38
rs905003463 Chr.10: 125841923-125841923 on Build GRCh38
rs551136425 Chr.10: 125841686-125841686 on Build GRCh38
rs755621661 Chr.10: 125841769-125841769 on Build GRCh38
rs532560567 Chr.10: 125841681-125841681 on Build GRCh38
rs773275564 Chr.10: 125841806-125841806 on Build GRCh38
rs989560306 Chr.10: 125841590-125841590 on Build GRCh38
rs750921125 Chr.10: 125841866-125841866 on Build GRCh38
rs972751530 Chr.10: 125841736-125841736 on Build GRCh38
rs939592831 Chr.10: 125841650-125841650 on Build GRCh38
rs753407497 Chr.10: 125841893-125841893 on Build GRCh38
rs1022049315 Chr.10: 125841511-125841511 on Build GRCh38
rs769971747 Chr.10: 125841827-125841827 on Build GRCh38
rs759691622 Chr.10: 125841707-125841707 on Build GRCh38
rs749871159 Chr.10: 125841762-125841762 on Build GRCh38
rs762757159 Chr.10: 125841837-125841838 on Build GRCh38
rs752953061 Chr.10: 125841743-125841743 on Build GRCh38
rs370172603 Chr.10: 125841697-125841697 on Build GRCh38
rs1001935200 Chr.10: 125841683-125841683 on Build GRCh38
rs541724640 Chr.10: 125841512-125841512 on Build GRCh38
rs778629239 Chr.10: 125841909-125841909 on Build GRCh38
rs3208565 Chr.10: 125841756-125841756 on Build GRCh38
rs536342432 Chr.10: 125841758-125841758 on Build GRCh38
rs372817769 Chr.10: 125841704-125841704 on Build GRCh38
rs1017977519 Chr.10: 125841884-125841884 on Build GRCh38
rs111767995 Chr.10: 125841842-125841842 on Build GRCh38
rs574953919 Chr.10: 125841798-125841798 on Build GRCh38
rs374870912 Chr.10: 125841813-125841813 on Build GRCh38
rs915360892 Chr.10: 125841599-125841599 on Build GRCh38
rs781675809 Chr.10: 125841759-125841759 on Build GRCh38
rs765477262 Chr.10: 125841726-125841726 on Build GRCh38
rs763285087 Chr.10: 125841754-125841754 on Build GRCh38
rs770787508 Chr.10: 125841600-125841603 on Build GRCh38
rs1042604442 Chr.10: 125841635-125841635 on Build GRCh38
rs137998421 Chr.10: 125841847-125841847 on Build GRCh38

View Full Product Details