Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 76826402-76826947 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC74A
Entrez Gene ID
145497
Gene Name
leucine rich repeat containing 74A
Gene Aliases
C14orf166B, LRRC74
UniGene
Hs.147276
Gene Chromosome Location
Chr.14: 76826370-76870302 on Build GRCh38
Transcripts
NM_194287.2 XM_011536479.1 XM_011536483.1 XM_011536487.1 XM_017021032.1 XM_017021033.1 XM_017021034.1 XM_017021035.1 XM_017021036.1 XM_017021037.1 XM_017021038.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4649885 c.1A>T p.M1L Chr.14: 76826496-76826496 on Build GRCh38
4052558 c.45C>A p.C15* Chr.14: 76826540-76826540 on Build GRCh38
3401479 c.20C>G p.S7* Chr.14: 76826515-76826515 on Build GRCh38
4927783 c.19T>C p.S7P Chr.14: 76826514-76826514 on Build GRCh38
5580388 c.34C>T p.P12S Chr.14: 76826529-76826529 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781522834 Chr.14: 76826734-76826734 on Build GRCh38
rs1016091302 Chr.14: 76826724-76826724 on Build GRCh38
rs937485572 Chr.14: 76826815-76826815 on Build GRCh38
rs754764975 Chr.14: 76826657-76826657 on Build GRCh38
rs767076001 Chr.14: 76826637-76826637 on Build GRCh38
rs777679049 Chr.14: 76826683-76826683 on Build GRCh38
rs1002822573 Chr.14: 76826796-76826796 on Build GRCh38
rs374629334 Chr.14: 76826568-76826568 on Build GRCh38
rs377665097 Chr.14: 76826456-76826456 on Build GRCh38
rs1047388497 Chr.14: 76826900-76826900 on Build GRCh38
rs761465229 Chr.14: 76826631-76826631 on Build GRCh38
rs545249817 Chr.14: 76826567-76826567 on Build GRCh38
rs548915561 Chr.14: 76826853-76826853 on Build GRCh38
rs755948145 Chr.14: 76826554-76826554 on Build GRCh38
rs371403927 Chr.14: 76826559-76826559 on Build GRCh38
rs753616357 Chr.14: 76826655-76826655 on Build GRCh38
rs977549203 Chr.14: 76826679-76826679 on Build GRCh38
rs958952001 Chr.14: 76826840-76826840 on Build GRCh38
rs758480102 Chr.14: 76826672-76826672 on Build GRCh38
rs188303516 Chr.14: 76826426-76826426 on Build GRCh38
rs766836556 Chr.14: 76826625-76826626 on Build GRCh38
rs775216415 Chr.14: 76826502-76826502 on Build GRCh38
rs924322433 Chr.14: 76826692-76826692 on Build GRCh38
rs893914971 Chr.14: 76826565-76826566 on Build GRCh38
rs747577358 Chr.14: 76826684-76826684 on Build GRCh38
rs748205035 Chr.14: 76826482-76826482 on Build GRCh38
rs776808323 Chr.14: 76826590-76826590 on Build GRCh38
rs754120975 Chr.14: 76826560-76826560 on Build GRCh38
rs898964537 Chr.14: 76826766-76826766 on Build GRCh38
rs1006422890 Chr.14: 76826689-76826689 on Build GRCh38
rs964655639 Chr.14: 76826453-76826453 on Build GRCh38
rs992968927 Chr.14: 76826844-76826844 on Build GRCh38
rs746217752 Chr.14: 76826599-76826599 on Build GRCh38
rs559201692 Chr.14: 76826858-76826858 on Build GRCh38
rs917464556 Chr.14: 76826862-76826862 on Build GRCh38
rs930230680 Chr.14: 76826876-76826876 on Build GRCh38
rs778916208 Chr.14: 76826477-76826477 on Build GRCh38
rs763605653 Chr.14: 76826599-76826600 on Build GRCh38
rs757945405 Chr.14: 76826563-76826563 on Build GRCh38
rs759982374 Chr.14: 76826654-76826654 on Build GRCh38
rs774157975 Chr.14: 76826747-76826747 on Build GRCh38
rs781699597 Chr.14: 76826709-76826709 on Build GRCh38
rs761372919 Chr.14: 76826533-76826533 on Build GRCh38
rs751213862 Chr.14: 76826696-76826696 on Build GRCh38
rs1030231279 Chr.14: 76826744-76826744 on Build GRCh38
rs771738701 Chr.14: 76826767-76826767 on Build GRCh38
rs771145083 Chr.14: 76826497-76826497 on Build GRCh38
rs886149739 Chr.14: 76826688-76826688 on Build GRCh38
rs943226601 Chr.14: 76826763-76826763 on Build GRCh38
rs768785566 Chr.14: 76826466-76826466 on Build GRCh38
rs746299042 Chr.14: 76826725-76826725 on Build GRCh38
rs887138388 Chr.14: 76826791-76826791 on Build GRCh38
rs926120 Chr.14: 76826649-76826649 on Build GRCh38
rs774922810 Chr.14: 76826635-76826636 on Build GRCh38
rs1035892190 Chr.14: 76826588-76826588 on Build GRCh38
rs752860672 Chr.14: 76826667-76826667 on Build GRCh38
rs762477739 Chr.14: 76826513-76826513 on Build GRCh38
rs749412696 Chr.14: 76826458-76826458 on Build GRCh38
rs768460355 Chr.14: 76826616-76826616 on Build GRCh38
rs530028648 Chr.14: 76826765-76826765 on Build GRCh38
rs1051660661 Chr.14: 76826429-76826429 on Build GRCh38
rs573523748 Chr.14: 76826531-76826531 on Build GRCh38
rs991671309 Chr.14: 76826624-76826624 on Build GRCh38
rs72737548 Chr.14: 76826729-76826729 on Build GRCh38
rs765503900 Chr.14: 76826662-76826662 on Build GRCh38
rs772609138 Chr.14: 76826485-76826485 on Build GRCh38
rs202183792 Chr.14: 76826530-76826530 on Build GRCh38
rs1047573183 Chr.14: 76826777-76826777 on Build GRCh38
rs950532069 Chr.14: 76826433-76826433 on Build GRCh38
rs544648090 Chr.14: 76826699-76826699 on Build GRCh38
rs767625989 Chr.14: 76826544-76826544 on Build GRCh38
rs1035944248 Chr.14: 76826814-76826814 on Build GRCh38
rs927484926 Chr.14: 76826642-76826642 on Build GRCh38
rs367555116 Chr.14: 76826617-76826617 on Build GRCh38
rs79863016 Chr.14: 76826650-76826650 on Build GRCh38
rs200362790 Chr.14: 76826496-76826496 on Build GRCh38
rs755244062 Chr.14: 76826562-76826562 on Build GRCh38
rs981683540 Chr.14: 76826571-76826571 on Build GRCh38
rs528326698 Chr.14: 76826878-76826878 on Build GRCh38
rs771362058 Chr.14: 76826583-76826583 on Build GRCh38
rs1016513747 Chr.14: 76826435-76826435 on Build GRCh38
rs927463977 Chr.14: 76826797-76826797 on Build GRCh38
rs573631480 Chr.14: 76826491-76826491 on Build GRCh38
rs758461408 Chr.14: 76826577-76826577 on Build GRCh38
rs773206185 Chr.14: 76826646-76826646 on Build GRCh38
rs537112917 Chr.14: 76826434-76826434 on Build GRCh38
rs772548127 Chr.14: 76826770-76826770 on Build GRCh38
rs750312780 Chr.14: 76826547-76826547 on Build GRCh38
rs747410665 Chr.14: 76826581-76826581 on Build GRCh38

View Full Product Details