Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37180852-37181410 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QTNF6
Entrez Gene ID
114904
Gene Name
C1q and tumor necrosis factor related protein 6
Gene Aliases
CTFP6, CTRP6, ZACRP6
UniGene
Hs.22011
Gene Chromosome Location
Chr.22: 37180166-37199400 on Build GRCh38
Transcripts
NM_031910.3 NM_182486.1 XM_011529857.1 XM_017028569.1 XM_017028570.1 XM_017028571.1 XM_017028572.1 XM_017028573.1 XM_017028574.1

SNP Detail

SNP ID Chromosome Location
rs951965085 Chr.22: 37181100-37181100 on Build GRCh38
rs1041956084 Chr.22: 37181370-37181370 on Build GRCh38
rs5756538 Chr.22: 37181103-37181103 on Build GRCh38
rs952465433 Chr.22: 37180917-37180917 on Build GRCh38
rs1034731242 Chr.22: 37181230-37181230 on Build GRCh38
rs952066633 Chr.22: 37181261-37181261 on Build GRCh38
rs922133814 Chr.22: 37181137-37181137 on Build GRCh38
rs754296258 Chr.22: 37180878-37180878 on Build GRCh38
rs552409285 Chr.22: 37181063-37181063 on Build GRCh38
rs755827426 Chr.22: 37181111-37181111 on Build GRCh38
rs983372910 Chr.22: 37181269-37181269 on Build GRCh38
rs6000597 Chr.22: 37181298-37181298 on Build GRCh38
rs12169573 Chr.22: 37181361-37181361 on Build GRCh38
rs922018789 Chr.22: 37181333-37181333 on Build GRCh38
rs944800110 Chr.22: 37181121-37181121 on Build GRCh38
rs1003219628 Chr.22: 37181028-37181028 on Build GRCh38
rs780084575 Chr.22: 37181082-37181082 on Build GRCh38
rs925820788 Chr.22: 37181273-37181273 on Build GRCh38
rs966451582 Chr.22: 37181027-37181027 on Build GRCh38
rs867394149 Chr.22: 37181179-37181179 on Build GRCh38
rs5756539 Chr.22: 37181105-37181105 on Build GRCh38
rs117552370 Chr.22: 37180909-37180909 on Build GRCh38
rs894711827 Chr.22: 37180883-37180883 on Build GRCh38
rs117040702 Chr.22: 37181012-37181012 on Build GRCh38
rs373603847 Chr.22: 37181189-37181189 on Build GRCh38
rs867233600 Chr.22: 37181357-37181357 on Build GRCh38
rs990792743 Chr.22: 37181079-37181079 on Build GRCh38
rs6000596 Chr.22: 37181203-37181203 on Build GRCh38
rs557830831 Chr.22: 37181279-37181279 on Build GRCh38
rs545969758 Chr.22: 37180998-37180998 on Build GRCh38
rs770602223 Chr.22: 37181330-37181330 on Build GRCh38
rs893505477 Chr.22: 37180976-37180976 on Build GRCh38
rs56271867 Chr.22: 37181214-37181214 on Build GRCh38
rs976144895 Chr.22: 37181136-37181136 on Build GRCh38
rs184455066 Chr.22: 37180939-37180939 on Build GRCh38
rs946320991 Chr.22: 37181363-37181363 on Build GRCh38
rs1040575277 Chr.22: 37180904-37180904 on Build GRCh38
rs539587324 Chr.22: 37181249-37181249 on Build GRCh38
rs990742656 Chr.22: 37181344-37181344 on Build GRCh38
rs764665676 Chr.22: 37180874-37180874 on Build GRCh38
rs757854101 Chr.22: 37181167-37181167 on Build GRCh38
rs1003271087 Chr.22: 37181220-37181220 on Build GRCh38
rs950114766 Chr.22: 37181350-37181350 on Build GRCh38
rs967290237 Chr.22: 37181219-37181219 on Build GRCh38
rs749212452 Chr.22: 37181312-37181312 on Build GRCh38
rs560013242 Chr.22: 37181040-37181040 on Build GRCh38
rs186098949 Chr.22: 37181180-37181180 on Build GRCh38
rs1049288949 Chr.22: 37181125-37181125 on Build GRCh38
rs944747800 Chr.22: 37180903-37180903 on Build GRCh38
rs895018754 Chr.22: 37181244-37181244 on Build GRCh38
rs566052901 Chr.22: 37181247-37181247 on Build GRCh38
rs932128413 Chr.22: 37181143-37181143 on Build GRCh38
rs900658043 Chr.22: 37180921-37180921 on Build GRCh38
rs1055789599 Chr.22: 37181062-37181062 on Build GRCh38
rs957447290 Chr.22: 37181290-37181290 on Build GRCh38
rs922426351 Chr.22: 37180992-37180992 on Build GRCh38
rs549834109 Chr.22: 37181130-37181130 on Build GRCh38
rs898005851 Chr.22: 37180904-37180904 on Build GRCh38
rs188832908 Chr.22: 37180945-37180945 on Build GRCh38
rs376914283 Chr.22: 37181299-37181299 on Build GRCh38
rs976198659 Chr.22: 37181303-37181303 on Build GRCh38
rs181892625 Chr.22: 37181055-37181055 on Build GRCh38
rs915886996 Chr.22: 37181335-37181335 on Build GRCh38
rs181195839 Chr.22: 37180936-37180936 on Build GRCh38
rs1025419152 Chr.22: 37180913-37180913 on Build GRCh38
rs914718554 Chr.22: 37181362-37181362 on Build GRCh38
rs9622563 Chr.22: 37181106-37181106 on Build GRCh38
rs977077941 Chr.22: 37180983-37180983 on Build GRCh38
rs1049256503 Chr.22: 37181368-37181368 on Build GRCh38
rs963992128 Chr.22: 37180953-37180953 on Build GRCh38
rs138548100 Chr.22: 37181336-37181336 on Build GRCh38
rs907961938 Chr.22: 37180891-37180891 on Build GRCh38

View Full Product Details