Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 29091230-29091804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP3K7CL
Entrez Gene ID
56911
Gene Name
MAP3K7 C-terminal like
Gene Aliases
C21orf7, HC21ORF7, TAK1L, TAKL, TAKL-1, TAKL-2, TAKL-4
UniGene
Hs.222802
Gene Chromosome Location
Chr.21: 29077471-29175889 on Build GRCh38
Transcripts
NM_001286617.1 NM_001286618.1 NM_001286622.1 NM_001286623.1 NM_001286634.1 NM_020152.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3396394 c.74G>C p.R25P Chr.21: 29091517-29091517 on Build GRCh38
3396395 c.74G>C p.R25P Chr.21: 29091517-29091517 on Build GRCh38
6371498 c.174C>A p.H58Q Chr.21: 29091708-29091708 on Build GRCh38
5701788 c.219_220GG>TT p.L73_D74>FY Chr.21: 29091753-29091754 on Build GRCh38
5701789 c.219_220GG>TT p.L73_D74>FY Chr.21: 29091753-29091754 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756387910 Chr.21: 29091673-29091673 on Build GRCh38
rs774493470 Chr.21: 29091706-29091706 on Build GRCh38
rs566825852 Chr.21: 29091653-29091654 on Build GRCh38
rs565664997 Chr.21: 29091618-29091618 on Build GRCh38
rs762219439 Chr.21: 29091477-29091477 on Build GRCh38
rs372992693 Chr.21: 29091475-29091475 on Build GRCh38
rs79182116 Chr.21: 29091328-29091328 on Build GRCh38
rs762330122 Chr.21: 29091590-29091590 on Build GRCh38
rs182989054 Chr.21: 29091764-29091764 on Build GRCh38
rs985284592 Chr.21: 29091517-29091517 on Build GRCh38
rs1020986024 Chr.21: 29091451-29091451 on Build GRCh38
rs770976426 Chr.21: 29091696-29091696 on Build GRCh38
rs1036726659 Chr.21: 29091274-29091274 on Build GRCh38
rs530244181 Chr.21: 29091291-29091291 on Build GRCh38
rs760207629 Chr.21: 29091740-29091740 on Build GRCh38
rs1051519861 Chr.21: 29091261-29091261 on Build GRCh38
rs8131128 Chr.21: 29091304-29091304 on Build GRCh38
rs563349236 Chr.21: 29091486-29091486 on Build GRCh38
rs774886661 Chr.21: 29091507-29091507 on Build GRCh38
rs940580502 Chr.21: 29091269-29091269 on Build GRCh38
rs1039966732 Chr.21: 29091316-29091316 on Build GRCh38
rs995634309 Chr.21: 29091334-29091334 on Build GRCh38
rs917855867 Chr.21: 29091593-29091593 on Build GRCh38
rs886202665 Chr.21: 29091372-29091372 on Build GRCh38
rs893295689 Chr.21: 29091419-29091419 on Build GRCh38
rs571100586 Chr.21: 29091680-29091680 on Build GRCh38
rs1034238142 Chr.21: 29091690-29091690 on Build GRCh38
rs775091641 Chr.21: 29091734-29091734 on Build GRCh38
rs745486791 Chr.21: 29091654-29091654 on Build GRCh38
rs967846227 Chr.21: 29091478-29091479 on Build GRCh38
rs760915613 Chr.21: 29091440-29091440 on Build GRCh38
rs191243964 Chr.21: 29091695-29091695 on Build GRCh38
rs971991189 Chr.21: 29091576-29091576 on Build GRCh38
rs150407882 Chr.21: 29091724-29091724 on Build GRCh38
rs909048357 Chr.21: 29091255-29091255 on Build GRCh38
rs904926247 Chr.21: 29091327-29091327 on Build GRCh38
rs1028960623 Chr.21: 29091719-29091719 on Build GRCh38
rs766673568 Chr.21: 29091641-29091641 on Build GRCh38
rs758836784 Chr.21: 29091611-29091611 on Build GRCh38
rs940553472 Chr.21: 29091564-29091564 on Build GRCh38
rs2832168 Chr.21: 29091374-29091374 on Build GRCh38
rs768748833 Chr.21: 29091461-29091461 on Build GRCh38
rs548106892 Chr.21: 29091621-29091621 on Build GRCh38
rs529000847 Chr.21: 29091512-29091512 on Build GRCh38
rs753957461 Chr.21: 29091653-29091653 on Build GRCh38
rs533140481 Chr.21: 29091427-29091427 on Build GRCh38
rs909045296 Chr.21: 29091535-29091535 on Build GRCh38
rs957154572 Chr.21: 29091466-29091466 on Build GRCh38
rs1044680204 Chr.21: 29091604-29091604 on Build GRCh38
rs572138711 Chr.21: 29091312-29091312 on Build GRCh38
rs1010847851 Chr.21: 29091631-29091631 on Build GRCh38
rs1045521448 Chr.21: 29091669-29091669 on Build GRCh38
rs186377134 Chr.21: 29091620-29091620 on Build GRCh38
rs957259729 Chr.21: 29091694-29091694 on Build GRCh38
rs987255915 Chr.21: 29091751-29091751 on Build GRCh38
rs989993585 Chr.21: 29091471-29091472 on Build GRCh38
rs763814375 Chr.21: 29091775-29091775 on Build GRCh38
rs750833790 Chr.21: 29091563-29091563 on Build GRCh38
rs181650933 Chr.21: 29091597-29091597 on Build GRCh38
rs748269074 Chr.21: 29091637-29091637 on Build GRCh38
rs76218876 Chr.21: 29091275-29091275 on Build GRCh38
rs1027337723 Chr.21: 29091368-29091368 on Build GRCh38
rs986943461 Chr.21: 29091502-29091502 on Build GRCh38
rs781452941 Chr.21: 29091630-29091630 on Build GRCh38
rs773567099 Chr.21: 29091710-29091710 on Build GRCh38
rs140863907 Chr.21: 29091683-29091683 on Build GRCh38

View Full Product Details