Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 179852901-179853446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf45
Entrez Gene ID
51149
Gene Name
chromosome 5 open reading frame 45
Gene Aliases
-
UniGene
Hs.741169
Gene Chromosome Location
Chr.5: 179837266-179858840 on Build GRCh38
Transcripts
NM_001017987.2 NM_016175.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4842344 c.173-7C>G p.? Chr.5: 179853283-179853283 on Build GRCh38
6616319 c.122delT p.L41fs*19 Chr.5: 179853382-179853382 on Build GRCh38
6616320 c.168delT p.F56fs*46 Chr.5: 179853382-179853382 on Build GRCh38
4406982 c.112C>T p.Q38* Chr.5: 179853392-179853392 on Build GRCh38
6616327 c.169G>T p.A57S Chr.5: 179853381-179853381 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs938304650 Chr.5: 179853077-179853077 on Build GRCh38
rs899340647 Chr.5: 179853015-179853015 on Build GRCh38
rs920834825 Chr.5: 179853377-179853377 on Build GRCh38
rs370491827 Chr.5: 179853281-179853281 on Build GRCh38
rs1046810898 Chr.5: 179852933-179852933 on Build GRCh38
rs756608240 Chr.5: 179853383-179853383 on Build GRCh38
rs966369310 Chr.5: 179853161-179853161 on Build GRCh38
rs949393663 Chr.5: 179852927-179852927 on Build GRCh38
rs867147285 Chr.5: 179852942-179852942 on Build GRCh38
rs772138448 Chr.5: 179853327-179853327 on Build GRCh38
rs774696814 Chr.5: 179853211-179853211 on Build GRCh38
rs775083784 Chr.5: 179852988-179852989 on Build GRCh38
rs996274208 Chr.5: 179853017-179853017 on Build GRCh38
rs1042120435 Chr.5: 179853135-179853135 on Build GRCh38
rs1028060093 Chr.5: 179853046-179853046 on Build GRCh38
rs745898421 Chr.5: 179853419-179853419 on Build GRCh38
rs929285726 Chr.5: 179853038-179853038 on Build GRCh38
rs554478180 Chr.5: 179853334-179853334 on Build GRCh38
rs775209562 Chr.5: 179853231-179853231 on Build GRCh38
rs1032236658 Chr.5: 179853247-179853247 on Build GRCh38
rs1019692496 Chr.5: 179853127-179853127 on Build GRCh38
rs757650592 Chr.5: 179853408-179853408 on Build GRCh38
rs990637897 Chr.5: 179853306-179853306 on Build GRCh38
rs269462 Chr.5: 179852949-179852949 on Build GRCh38
rs748997047 Chr.5: 179853280-179853280 on Build GRCh38
rs773725615 Chr.5: 179853283-179853284 on Build GRCh38
rs756512181 Chr.5: 179853286-179853286 on Build GRCh38
rs770897667 Chr.5: 179853210-179853210 on Build GRCh38
rs1650893 Chr.5: 179853379-179853379 on Build GRCh38
rs1007934455 Chr.5: 179853062-179853062 on Build GRCh38
rs748676157 Chr.5: 179853150-179853150 on Build GRCh38
rs552202655 Chr.5: 179853168-179853168 on Build GRCh38
rs568825732 Chr.5: 179852959-179852959 on Build GRCh38
rs112867427 Chr.5: 179853392-179853392 on Build GRCh38
rs778468372 Chr.5: 179853287-179853287 on Build GRCh38
rs188462274 Chr.5: 179853006-179853006 on Build GRCh38
rs753745249 Chr.5: 179853262-179853262 on Build GRCh38
rs748480330 Chr.5: 179853285-179853285 on Build GRCh38
rs57446717 Chr.5: 179853345-179853345 on Build GRCh38
rs766187518 Chr.5: 179853188-179853188 on Build GRCh38
rs532027222 Chr.5: 179853050-179853050 on Build GRCh38
rs764647781 Chr.5: 179853264-179853264 on Build GRCh38
rs766165449 Chr.5: 179853374-179853374 on Build GRCh38
rs946882295 Chr.5: 179853122-179853122 on Build GRCh38
rs767352215 Chr.5: 179853224-179853224 on Build GRCh38
rs757928523 Chr.5: 179853274-179853274 on Build GRCh38
rs35200590 Chr.5: 179853120-179853121 on Build GRCh38
rs778282264 Chr.5: 179853386-179853386 on Build GRCh38
rs963414440 Chr.5: 179852950-179852950 on Build GRCh38
rs774266403 Chr.5: 179853301-179853302 on Build GRCh38
rs759676113 Chr.5: 179853216-179853216 on Build GRCh38
rs750093441 Chr.5: 179853270-179853270 on Build GRCh38
rs766956244 Chr.5: 179852943-179852943 on Build GRCh38
rs916689576 Chr.5: 179853314-179853314 on Build GRCh38
rs758601317 Chr.5: 179853245-179853245 on Build GRCh38
rs575281113 Chr.5: 179853318-179853318 on Build GRCh38
rs556937992 Chr.5: 179853290-179853290 on Build GRCh38
rs982530466 Chr.5: 179853051-179853051 on Build GRCh38
rs753694573 Chr.5: 179853191-179853191 on Build GRCh38
rs902270472 Chr.5: 179853166-179853166 on Build GRCh38
rs915441161 Chr.5: 179853109-179853109 on Build GRCh38
rs568641724 Chr.5: 179853196-179853196 on Build GRCh38
rs778795993 Chr.5: 179853307-179853307 on Build GRCh38
rs779074536 Chr.5: 179853413-179853413 on Build GRCh38
rs760629285 Chr.5: 179853253-179853253 on Build GRCh38
rs762695729 Chr.5: 179853346-179853346 on Build GRCh38
rs538571801 Chr.5: 179853278-179853281 on Build GRCh38
rs761443950 Chr.5: 179853364-179853365 on Build GRCh38
rs182824285 Chr.5: 179853263-179853263 on Build GRCh38
rs771112890 Chr.5: 179853301-179853301 on Build GRCh38
rs1055281720 Chr.5: 179853096-179853096 on Build GRCh38
rs190351989 Chr.5: 179853107-179853107 on Build GRCh38
rs200458264 Chr.5: 179853343-179853343 on Build GRCh38
rs775296122 Chr.5: 179853328-179853328 on Build GRCh38
rs529713110 Chr.5: 179852994-179852994 on Build GRCh38
rs763979477 Chr.5: 179853261-179853261 on Build GRCh38
rs773453254 Chr.5: 179853192-179853192 on Build GRCh38
rs567935970 Chr.5: 179853086-179853086 on Build GRCh38
rs760504435 Chr.5: 179853332-179853332 on Build GRCh38
rs1038374686 Chr.5: 179853366-179853366 on Build GRCh38
rs76805955 Chr.5: 179853333-179853333 on Build GRCh38
rs761588716 Chr.5: 179853337-179853337 on Build GRCh38

View Full Product Details