Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 87158011-87158559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf170
Entrez Gene ID
401535
Gene Name
chromosome 9 open reading frame 170
Gene Aliases
-
UniGene
Hs.657740
Gene Chromosome Location
Chr.9: 87148644-87159726 on Build GRCh38
Transcripts
NM_001001709.2

SNP Detail

SNP ID Chromosome Location
rs772377304 Chr.9: 87158291-87158291 on Build GRCh38
rs903145089 Chr.9: 87158455-87158455 on Build GRCh38
rs753034320 Chr.9: 87158225-87158226 on Build GRCh38
rs895288445 Chr.9: 87158500-87158500 on Build GRCh38
rs748269409 Chr.9: 87158180-87158180 on Build GRCh38
rs915030874 Chr.9: 87158047-87158047 on Build GRCh38
rs911184046 Chr.9: 87158039-87158039 on Build GRCh38
rs918668399 Chr.9: 87158091-87158091 on Build GRCh38
rs1025237705 Chr.9: 87158304-87158304 on Build GRCh38
rs901483242 Chr.9: 87158170-87158170 on Build GRCh38
rs138769842 Chr.9: 87158371-87158371 on Build GRCh38
rs958373708 Chr.9: 87158290-87158290 on Build GRCh38
rs573544418 Chr.9: 87158038-87158038 on Build GRCh38
rs970769849 Chr.9: 87158057-87158057 on Build GRCh38
rs909080532 Chr.9: 87158143-87158143 on Build GRCh38
rs989923731 Chr.9: 87158037-87158037 on Build GRCh38
rs377617648 Chr.9: 87158410-87158410 on Build GRCh38
rs936385099 Chr.9: 87158492-87158492 on Build GRCh38
rs998696436 Chr.9: 87158483-87158483 on Build GRCh38
rs112518186 Chr.9: 87158195-87158195 on Build GRCh38
rs771422510 Chr.9: 87158397-87158397 on Build GRCh38
rs943233171 Chr.9: 87158145-87158145 on Build GRCh38
rs986354314 Chr.9: 87158500-87158500 on Build GRCh38
rs558399165 Chr.9: 87158312-87158312 on Build GRCh38
rs895319379 Chr.9: 87158274-87158274 on Build GRCh38
rs144909664 Chr.9: 87158352-87158352 on Build GRCh38
rs568270863 Chr.9: 87158311-87158311 on Build GRCh38
rs1014140061 Chr.9: 87158211-87158211 on Build GRCh38
rs578195856 Chr.9: 87158322-87158322 on Build GRCh38
rs565650174 Chr.9: 87158307-87158307 on Build GRCh38
rs542694653 Chr.9: 87158385-87158385 on Build GRCh38
rs1032840057 Chr.9: 87158279-87158279 on Build GRCh38
rs933270966 Chr.9: 87158431-87158431 on Build GRCh38
rs971417242 Chr.9: 87158306-87158306 on Build GRCh38
rs1053486125 Chr.9: 87158499-87158499 on Build GRCh38
rs182319680 Chr.9: 87158190-87158190 on Build GRCh38
rs62562338 Chr.9: 87158336-87158336 on Build GRCh38
rs906783140 Chr.9: 87158526-87158526 on Build GRCh38
rs73486877 Chr.9: 87158518-87158518 on Build GRCh38
rs528458165 Chr.9: 87158515-87158515 on Build GRCh38
rs1012357333 Chr.9: 87158505-87158505 on Build GRCh38
rs967251123 Chr.9: 87158272-87158272 on Build GRCh38
rs922887178 Chr.9: 87158309-87158309 on Build GRCh38
rs277753 Chr.9: 87158101-87158101 on Build GRCh38
rs902185006 Chr.9: 87158452-87158452 on Build GRCh38
rs1013901455 Chr.9: 87158432-87158432 on Build GRCh38
rs548370888 Chr.9: 87158082-87158082 on Build GRCh38
rs1041287894 Chr.9: 87158064-87158064 on Build GRCh38
rs911224750 Chr.9: 87158297-87158297 on Build GRCh38
rs1018381513 Chr.9: 87158502-87158502 on Build GRCh38
rs186979489 Chr.9: 87158196-87158196 on Build GRCh38
rs534248331 Chr.9: 87158447-87158447 on Build GRCh38
rs1048925475 Chr.9: 87158119-87158119 on Build GRCh38
rs1042470194 Chr.9: 87158181-87158181 on Build GRCh38
rs1024205686 Chr.9: 87158230-87158230 on Build GRCh38
rs538509173 Chr.9: 87158161-87158161 on Build GRCh38
rs894203481 Chr.9: 87158376-87158376 on Build GRCh38

View Full Product Details