Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53988776-53989330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNG8
Entrez Gene ID
59283
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 8
Gene Aliases
-
UniGene
Hs.631595
Gene Chromosome Location
Chr.19: 53963036-53990215 on Build GRCh38
Transcripts
NM_031895.5

SNP Detail

SNP ID Chromosome Location
rs1019520562 Chr.19: 53988863-53988863 on Build GRCh38
rs567031203 Chr.19: 53989127-53989127 on Build GRCh38
rs751731658 Chr.19: 53989071-53989071 on Build GRCh38
rs764869851 Chr.19: 53989297-53989297 on Build GRCh38
rs951588986 Chr.19: 53989155-53989155 on Build GRCh38
rs527456234 Chr.19: 53988898-53988898 on Build GRCh38
rs946352224 Chr.19: 53988978-53988978 on Build GRCh38
rs979049482 Chr.19: 53989274-53989277 on Build GRCh38
rs529949486 Chr.19: 53988865-53988865 on Build GRCh38
rs1008037047 Chr.19: 53989096-53989096 on Build GRCh38
rs12979217 Chr.19: 53989192-53989192 on Build GRCh38
rs886921701 Chr.19: 53989083-53989083 on Build GRCh38
rs1019876217 Chr.19: 53989138-53989138 on Build GRCh38
rs759884425 Chr.19: 53988928-53988928 on Build GRCh38
rs761829886 Chr.19: 53989172-53989172 on Build GRCh38
rs916323292 Chr.19: 53989258-53989258 on Build GRCh38
rs562540687 Chr.19: 53988864-53988864 on Build GRCh38
rs1055424957 Chr.19: 53989022-53989022 on Build GRCh38
rs988168435 Chr.19: 53989206-53989206 on Build GRCh38
rs542068104 Chr.19: 53988867-53988867 on Build GRCh38
rs574742890 Chr.19: 53989267-53989267 on Build GRCh38
rs559389754 Chr.19: 53989087-53989087 on Build GRCh38
rs112369042 Chr.19: 53989276-53989289 on Build GRCh38
rs958146258 Chr.19: 53988893-53988893 on Build GRCh38
rs181172644 Chr.19: 53988871-53988871 on Build GRCh38
rs1020970345 Chr.19: 53989207-53989207 on Build GRCh38
rs761213111 Chr.19: 53989160-53989160 on Build GRCh38
rs555213650 Chr.19: 53989106-53989106 on Build GRCh38
rs112349253 Chr.19: 53988860-53988860 on Build GRCh38
rs556148545 Chr.19: 53989245-53989245 on Build GRCh38
rs1046874433 Chr.19: 53989077-53989077 on Build GRCh38
rs995496773 Chr.19: 53989109-53989109 on Build GRCh38
rs988219329 Chr.19: 53988902-53988902 on Build GRCh38
rs141526501 Chr.19: 53989173-53989173 on Build GRCh38
rs112793050 Chr.19: 53988861-53988861 on Build GRCh38
rs899710989 Chr.19: 53989149-53989149 on Build GRCh38
rs1043659889 Chr.19: 53988993-53988993 on Build GRCh38
rs184193581 Chr.19: 53989283-53989283 on Build GRCh38
rs923889825 Chr.19: 53989298-53989303 on Build GRCh38
rs1046565662 Chr.19: 53988939-53988939 on Build GRCh38
rs570597403 Chr.19: 53988933-53988933 on Build GRCh38
rs925280796 Chr.19: 53988881-53988881 on Build GRCh38
rs571715197 Chr.19: 53989300-53989300 on Build GRCh38
rs1050936302 Chr.19: 53989078-53989078 on Build GRCh38
rs536557888 Chr.19: 53989062-53989062 on Build GRCh38
rs916266822 Chr.19: 53988929-53988929 on Build GRCh38
rs534438089 Chr.19: 53989137-53989137 on Build GRCh38
rs535156515 Chr.19: 53989140-53989140 on Build GRCh38
rs531427059 Chr.19: 53988951-53988951 on Build GRCh38
rs935190390 Chr.19: 53989037-53989037 on Build GRCh38
rs914060157 Chr.19: 53988917-53988917 on Build GRCh38
rs552380872 Chr.19: 53988927-53988927 on Build GRCh38
rs776151907 Chr.19: 53988973-53988973 on Build GRCh38
rs549804206 Chr.19: 53988997-53988997 on Build GRCh38
rs1018738121 Chr.19: 53989097-53989097 on Build GRCh38
rs982210057 Chr.19: 53989261-53989261 on Build GRCh38
rs151332769 Chr.19: 53989046-53989046 on Build GRCh38
rs767924869 Chr.19: 53988961-53988961 on Build GRCh38

View Full Product Details