Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 104353654-104354156 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP58
Entrez Gene ID
159686
Gene Name
cilia and flagella associated protein 58
Gene Aliases
C10orf80, CCDC147, bA127L20.4, bA127L20.5, bA554P13.1
UniGene
Hs.253576
Gene Chromosome Location
Chr.10: 104339124-104455090 on Build GRCh38
Transcripts
NM_001008723.1 XM_011539377.2 XM_017015787.1 XM_017015788.1 XM_017015789.1 XM_017015790.1

SNP Detail

SNP ID Chromosome Location
rs767881945 Chr.10: 104353872-104353872 on Build GRCh38
rs763907154 Chr.10: 104353945-104353945 on Build GRCh38
rs1002612293 Chr.10: 104354098-104354098 on Build GRCh38
rs774846637 Chr.10: 104353929-104353929 on Build GRCh38
rs376512013 Chr.10: 104353967-104353967 on Build GRCh38
rs79858905 Chr.10: 104353720-104353720 on Build GRCh38
rs917157105 Chr.10: 104353764-104353764 on Build GRCh38
rs1015059187 Chr.10: 104353792-104353792 on Build GRCh38
rs527844094 Chr.10: 104353680-104353680 on Build GRCh38
rs762994676 Chr.10: 104353933-104353933 on Build GRCh38
rs922082824 Chr.10: 104353992-104353992 on Build GRCh38
rs932162600 Chr.10: 104354010-104354010 on Build GRCh38
rs978128662 Chr.10: 104353863-104353863 on Build GRCh38
rs1030400429 Chr.10: 104353703-104353703 on Build GRCh38
rs80148976 Chr.10: 104354021-104354021 on Build GRCh38
rs900713564 Chr.10: 104353748-104353748 on Build GRCh38
rs750626097 Chr.10: 104353873-104353873 on Build GRCh38
rs772975648 Chr.10: 104354049-104354049 on Build GRCh38
rs113713062 Chr.10: 104353742-104353742 on Build GRCh38
rs185574994 Chr.10: 104354070-104354070 on Build GRCh38
rs190286212 Chr.10: 104354129-104354129 on Build GRCh38
rs532036322 Chr.10: 104353749-104353749 on Build GRCh38
rs757592921 Chr.10: 104353870-104353870 on Build GRCh38
rs745453821 Chr.10: 104353920-104353920 on Build GRCh38
rs753485122 Chr.10: 104353727-104353727 on Build GRCh38
rs761724145 Chr.10: 104353952-104353952 on Build GRCh38
rs568845513 Chr.10: 104353784-104353784 on Build GRCh38
rs200365700 Chr.10: 104353885-104353885 on Build GRCh38
rs1021513379 Chr.10: 104353837-104353837 on Build GRCh38
rs906897266 Chr.10: 104354094-104354094 on Build GRCh38
rs200898309 Chr.10: 104353898-104353898 on Build GRCh38
rs774389604 Chr.10: 104353946-104353946 on Build GRCh38
rs944838498 Chr.10: 104353740-104353740 on Build GRCh38
rs921465109 Chr.10: 104353973-104353973 on Build GRCh38
rs990058621 Chr.10: 104353704-104353704 on Build GRCh38
rs772389016 Chr.10: 104353913-104353913 on Build GRCh38
rs999983270 Chr.10: 104354069-104354069 on Build GRCh38
rs560802605 Chr.10: 104354118-104354118 on Build GRCh38
rs556160360 Chr.10: 104354109-104354109 on Build GRCh38
rs182377907 Chr.10: 104353821-104353821 on Build GRCh38
rs761555496 Chr.10: 104353812-104353812 on Build GRCh38
rs373922648 Chr.10: 104353869-104353869 on Build GRCh38
rs1003074558 Chr.10: 104353796-104353796 on Build GRCh38
rs1009884284 Chr.10: 104354038-104354038 on Build GRCh38
rs946740706 Chr.10: 104353721-104353721 on Build GRCh38
rs758250894 Chr.10: 104354089-104354089 on Build GRCh38
rs971614982 Chr.10: 104353686-104353686 on Build GRCh38
rs1034640828 Chr.10: 104353691-104353691 on Build GRCh38
rs967325193 Chr.10: 104354123-104354123 on Build GRCh38
rs978574975 Chr.10: 104353733-104353733 on Build GRCh38
rs771764245 Chr.10: 104353723-104353723 on Build GRCh38
rs763566721 Chr.10: 104353697-104353697 on Build GRCh38
rs983200539 Chr.10: 104353809-104353809 on Build GRCh38
rs750242346 Chr.10: 104354031-104354031 on Build GRCh38
rs887424193 Chr.10: 104353772-104353772 on Build GRCh38
rs531439529 Chr.10: 104353817-104353817 on Build GRCh38
rs1004571795 Chr.10: 104353787-104353787 on Build GRCh38
rs906953545 Chr.10: 104353795-104353795 on Build GRCh38
rs376272261 Chr.10: 104353888-104353888 on Build GRCh38
rs932215022 Chr.10: 104353752-104353752 on Build GRCh38
rs552169961 Chr.10: 104353760-104353760 on Build GRCh38
rs199569018 Chr.10: 104353931-104353931 on Build GRCh38
rs892717254 Chr.10: 104354036-104354036 on Build GRCh38
rs749874622 Chr.10: 104353881-104353881 on Build GRCh38
rs976609299 Chr.10: 104353956-104353956 on Build GRCh38
rs530493400 Chr.10: 104354106-104354106 on Build GRCh38
rs756232343 Chr.10: 104353875-104353875 on Build GRCh38
rs967990545 Chr.10: 104353840-104353840 on Build GRCh38
rs776275853 Chr.10: 104353914-104353914 on Build GRCh38
rs764726960 Chr.10: 104353829-104353829 on Build GRCh38
rs1034137542 Chr.10: 104353807-104353807 on Build GRCh38
rs981305883 Chr.10: 104353690-104353690 on Build GRCh38
rs879092527 Chr.10: 104354058-104354058 on Build GRCh38
rs934232385 Chr.10: 104354002-104354002 on Build GRCh38
rs1011375575 Chr.10: 104354116-104354116 on Build GRCh38
rs769410990 Chr.10: 104353928-104353928 on Build GRCh38
rs779933997 Chr.10: 104353879-104353879 on Build GRCh38
rs904262394 Chr.10: 104354061-104354061 on Build GRCh38
rs924060647 Chr.10: 104353728-104353728 on Build GRCh38

View Full Product Details