Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 169654056-169654549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC173
Entrez Gene ID
129881
Gene Name
coiled-coil domain containing 173
Gene Aliases
C2orf77
UniGene
Hs.370111
Gene Chromosome Location
Chr.2: 169645425-169694421 on Build GRCh38
Transcripts
NM_001085447.1 XM_011510589.1 XM_011510590.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4512456 c.903C>G p.I301M Chr.2: 169654131-169654131 on Build GRCh38
3569838 c.874G>A p.E292K Chr.2: 169654160-169654160 on Build GRCh38
279383 c.888G>T p.K296N Chr.2: 169654146-169654146 on Build GRCh38
6010376 c.877G>A p.E293K Chr.2: 169654157-169654157 on Build GRCh38
5717054 c.873G>A p.E291E Chr.2: 169654161-169654161 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747958128 Chr.2: 169654223-169654223 on Build GRCh38
rs781065621 Chr.2: 169654200-169654201 on Build GRCh38
rs760741689 Chr.2: 169654182-169654182 on Build GRCh38
rs929793599 Chr.2: 169654441-169654441 on Build GRCh38
rs910907883 Chr.2: 169654370-169654370 on Build GRCh38
rs929366920 Chr.2: 169654233-169654233 on Build GRCh38
rs769388764 Chr.2: 169654462-169654462 on Build GRCh38
rs187139720 Chr.2: 169654522-169654522 on Build GRCh38
rs976258278 Chr.2: 169654296-169654296 on Build GRCh38
rs535669579 Chr.2: 169654379-169654379 on Build GRCh38
rs955061215 Chr.2: 169654454-169654454 on Build GRCh38
rs530807316 Chr.2: 169654118-169654118 on Build GRCh38
rs368282667 Chr.2: 169654427-169654427 on Build GRCh38
rs555498127 Chr.2: 169654490-169654490 on Build GRCh38
rs780776660 Chr.2: 169654227-169654227 on Build GRCh38
rs966551743 Chr.2: 169654406-169654406 on Build GRCh38
rs1022109250 Chr.2: 169654378-169654378 on Build GRCh38
rs977796719 Chr.2: 169654409-169654409 on Build GRCh38
rs895085192 Chr.2: 169654356-169654356 on Build GRCh38
rs973641070 Chr.2: 169654521-169654521 on Build GRCh38
rs767701013 Chr.2: 169654176-169654176 on Build GRCh38
rs953658345 Chr.2: 169654343-169654343 on Build GRCh38
rs748129044 Chr.2: 169654126-169654126 on Build GRCh38
rs910787182 Chr.2: 169654469-169654469 on Build GRCh38
rs776349478 Chr.2: 169654450-169654451 on Build GRCh38
rs146339584 Chr.2: 169654089-169654089 on Build GRCh38
rs183570726 Chr.2: 169654117-169654117 on Build GRCh38
rs115940388 Chr.2: 169654245-169654245 on Build GRCh38
rs985570843 Chr.2: 169654464-169654464 on Build GRCh38
rs754244427 Chr.2: 169654188-169654188 on Build GRCh38
rs765478715 Chr.2: 169654200-169654200 on Build GRCh38
rs943652362 Chr.2: 169654512-169654512 on Build GRCh38
rs956859729 Chr.2: 169654277-169654277 on Build GRCh38
rs764242607 Chr.2: 169654183-169654183 on Build GRCh38
rs1023667714 Chr.2: 169654235-169654235 on Build GRCh38
rs746662853 Chr.2: 169654221-169654221 on Build GRCh38
rs200463219 Chr.2: 169654517-169654521 on Build GRCh38
rs1003986486 Chr.2: 169654292-169654292 on Build GRCh38
rs754479830 Chr.2: 169654225-169654225 on Build GRCh38
rs775749803 Chr.2: 169654181-169654181 on Build GRCh38
rs1012180948 Chr.2: 169654524-169654527 on Build GRCh38
rs769519252 Chr.2: 169654242-169654243 on Build GRCh38
rs777618032 Chr.2: 169654254-169654254 on Build GRCh38
rs769735414 Chr.2: 169654251-169654251 on Build GRCh38
rs779847324 Chr.2: 169654217-169654217 on Build GRCh38
rs16857114 Chr.2: 169654222-169654222 on Build GRCh38
rs868672729 Chr.2: 169654205-169654205 on Build GRCh38
rs61741435 Chr.2: 169654141-169654141 on Build GRCh38
rs1033597029 Chr.2: 169654329-169654329 on Build GRCh38
rs1029833657 Chr.2: 169654431-169654431 on Build GRCh38
rs371725102 Chr.2: 169654211-169654211 on Build GRCh38
rs747592486 Chr.2: 169654430-169654430 on Build GRCh38
rs367772135 Chr.2: 169654174-169654174 on Build GRCh38
rs762774687 Chr.2: 169654175-169654175 on Build GRCh38
rs1010729316 Chr.2: 169654360-169654360 on Build GRCh38
rs754877345 Chr.2: 169654192-169654193 on Build GRCh38
rs757806315 Chr.2: 169654198-169654198 on Build GRCh38
rs750770589 Chr.2: 169654202-169654202 on Build GRCh38

View Full Product Details