Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 112922773-112923274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD200R1
Entrez Gene ID
131450
Gene Name
CD200 receptor 1
Gene Aliases
CD200R, HCRTR2, MOX2R, OX2R
UniGene
Hs.309158
Gene Chromosome Location
Chr.3: 112922685-112975090 on Build GRCh38
Transcripts
NM_138806.3 NM_170780.2

SNP Detail

SNP ID Chromosome Location
rs559650451 Chr.3: 112923017-112923017 on Build GRCh38
rs904510821 Chr.3: 112922981-112922981 on Build GRCh38
rs1052227241 Chr.3: 112922857-112922857 on Build GRCh38
rs376958365 Chr.3: 112923177-112923177 on Build GRCh38
rs753178353 Chr.3: 112923211-112923211 on Build GRCh38
rs550223339 Chr.3: 112923233-112923233 on Build GRCh38
rs962544067 Chr.3: 112923176-112923176 on Build GRCh38
rs937865586 Chr.3: 112922989-112922989 on Build GRCh38
rs1038453378 Chr.3: 112923150-112923152 on Build GRCh38
rs568109848 Chr.3: 112923180-112923180 on Build GRCh38
rs887132601 Chr.3: 112923048-112923048 on Build GRCh38
rs1043558972 Chr.3: 112922958-112922958 on Build GRCh38
rs929230709 Chr.3: 112923044-112923044 on Build GRCh38
rs979203757 Chr.3: 112922965-112922965 on Build GRCh38
rs973161671 Chr.3: 112923207-112923207 on Build GRCh38
rs1021057994 Chr.3: 112922924-112922924 on Build GRCh38
rs1009036748 Chr.3: 112923066-112923066 on Build GRCh38
rs528309783 Chr.3: 112923068-112923068 on Build GRCh38
rs926351084 Chr.3: 112922985-112922985 on Build GRCh38
rs941358288 Chr.3: 112923098-112923098 on Build GRCh38
rs955817097 Chr.3: 112922900-112922900 on Build GRCh38
rs996963042 Chr.3: 112922837-112922837 on Build GRCh38
rs891850883 Chr.3: 112922926-112922926 on Build GRCh38
rs35099230 Chr.3: 112923228-112923228 on Build GRCh38
rs939148627 Chr.3: 112922815-112922815 on Build GRCh38
rs996614768 Chr.3: 112922990-112922990 on Build GRCh38
rs573138021 Chr.3: 112923022-112923022 on Build GRCh38
rs188311290 Chr.3: 112923117-112923117 on Build GRCh38
rs1016958317 Chr.3: 112922798-112922798 on Build GRCh38
rs557777000 Chr.3: 112922845-112922845 on Build GRCh38
rs1029908452 Chr.3: 112922859-112922859 on Build GRCh38
rs954946437 Chr.3: 112923047-112923047 on Build GRCh38
rs988381220 Chr.3: 112922904-112922905 on Build GRCh38
rs1010340158 Chr.3: 112922942-112922942 on Build GRCh38

View Full Product Details