Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151700982-151701492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF3
Entrez Gene ID
11189
Gene Name
CUGBP, Elav-like family member 3
Gene Aliases
BRUNOL1, CAGH4, ERDA4, ETR-1, TNRC4
UniGene
Hs.26047
Gene Chromosome Location
Chr.1: 151700058-151716821 on Build GRCh38
Transcripts
NM_001172648.3 NM_001172649.3 NM_001291106.1 NM_001291107.1 NM_007185.6 XM_006711139.2 XM_006711141.2 XM_011509111.2 XM_011509112.2 XM_011509114.2 XM_017000145.1 XM_017000146.1

SNP Detail

SNP ID Chromosome Location
rs921888473 Chr.1: 151701438-151701438 on Build GRCh38
rs1027034683 Chr.1: 151701250-151701250 on Build GRCh38
rs543716309 Chr.1: 151701430-151701430 on Build GRCh38
rs1030539564 Chr.1: 151701281-151701282 on Build GRCh38
rs996823226 Chr.1: 151701275-151701275 on Build GRCh38
rs771577391 Chr.1: 151701099-151701099 on Build GRCh38
rs532045283 Chr.1: 151701330-151701330 on Build GRCh38
rs189207685 Chr.1: 151701342-151701342 on Build GRCh38
rs528072239 Chr.1: 151701159-151701159 on Build GRCh38
rs780003994 Chr.1: 151701128-151701128 on Build GRCh38
rs534086768 Chr.1: 151701415-151701415 on Build GRCh38
rs181162482 Chr.1: 151701367-151701367 on Build GRCh38
rs937974181 Chr.1: 151701424-151701424 on Build GRCh38
rs563072694 Chr.1: 151701225-151701225 on Build GRCh38
rs78636920 Chr.1: 151701137-151701137 on Build GRCh38
rs571510048 Chr.1: 151701413-151701413 on Build GRCh38
rs540742656 Chr.1: 151701049-151701049 on Build GRCh38
rs954843493 Chr.1: 151701283-151701283 on Build GRCh38
rs968768780 Chr.1: 151701340-151701340 on Build GRCh38
rs982016228 Chr.1: 151701047-151701069 on Build GRCh38
rs560902474 Chr.1: 151701119-151701119 on Build GRCh38
rs182085212 Chr.1: 151701016-151701016 on Build GRCh38
rs369797253 Chr.1: 151701052-151701052 on Build GRCh38
rs549335571 Chr.1: 151701148-151701148 on Build GRCh38
rs964999510 Chr.1: 151701274-151701274 on Build GRCh38
rs572224833 Chr.1: 151701042-151701042 on Build GRCh38
rs913333911 Chr.1: 151701334-151701334 on Build GRCh38
rs749211135 Chr.1: 151701350-151701350 on Build GRCh38
rs186011019 Chr.1: 151701143-151701143 on Build GRCh38
rs765459472 Chr.1: 151701351-151701351 on Build GRCh38
rs991140177 Chr.1: 151701436-151701436 on Build GRCh38
rs988925961 Chr.1: 151701321-151701321 on Build GRCh38
rs1004692675 Chr.1: 151701353-151701353 on Build GRCh38

View Full Product Details