Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 90405366-90405822 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN12
Entrez Gene ID
9069
Gene Name
claudin 12
Gene Aliases
-
UniGene
Hs.731917
Gene Chromosome Location
Chr.7: 90403334-90415954 on Build GRCh38
Transcripts
NM_001185072.2 NM_001185073.2 NM_012129.4 XM_017012892.1 XM_017012893.1

Gene Symbol
LOC102723899
Entrez Gene ID
102723899
Gene Name
pituitary tumor-transforming gene 1 protein-interacting protein
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.7: 90403379-90513397 on Build GRCh38
Transcripts
NM_001185072.2 NM_001185073.2 NM_012129.4 XM_017012892.1 XM_017012893.1

SNP Detail

SNP ID Chromosome Location
rs528043700 Chr.7: 90405773-90405773 on Build GRCh38
rs905259000 Chr.7: 90405629-90405629 on Build GRCh38
rs910285876 Chr.7: 90405554-90405554 on Build GRCh38
rs899296920 Chr.7: 90405736-90405736 on Build GRCh38
rs780226842 Chr.7: 90405413-90405413 on Build GRCh38
rs972278194 Chr.7: 90405533-90405533 on Build GRCh38
rs949381040 Chr.7: 90405619-90405619 on Build GRCh38
rs911440597 Chr.7: 90405577-90405577 on Build GRCh38
rs895027578 Chr.7: 90405734-90405734 on Build GRCh38
rs755004916 Chr.7: 90405578-90405578 on Build GRCh38
rs1002773415 Chr.7: 90405474-90405474 on Build GRCh38
rs773441268 Chr.7: 90405712-90405712 on Build GRCh38
rs116804658 Chr.7: 90405535-90405535 on Build GRCh38
rs907210025 Chr.7: 90405705-90405705 on Build GRCh38
rs1035350388 Chr.7: 90405730-90405730 on Build GRCh38
rs548104500 Chr.7: 90405624-90405624 on Build GRCh38
rs773152966 Chr.7: 90405538-90405539 on Build GRCh38
rs559654252 Chr.7: 90405492-90405492 on Build GRCh38
rs564506207 Chr.7: 90405721-90405721 on Build GRCh38
rs987257325 Chr.7: 90405553-90405553 on Build GRCh38
rs553754996 Chr.7: 90405517-90405517 on Build GRCh38
rs1046507142 Chr.7: 90405627-90405627 on Build GRCh38
rs925780228 Chr.7: 90405600-90405600 on Build GRCh38
rs539220571 Chr.7: 90405617-90405617 on Build GRCh38
rs1002267391 Chr.7: 90405655-90405655 on Build GRCh38
rs1052929005 Chr.7: 90405613-90405613 on Build GRCh38
rs558237432 Chr.7: 90405675-90405676 on Build GRCh38
rs963982658 Chr.7: 90405524-90405524 on Build GRCh38
rs1026564265 Chr.7: 90405776-90405776 on Build GRCh38
rs185475099 Chr.7: 90405603-90405603 on Build GRCh38
rs548604300 Chr.7: 90405701-90405701 on Build GRCh38
rs781426929 Chr.7: 90405598-90405598 on Build GRCh38
rs760822911 Chr.7: 90405789-90405789 on Build GRCh38
rs770200022 Chr.7: 90405690-90405690 on Build GRCh38
rs1016542040 Chr.7: 90405506-90405506 on Build GRCh38
rs954322006 Chr.7: 90405539-90405539 on Build GRCh38
rs905395587 Chr.7: 90405403-90405403 on Build GRCh38
rs6963257 Chr.7: 90405796-90405796 on Build GRCh38
rs934587691 Chr.7: 90405608-90405608 on Build GRCh38
rs7792018 Chr.7: 90405395-90405395 on Build GRCh38
rs1025478980 Chr.7: 90405402-90405402 on Build GRCh38
rs962426667 Chr.7: 90405400-90405400 on Build GRCh38
rs551241265 Chr.7: 90405582-90405582 on Build GRCh38

View Full Product Details